Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41746535-41747037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR6814
Entrez Gene ID
102465488
Gene Name
microRNA 6814
Gene Aliases
hsa-mir-6814
UniGene
Gene Chromosome Location
Chr.21: 41746772-41746841 on Build GRCh38
Transcripts
NR_106872.1 NM_020639.2

Gene Symbol
RIPK4
Entrez Gene ID
54101
Gene Name
receptor interacting serine/threonine kinase 4
Gene Aliases
ANKK2, ANKRD3, DIK, NKRD3, PKK, PPS2, RIP4
UniGene
Hs.517310
Gene Chromosome Location
Chr.21: 41739369-41767089 on Build GRCh38
Transcripts
NR_106872.1 NM_020639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5858653 c.809C>T p.P270L Chr.21: 41746636-41746636 on Build GRCh38
4637037 c.809C>A p.P270Q Chr.21: 41746636-41746636 on Build GRCh38
6478341 c.747A>G p.R249R Chr.21: 41746698-41746698 on Build GRCh38
296862 c.812G>A p.R271Q Chr.21: 41746633-41746633 on Build GRCh38
7331076 c.811C>T p.R271* Chr.21: 41746634-41746634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541790408 Chr.21: 41746882-41746882 on Build GRCh38
rs377254301 Chr.21: 41746686-41746686 on Build GRCh38
rs780110086 Chr.21: 41746857-41746857 on Build GRCh38
rs762385012 Chr.21: 41746718-41746718 on Build GRCh38
rs144981787 Chr.21: 41746723-41746724 on Build GRCh38
rs992479936 Chr.21: 41746935-41746935 on Build GRCh38
rs905851485 Chr.21: 41746899-41746899 on Build GRCh38
rs773372178 Chr.21: 41746729-41746730 on Build GRCh38
rs891831110 Chr.21: 41746723-41746723 on Build GRCh38
rs537366804 Chr.21: 41746842-41746842 on Build GRCh38
rs574762421 Chr.21: 41746903-41746903 on Build GRCh38
rs1014622365 Chr.21: 41746653-41746653 on Build GRCh38
rs772122950 Chr.21: 41746632-41746632 on Build GRCh38
rs768512721 Chr.21: 41746622-41746622 on Build GRCh38
rs965701686 Chr.21: 41746665-41746665 on Build GRCh38
rs200917164 Chr.21: 41746774-41746774 on Build GRCh38
rs188327126 Chr.21: 41746911-41746911 on Build GRCh38
rs146341788 Chr.21: 41746611-41746611 on Build GRCh38
rs758807765 Chr.21: 41746647-41746647 on Build GRCh38
rs539297855 Chr.21: 41746869-41746869 on Build GRCh38
rs376489105 Chr.21: 41746591-41746591 on Build GRCh38
rs143840048 Chr.21: 41746854-41746854 on Build GRCh38
rs940170033 Chr.21: 41747006-41747006 on Build GRCh38
rs764473830 Chr.21: 41746654-41746654 on Build GRCh38
rs768045983 Chr.21: 41746828-41746828 on Build GRCh38
rs770860869 Chr.21: 41746690-41746690 on Build GRCh38
rs753148439 Chr.21: 41746605-41746605 on Build GRCh38
rs754584153 Chr.21: 41746733-41746733 on Build GRCh38
rs1053077537 Chr.21: 41746575-41746575 on Build GRCh38
rs753385569 Chr.21: 41746728-41746728 on Build GRCh38
rs548574974 Chr.21: 41747009-41747009 on Build GRCh38
rs73906705 Chr.21: 41746895-41746895 on Build GRCh38
rs764775567 Chr.21: 41746878-41746878 on Build GRCh38
rs138515359 Chr.21: 41746875-41746875 on Build GRCh38
rs924396478 Chr.21: 41746828-41746828 on Build GRCh38
rs145556669 Chr.21: 41746688-41746688 on Build GRCh38
rs932839266 Chr.21: 41746682-41746682 on Build GRCh38
rs769995090 Chr.21: 41746707-41746707 on Build GRCh38
rs767637884 Chr.21: 41746590-41746590 on Build GRCh38
rs926108742 Chr.21: 41746693-41746693 on Build GRCh38
rs552648714 Chr.21: 41746687-41746687 on Build GRCh38
rs953070177 Chr.21: 41746986-41746986 on Build GRCh38
rs765614531 Chr.21: 41746602-41746602 on Build GRCh38
rs777639039 Chr.21: 41746775-41746775 on Build GRCh38
rs767713104 Chr.21: 41746576-41746576 on Build GRCh38
rs559048535 Chr.21: 41746620-41746620 on Build GRCh38
rs775637459 Chr.21: 41746799-41746799 on Build GRCh38
rs766489200 Chr.21: 41746598-41746598 on Build GRCh38
rs375435761 Chr.21: 41746599-41746599 on Build GRCh38
rs139607246 Chr.21: 41746655-41746655 on Build GRCh38
rs759077398 Chr.21: 41746837-41746837 on Build GRCh38
rs528593289 Chr.21: 41746974-41746974 on Build GRCh38
rs149032082 Chr.21: 41746640-41746640 on Build GRCh38
rs764538906 Chr.21: 41746711-41746711 on Build GRCh38
rs572876681 Chr.21: 41746879-41746879 on Build GRCh38
rs989029560 Chr.21: 41746885-41746885 on Build GRCh38
rs977125293 Chr.21: 41746810-41746810 on Build GRCh38
rs901297422 Chr.21: 41746874-41746874 on Build GRCh38
rs779530950 Chr.21: 41746612-41746612 on Build GRCh38
rs543708665 Chr.21: 41746905-41746905 on Build GRCh38
rs534995730 Chr.21: 41746725-41746725 on Build GRCh38
rs937446152 Chr.21: 41746904-41746904 on Build GRCh38
rs868493837 Chr.21: 41746948-41746948 on Build GRCh38
rs144179743 Chr.21: 41746633-41746633 on Build GRCh38
rs776699593 Chr.21: 41746694-41746694 on Build GRCh38
rs775725385 Chr.21: 41746709-41746709 on Build GRCh38
rs528547745 Chr.21: 41746953-41746953 on Build GRCh38
rs774484584 Chr.21: 41746868-41746868 on Build GRCh38
rs921644618 Chr.21: 41746668-41746668 on Build GRCh38
rs764863379 Chr.21: 41746838-41746838 on Build GRCh38
rs563560488 Chr.21: 41746685-41746685 on Build GRCh38
rs759682809 Chr.21: 41746706-41746706 on Build GRCh38
rs775801568 Chr.21: 41746674-41746674 on Build GRCh38
rs552541664 Chr.21: 41746926-41746926 on Build GRCh38
rs878880423 Chr.21: 41746650-41746650 on Build GRCh38
rs770900342 Chr.21: 41746788-41746788 on Build GRCh38
rs949002707 Chr.21: 41746851-41746851 on Build GRCh38
rs778678584 Chr.21: 41746737-41746737 on Build GRCh38
rs538806393 Chr.21: 41746989-41746989 on Build GRCh38
rs761853253 Chr.21: 41746589-41746589 on Build GRCh38
rs976783887 Chr.21: 41746719-41746719 on Build GRCh38
rs549925444 Chr.21: 41746666-41746666 on Build GRCh38
rs768908446 Chr.21: 41746866-41746866 on Build GRCh38
rs185472832 Chr.21: 41746830-41746830 on Build GRCh38
rs760867599 Chr.21: 41746595-41746595 on Build GRCh38
rs530359106 Chr.21: 41746641-41746641 on Build GRCh38
rs751359756 Chr.21: 41746845-41746845 on Build GRCh38
rs13048502 Chr.21: 41746906-41746906 on Build GRCh38
rs774291469 Chr.21: 41746624-41746624 on Build GRCh38
rs753060606 Chr.21: 41746642-41746642 on Build GRCh38
rs915581413 Chr.21: 41746952-41746952 on Build GRCh38
rs769889642 Chr.21: 41746790-41746790 on Build GRCh38
rs768835121 Chr.21: 41746803-41746803 on Build GRCh38
rs566081548 Chr.21: 41746713-41746713 on Build GRCh38
rs762140954 Chr.21: 41746821-41746821 on Build GRCh38
rs914292273 Chr.21: 41746635-41746635 on Build GRCh38
rs746991099 Chr.21: 41746689-41746689 on Build GRCh38
rs757054568 Chr.21: 41746852-41746852 on Build GRCh38
rs149749209 Chr.21: 41746667-41746667 on Build GRCh38
rs751198551 Chr.21: 41746890-41746890 on Build GRCh38
rs563651173 Chr.21: 41746683-41746683 on Build GRCh38
rs139503982 Chr.21: 41746626-41746626 on Build GRCh38
rs1014854989 Chr.21: 41746958-41746958 on Build GRCh38
rs745345054 Chr.21: 41746608-41746608 on Build GRCh38
rs755297456 Chr.21: 41746601-41746601 on Build GRCh38
rs752429815 Chr.21: 41746753-41746753 on Build GRCh38
rs760641174 Chr.21: 41746636-41746636 on Build GRCh38
rs982749754 Chr.21: 41746558-41746558 on Build GRCh38
rs762432751 Chr.21: 41746880-41746880 on Build GRCh38
rs763261232 Chr.21: 41746710-41746710 on Build GRCh38
rs147290898 Chr.21: 41746959-41746959 on Build GRCh38
rs370707445 Chr.21: 41747002-41747002 on Build GRCh38
rs761278858 Chr.21: 41746877-41746877 on Build GRCh38
rs749394094 Chr.21: 41746865-41746865 on Build GRCh38
rs116839817 Chr.21: 41746855-41746855 on Build GRCh38
rs774772567 Chr.21: 41746808-41746808 on Build GRCh38
rs757024529 Chr.21: 41746891-41746891 on Build GRCh38
rs1046115040 Chr.21: 41746896-41746896 on Build GRCh38
rs772597586 Chr.21: 41746870-41746870 on Build GRCh38
rs778398173 Chr.21: 41746606-41746606 on Build GRCh38
rs774381110 Chr.21: 41746562-41746562 on Build GRCh38
rs547105895 Chr.21: 41746954-41746954 on Build GRCh38
rs780865560 Chr.21: 41746965-41746965 on Build GRCh38
rs781168270 Chr.21: 41746789-41746789 on Build GRCh38
rs746858488 Chr.21: 41746781-41746781 on Build GRCh38
rs758078007 Chr.21: 41746843-41746843 on Build GRCh38

View Full Product Details