Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 27982753-27983263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC28
Entrez Gene ID
23331
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
TPRBK
UniGene
Hs.387856
Gene Chromosome Location
Chr.22: 27978014-28679865 on Build GRCh38
Transcripts
NM_001145418.1 XM_005261405.2 XM_006724171.3 XM_011530018.2 XM_017028673.1 NR_026963.1

Gene Symbol
TTC28-AS1
Entrez Gene ID
284900
Gene Name
TTC28 antisense RNA 1
Gene Aliases
TTC28-AS, TTC28AS, dJ353E16.2
UniGene
Hs.602319
Gene Chromosome Location
Chr.22: 27919376-28002679 on Build GRCh38
Transcripts
NM_001145418.1 XM_005261405.2 XM_006724171.3 XM_011530018.2 XM_017028673.1 NR_026963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6356302 c.6788C>T p.P2263L Chr.22: 27982879-27982879 on Build GRCh38
1033039 c.6822C>T p.C2274C Chr.22: 27982845-27982845 on Build GRCh38
1616322 c.6903A>G p.K2301K Chr.22: 27982764-27982764 on Build GRCh38
5344076 c.6589G>A p.V2197I Chr.22: 27983078-27983078 on Build GRCh38
1033040 c.6579T>C p.P2193P Chr.22: 27983088-27983088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931963418 Chr.22: 27982917-27982917 on Build GRCh38
rs1040126300 Chr.22: 27982951-27982951 on Build GRCh38
rs751128994 Chr.22: 27983007-27983007 on Build GRCh38
rs777717051 Chr.22: 27983049-27983049 on Build GRCh38
rs763865377 Chr.22: 27982946-27982946 on Build GRCh38
rs896896675 Chr.22: 27982821-27982821 on Build GRCh38
rs372409227 Chr.22: 27983139-27983139 on Build GRCh38
rs760016036 Chr.22: 27982925-27982925 on Build GRCh38
rs1025708079 Chr.22: 27983003-27983003 on Build GRCh38
rs753111680 Chr.22: 27982801-27982801 on Build GRCh38
rs542940557 Chr.22: 27982828-27982828 on Build GRCh38
rs745425991 Chr.22: 27983043-27983043 on Build GRCh38
rs758369722 Chr.22: 27983136-27983136 on Build GRCh38
rs768505455 Chr.22: 27982898-27982898 on Build GRCh38
rs1050667308 Chr.22: 27982780-27982780 on Build GRCh38
rs1005612701 Chr.22: 27983238-27983238 on Build GRCh38
rs774233407 Chr.22: 27983099-27983099 on Build GRCh38
rs760927930 Chr.22: 27982943-27982943 on Build GRCh38
rs753205814 Chr.22: 27982944-27982944 on Build GRCh38
rs144350540 Chr.22: 27982850-27982850 on Build GRCh38
rs750107371 Chr.22: 27983236-27983236 on Build GRCh38
rs758631680 Chr.22: 27982964-27982964 on Build GRCh38
rs753960601 Chr.22: 27982924-27982924 on Build GRCh38
rs373286553 Chr.22: 27982845-27982845 on Build GRCh38
rs374544992 Chr.22: 27982879-27982879 on Build GRCh38
rs189781469 Chr.22: 27983028-27983028 on Build GRCh38
rs370917225 Chr.22: 27982851-27982851 on Build GRCh38
rs184835480 Chr.22: 27982878-27982878 on Build GRCh38
rs17482652 Chr.22: 27983010-27983010 on Build GRCh38
rs551506894 Chr.22: 27982911-27982911 on Build GRCh38
rs747827727 Chr.22: 27983037-27983037 on Build GRCh38
rs538752652 Chr.22: 27983078-27983078 on Build GRCh38
rs773254804 Chr.22: 27982905-27982905 on Build GRCh38
rs755511418 Chr.22: 27982779-27982779 on Build GRCh38
rs771812441 Chr.22: 27983044-27983044 on Build GRCh38
rs745766686 Chr.22: 27982861-27982861 on Build GRCh38
rs901149944 Chr.22: 27982953-27982953 on Build GRCh38
rs764781324 Chr.22: 27983098-27983098 on Build GRCh38
rs970407508 Chr.22: 27983112-27983112 on Build GRCh38
rs572223686 Chr.22: 27982823-27982823 on Build GRCh38
rs967977216 Chr.22: 27983079-27983079 on Build GRCh38
rs764330777 Chr.22: 27983140-27983140 on Build GRCh38
rs553471572 Chr.22: 27983114-27983114 on Build GRCh38
rs979279969 Chr.22: 27982885-27982885 on Build GRCh38
rs984285849 Chr.22: 27982818-27982818 on Build GRCh38
rs549693274 Chr.22: 27983006-27983006 on Build GRCh38
rs909480025 Chr.22: 27983228-27983228 on Build GRCh38
rs960331630 Chr.22: 27982855-27982855 on Build GRCh38
rs576263878 Chr.22: 27983220-27983220 on Build GRCh38
rs887946345 Chr.22: 27982782-27982782 on Build GRCh38
rs923669430 Chr.22: 27982862-27982862 on Build GRCh38
rs780105357 Chr.22: 27983150-27983150 on Build GRCh38
rs951575358 Chr.22: 27982800-27982800 on Build GRCh38
rs962210160 Chr.22: 27982832-27982832 on Build GRCh38
rs769207304 Chr.22: 27982778-27982778 on Build GRCh38
rs767347871 Chr.22: 27983229-27983229 on Build GRCh38
rs371102203 Chr.22: 27982792-27982792 on Build GRCh38
rs778483279 Chr.22: 27983029-27983029 on Build GRCh38
rs747169297 Chr.22: 27983108-27983108 on Build GRCh38
rs776681243 Chr.22: 27983117-27983117 on Build GRCh38
rs143432636 Chr.22: 27983132-27983132 on Build GRCh38
rs935398970 Chr.22: 27982901-27982901 on Build GRCh38
rs915054240 Chr.22: 27983172-27983172 on Build GRCh38
rs1046208266 Chr.22: 27982856-27982856 on Build GRCh38
rs759543750 Chr.22: 27983122-27983122 on Build GRCh38
rs1039650020 Chr.22: 27983202-27983202 on Build GRCh38
rs777222069 Chr.22: 27982836-27982836 on Build GRCh38
rs148825394 Chr.22: 27983115-27983115 on Build GRCh38
rs143483336 Chr.22: 27983082-27983082 on Build GRCh38
rs756843815 Chr.22: 27983190-27983190 on Build GRCh38
rs901079272 Chr.22: 27983207-27983209 on Build GRCh38
rs527964221 Chr.22: 27982948-27982948 on Build GRCh38
rs780074002 Chr.22: 27983071-27983071 on Build GRCh38
rs202068761 Chr.22: 27982822-27982822 on Build GRCh38
rs765313384 Chr.22: 27983131-27983131 on Build GRCh38
rs566886681 Chr.22: 27982926-27982926 on Build GRCh38
rs986137574 Chr.22: 27983184-27983184 on Build GRCh38
rs993932402 Chr.22: 27983030-27983030 on Build GRCh38
rs5762431 Chr.22: 27983070-27983070 on Build GRCh38
rs533331940 Chr.22: 27982881-27982881 on Build GRCh38
rs373188785 Chr.22: 27983118-27983118 on Build GRCh38
rs147787655 Chr.22: 27983052-27983052 on Build GRCh38
rs549696928 Chr.22: 27983187-27983187 on Build GRCh38
rs766406725 Chr.22: 27982914-27982914 on Build GRCh38

View Full Product Details