Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119212387-119212884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT4
Entrez Gene ID
8702
Gene Name
beta-1,4-galactosyltransferase 4
Gene Aliases
B4Gal-T4, beta4Gal-T4
UniGene
Hs.13225
Gene Chromosome Location
Chr.3: 119211742-119240946 on Build GRCh38
Transcripts
NM_003778.3 NM_212543.1 XM_005247855.1 XM_006713798.2 XM_006713799.2 XM_006713800.1 XM_006713801.2 XM_011513260.1 XM_017007401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3660112 c.1006A>G p.I336V Chr.3: 119212578-119212578 on Build GRCh38
1752823 c.933C>T p.V311V Chr.3: 119212651-119212651 on Build GRCh38
6609462 c.914T>C p.L305S Chr.3: 119212670-119212670 on Build GRCh38
1036682 c.928C>T p.R310* Chr.3: 119212656-119212656 on Build GRCh38
6511399 c.1010C>T p.T337I Chr.3: 119212574-119212574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150344141 Chr.3: 119212554-119212554 on Build GRCh38
rs150966320 Chr.3: 119212584-119212584 on Build GRCh38
rs760595977 Chr.3: 119212788-119212788 on Build GRCh38
rs773554968 Chr.3: 119212532-119212532 on Build GRCh38
rs990586756 Chr.3: 119212783-119212783 on Build GRCh38
rs6784208 Chr.3: 119212542-119212542 on Build GRCh38
rs750842521 Chr.3: 119212430-119212430 on Build GRCh38
rs72655940 Chr.3: 119212438-119212438 on Build GRCh38
rs371155191 Chr.3: 119212713-119212713 on Build GRCh38
rs751556702 Chr.3: 119212723-119212723 on Build GRCh38
rs188399911 Chr.3: 119212474-119212474 on Build GRCh38
rs950344842 Chr.3: 119212595-119212595 on Build GRCh38
rs760493860 Chr.3: 119212690-119212690 on Build GRCh38
rs200067522 Chr.3: 119212693-119212693 on Build GRCh38
rs17849860 Chr.3: 119212669-119212669 on Build GRCh38
rs1006884699 Chr.3: 119212715-119212715 on Build GRCh38
rs113574391 Chr.3: 119212484-119212484 on Build GRCh38
rs534561923 Chr.3: 119212854-119212854 on Build GRCh38
rs770427428 Chr.3: 119212540-119212540 on Build GRCh38
rs761097317 Chr.3: 119212537-119212537 on Build GRCh38
rs761462193 Chr.3: 119212725-119212725 on Build GRCh38
rs747321523 Chr.3: 119212683-119212683 on Build GRCh38
rs774668771 Chr.3: 119212517-119212517 on Build GRCh38
rs773364309 Chr.3: 119212655-119212655 on Build GRCh38
rs1053112886 Chr.3: 119212502-119212502 on Build GRCh38
rs748681703 Chr.3: 119212519-119212519 on Build GRCh38
rs780329693 Chr.3: 119212460-119212460 on Build GRCh38
rs776513526 Chr.3: 119212711-119212711 on Build GRCh38
rs772566300 Chr.3: 119212656-119212656 on Build GRCh38
rs72655938 Chr.3: 119212639-119212639 on Build GRCh38
rs772093606 Chr.3: 119212580-119212580 on Build GRCh38
rs559820184 Chr.3: 119212849-119212849 on Build GRCh38
rs778930636 Chr.3: 119212641-119212641 on Build GRCh38
rs537174060 Chr.3: 119212767-119212767 on Build GRCh38
rs757080098 Chr.3: 119212591-119212591 on Build GRCh38
rs768850722 Chr.3: 119212548-119212548 on Build GRCh38
rs6784717 Chr.3: 119212822-119212822 on Build GRCh38
rs937020695 Chr.3: 119212552-119212552 on Build GRCh38
rs1018782481 Chr.3: 119212717-119212717 on Build GRCh38
rs764055829 Chr.3: 119212714-119212714 on Build GRCh38
rs775125026 Chr.3: 119212710-119212710 on Build GRCh38
rs778260430 Chr.3: 119212668-119212668 on Build GRCh38
rs201736032 Chr.3: 119212529-119212529 on Build GRCh38
rs532563287 Chr.3: 119212752-119212752 on Build GRCh38
rs946586103 Chr.3: 119212442-119212444 on Build GRCh38
rs764184506 Chr.3: 119212569-119212569 on Build GRCh38
rs753058429 Chr.3: 119212623-119212625 on Build GRCh38
rs1027948260 Chr.3: 119212592-119212592 on Build GRCh38
rs769383261 Chr.3: 119212701-119212701 on Build GRCh38
rs780078091 Chr.3: 119212593-119212593 on Build GRCh38
rs779176185 Chr.3: 119212549-119212549 on Build GRCh38
rs891168711 Chr.3: 119212704-119212704 on Build GRCh38
rs755739155 Chr.3: 119212723-119212724 on Build GRCh38
rs555333982 Chr.3: 119212810-119212810 on Build GRCh38
rs72655939 Chr.3: 119212556-119212556 on Build GRCh38
rs754108910 Chr.3: 119212615-119212615 on Build GRCh38
rs913487791 Chr.3: 119212467-119212467 on Build GRCh38
rs1043619297 Chr.3: 119212863-119212863 on Build GRCh38
rs34555519 Chr.3: 119212747-119212748 on Build GRCh38
rs72655937 Chr.3: 119212694-119212694 on Build GRCh38
rs767372852 Chr.3: 119212730-119212730 on Build GRCh38
rs767808972 Chr.3: 119212620-119212622 on Build GRCh38

View Full Product Details