Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3444727-3445227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGFAC
Entrez Gene ID
3083
Gene Name
HGF activator
Gene Aliases
HGFA
UniGene
Hs.104
Gene Chromosome Location
Chr.4: 3441933-3449495 on Build GRCh38
Transcripts
NM_001297439.1 NM_001528.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209631 c.980C>T p.A327V Chr.4: 3444957-3444957 on Build GRCh38
3603394 c.936C>T p.S312S Chr.4: 3444913-3444913 on Build GRCh38
4959960 c.866G>A p.G289E Chr.4: 3444843-3444843 on Build GRCh38
5481887 c.975C>T p.G325G Chr.4: 3444952-3444952 on Build GRCh38
1054927 c.882C>A p.Y294* Chr.4: 3444859-3444859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750588653 Chr.4: 3444781-3444781 on Build GRCh38
rs771061781 Chr.4: 3444928-3444928 on Build GRCh38
rs1031834455 Chr.4: 3445092-3445092 on Build GRCh38
rs199924643 Chr.4: 3444937-3444937 on Build GRCh38
rs377539052 Chr.4: 3445007-3445007 on Build GRCh38
rs770592174 Chr.4: 3444752-3444752 on Build GRCh38
rs202090816 Chr.4: 3444958-3444958 on Build GRCh38
rs369980486 Chr.4: 3444989-3444989 on Build GRCh38
rs1023065109 Chr.4: 3445118-3445118 on Build GRCh38
rs371414500 Chr.4: 3444789-3444789 on Build GRCh38
rs3752439 Chr.4: 3445024-3445024 on Build GRCh38
rs374919117 Chr.4: 3444860-3444860 on Build GRCh38
rs748101009 Chr.4: 3444831-3444831 on Build GRCh38
rs545303850 Chr.4: 3444975-3444975 on Build GRCh38
rs746976598 Chr.4: 3444969-3444969 on Build GRCh38
rs1022724157 Chr.4: 3445096-3445096 on Build GRCh38
rs564886600 Chr.4: 3445000-3445000 on Build GRCh38
rs1018743491 Chr.4: 3444754-3444754 on Build GRCh38
rs147268113 Chr.4: 3444964-3444964 on Build GRCh38
rs1052823843 Chr.4: 3445195-3445195 on Build GRCh38
rs371968209 Chr.4: 3445004-3445004 on Build GRCh38
rs758127865 Chr.4: 3444999-3444999 on Build GRCh38
rs3752442 Chr.4: 3445156-3445156 on Build GRCh38
rs140054551 Chr.4: 3444904-3444904 on Build GRCh38
rs754713977 Chr.4: 3444886-3444886 on Build GRCh38
rs901469941 Chr.4: 3445086-3445086 on Build GRCh38
rs768457054 Chr.4: 3444772-3444772 on Build GRCh38
rs34665586 Chr.4: 3444807-3444808 on Build GRCh38
rs567299158 Chr.4: 3445170-3445170 on Build GRCh38
rs752365593 Chr.4: 3444995-3444995 on Build GRCh38
rs768217793 Chr.4: 3444872-3444872 on Build GRCh38
rs771972328 Chr.4: 3444913-3444913 on Build GRCh38
rs758995866 Chr.4: 3445026-3445026 on Build GRCh38
rs780375201 Chr.4: 3445015-3445015 on Build GRCh38
rs3752440 Chr.4: 3445027-3445027 on Build GRCh38
rs372328673 Chr.4: 3444773-3444773 on Build GRCh38
rs747650651 Chr.4: 3444759-3444767 on Build GRCh38
rs772076535 Chr.4: 3444847-3444847 on Build GRCh38
rs767610739 Chr.4: 3444776-3444776 on Build GRCh38
rs528349785 Chr.4: 3445108-3445108 on Build GRCh38
rs776940607 Chr.4: 3444856-3444856 on Build GRCh38
rs763712380 Chr.4: 3445033-3445033 on Build GRCh38
rs775145219 Chr.4: 3444760-3444760 on Build GRCh38
rs753278072 Chr.4: 3444795-3444795 on Build GRCh38
rs780993043 Chr.4: 3444749-3444753 on Build GRCh38
rs758966957 Chr.4: 3444865-3444865 on Build GRCh38
rs761845394 Chr.4: 3444774-3444774 on Build GRCh38
rs551722565 Chr.4: 3445109-3445109 on Build GRCh38
rs754623818 Chr.4: 3444956-3444956 on Build GRCh38
rs536476839 Chr.4: 3444957-3444957 on Build GRCh38
rs759843037 Chr.4: 3444938-3444938 on Build GRCh38
rs1008457507 Chr.4: 3445078-3445078 on Build GRCh38
rs746001572 Chr.4: 3444971-3444971 on Build GRCh38
rs1019208353 Chr.4: 3444901-3444901 on Build GRCh38
rs756210436 Chr.4: 3445101-3445101 on Build GRCh38
rs781328045 Chr.4: 3444970-3444970 on Build GRCh38
rs774227017 Chr.4: 3444830-3444830 on Build GRCh38
rs186940347 Chr.4: 3445146-3445146 on Build GRCh38
rs944235487 Chr.4: 3445178-3445178 on Build GRCh38
rs762662696 Chr.4: 3445030-3445030 on Build GRCh38
rs774666825 Chr.4: 3444985-3444985 on Build GRCh38
rs766476901 Chr.4: 3444855-3444855 on Build GRCh38
rs972799854 Chr.4: 3445140-3445140 on Build GRCh38
rs769319829 Chr.4: 3444814-3444814 on Build GRCh38
rs551357198 Chr.4: 3444870-3444870 on Build GRCh38
rs934925430 Chr.4: 3444821-3444821 on Build GRCh38
rs774778658 Chr.4: 3445018-3445018 on Build GRCh38
rs748006879 Chr.4: 3444908-3444908 on Build GRCh38
rs756767787 Chr.4: 3444805-3444805 on Build GRCh38
rs890030978 Chr.4: 3445063-3445063 on Build GRCh38
rs144021102 Chr.4: 3444955-3444955 on Build GRCh38
rs947640760 Chr.4: 3445169-3445169 on Build GRCh38
rs779807008 Chr.4: 3444815-3444815 on Build GRCh38
rs529061927 Chr.4: 3445062-3445062 on Build GRCh38
rs775982284 Chr.4: 3444946-3444946 on Build GRCh38
rs753187984 Chr.4: 3444861-3444861 on Build GRCh38
rs764667024 Chr.4: 3444947-3444947 on Build GRCh38
rs3752441 Chr.4: 3445093-3445093 on Build GRCh38
rs762865732 Chr.4: 3444762-3444762 on Build GRCh38
rs111607130 Chr.4: 3445100-3445100 on Build GRCh38
rs931467361 Chr.4: 3445031-3445031 on Build GRCh38
rs998564160 Chr.4: 3445087-3445087 on Build GRCh38
rs200605772 Chr.4: 3445017-3445017 on Build GRCh38
rs1046320110 Chr.4: 3444806-3444806 on Build GRCh38
rs536845424 Chr.4: 3445189-3445189 on Build GRCh38
rs772972316 Chr.4: 3445036-3445037 on Build GRCh38
rs1049846453 Chr.4: 3445159-3445159 on Build GRCh38
rs781039918 Chr.4: 3445005-3445005 on Build GRCh38
rs560624553 Chr.4: 3444823-3444823 on Build GRCh38
rs562460119 Chr.4: 3445194-3445194 on Build GRCh38
rs1041683303 Chr.4: 3445186-3445186 on Build GRCh38
rs889873680 Chr.4: 3444876-3444876 on Build GRCh38
rs769241863 Chr.4: 3444770-3444770 on Build GRCh38
rs763370594 Chr.4: 3444976-3444976 on Build GRCh38
rs201137715 Chr.4: 3444793-3444793 on Build GRCh38
rs145550937 Chr.4: 3444794-3444794 on Build GRCh38
rs762144888 Chr.4: 3445020-3445020 on Build GRCh38
rs755531367 Chr.4: 3444952-3444952 on Build GRCh38
rs79184809 Chr.4: 3444756-3444756 on Build GRCh38
rs777811625 Chr.4: 3445133-3445133 on Build GRCh38
rs748995859 Chr.4: 3444885-3444885 on Build GRCh38
rs749535855 Chr.4: 3445016-3445016 on Build GRCh38
rs770027069 Chr.4: 3445008-3445008 on Build GRCh38
rs367795146 Chr.4: 3444787-3444787 on Build GRCh38
rs1045731346 Chr.4: 3444833-3444833 on Build GRCh38
rs745518539 Chr.4: 3444753-3444753 on Build GRCh38
rs981394745 Chr.4: 3445013-3445013 on Build GRCh38
rs751376718 Chr.4: 3445003-3445003 on Build GRCh38
rs182798104 Chr.4: 3445082-3445082 on Build GRCh38
rs773163201 Chr.4: 3444848-3444848 on Build GRCh38
rs769088341 Chr.4: 3444980-3444980 on Build GRCh38
rs755317395 Chr.4: 3445045-3445045 on Build GRCh38
rs780853328 Chr.4: 3444808-3444808 on Build GRCh38
rs776289947 Chr.4: 3444921-3444921 on Build GRCh38
rs751024138 Chr.4: 3444993-3444993 on Build GRCh38
rs373478970 Chr.4: 3444992-3444992 on Build GRCh38
rs369359490 Chr.4: 3444919-3444919 on Build GRCh38
rs989989783 Chr.4: 3444807-3444807 on Build GRCh38
rs773239142 Chr.4: 3444914-3444914 on Build GRCh38
rs1011350994 Chr.4: 3445094-3445094 on Build GRCh38
rs17805201 Chr.4: 3444786-3444786 on Build GRCh38
rs765620892 Chr.4: 3444943-3444943 on Build GRCh38
rs573243423 Chr.4: 3444961-3444961 on Build GRCh38
rs534595424 Chr.4: 3444866-3444866 on Build GRCh38
rs375562342 Chr.4: 3444953-3444953 on Build GRCh38
rs1049879406 Chr.4: 3444871-3444871 on Build GRCh38

View Full Product Details