Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55219517-55220029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCIDAS
Entrez Gene ID
345643
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Gene Aliases
IDAS, MCI, MCIN
UniGene
Hs.394578
Gene Chromosome Location
Chr.5: 55219597-55227315 on Build GRCh38
Transcripts
NM_001190787.1

SNP Detail

SNP ID Chromosome Location
rs770901942 Chr.5: 55219932-55219932 on Build GRCh38
rs1033946813 Chr.5: 55219625-55219625 on Build GRCh38
rs939995426 Chr.5: 55219908-55219908 on Build GRCh38
rs34303601 Chr.5: 55219550-55219551 on Build GRCh38
rs891426906 Chr.5: 55219877-55219877 on Build GRCh38
rs184503003 Chr.5: 55219936-55219936 on Build GRCh38
rs999978492 Chr.5: 55219832-55219832 on Build GRCh38
rs368586145 Chr.5: 55219554-55219554 on Build GRCh38
rs776488742 Chr.5: 55219878-55219878 on Build GRCh38
rs917185931 Chr.5: 55219672-55219672 on Build GRCh38
rs977227413 Chr.5: 55219722-55219722 on Build GRCh38
rs1007459366 Chr.5: 55219897-55219897 on Build GRCh38
rs141631566 Chr.5: 55219952-55219955 on Build GRCh38
rs190485691 Chr.5: 55219665-55219665 on Build GRCh38
rs963116369 Chr.5: 55219921-55219921 on Build GRCh38
rs180824371 Chr.5: 55219778-55219778 on Build GRCh38
rs13167348 Chr.5: 55219573-55219573 on Build GRCh38
rs976520035 Chr.5: 55219818-55219818 on Build GRCh38
rs539885573 Chr.5: 55219765-55219765 on Build GRCh38
rs544360779 Chr.5: 55219581-55219581 on Build GRCh38
rs529535811 Chr.5: 55219783-55219783 on Build GRCh38
rs890070576 Chr.5: 55219891-55219891 on Build GRCh38
rs918626689 Chr.5: 55219646-55219646 on Build GRCh38
rs747599737 Chr.5: 55219705-55219705 on Build GRCh38
rs960664978 Chr.5: 55219970-55219970 on Build GRCh38
rs1017137177 Chr.5: 55219912-55219912 on Build GRCh38
rs1016079125 Chr.5: 55219561-55219562 on Build GRCh38
rs1029897937 Chr.5: 55219880-55219880 on Build GRCh38
rs202130095 Chr.5: 55219647-55219647 on Build GRCh38
rs776756893 Chr.5: 55219972-55219972 on Build GRCh38
rs904257756 Chr.5: 55219794-55219794 on Build GRCh38
rs992266055 Chr.5: 55219563-55219563 on Build GRCh38
rs972701401 Chr.5: 55219632-55219632 on Build GRCh38
rs760430457 Chr.5: 55219659-55219659 on Build GRCh38
rs962858208 Chr.5: 55219600-55219600 on Build GRCh38
rs968782520 Chr.5: 55219686-55219686 on Build GRCh38
rs1052765761 Chr.5: 55219863-55219863 on Build GRCh38
rs461103 Chr.5: 55219650-55219650 on Build GRCh38
rs115172095 Chr.5: 55219574-55219574 on Build GRCh38
rs185563800 Chr.5: 55219661-55219661 on Build GRCh38
rs954328565 Chr.5: 55219556-55219556 on Build GRCh38
rs372201515 Chr.5: 55219555-55219555 on Build GRCh38
rs959578778 Chr.5: 55219638-55219638 on Build GRCh38
rs972753864 Chr.5: 55219933-55219933 on Build GRCh38
rs997866615 Chr.5: 55219879-55219879 on Build GRCh38
rs1055261442 Chr.5: 55219987-55219987 on Build GRCh38
rs921181825 Chr.5: 55219869-55219869 on Build GRCh38
rs932609193 Chr.5: 55219871-55219871 on Build GRCh38

View Full Product Details