Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55222871-55223376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCIDAS
Entrez Gene ID
345643
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Gene Aliases
IDAS, MCI, MCIN
UniGene
Hs.394578
Gene Chromosome Location
Chr.5: 55219597-55227315 on Build GRCh38
Transcripts
NM_001190787.1 XM_017009439.1

SNP Detail

SNP ID Chromosome Location
rs902790990 Chr.5: 55223074-55223074 on Build GRCh38
rs766135658 Chr.5: 55223154-55223154 on Build GRCh38
rs542498059 Chr.5: 55223242-55223242 on Build GRCh38
rs551260620 Chr.5: 55223009-55223009 on Build GRCh38
rs943340536 Chr.5: 55223327-55223327 on Build GRCh38
rs77412861 Chr.5: 55223103-55223103 on Build GRCh38
rs937813679 Chr.5: 55222909-55222909 on Build GRCh38
rs893440498 Chr.5: 55222933-55222933 on Build GRCh38
rs1037803235 Chr.5: 55223340-55223340 on Build GRCh38
rs1051677967 Chr.5: 55222969-55222969 on Build GRCh38
rs116674414 Chr.5: 55223129-55223129 on Build GRCh38
rs957912084 Chr.5: 55223230-55223230 on Build GRCh38
rs965249833 Chr.5: 55223127-55223127 on Build GRCh38
rs1009466027 Chr.5: 55223094-55223094 on Build GRCh38
rs558142215 Chr.5: 55222923-55222923 on Build GRCh38
rs1033915071 Chr.5: 55223203-55223203 on Build GRCh38
rs183568273 Chr.5: 55223080-55223080 on Build GRCh38
rs761652759 Chr.5: 55223134-55223134 on Build GRCh38
rs933332259 Chr.5: 55222966-55222966 on Build GRCh38
rs1055283977 Chr.5: 55222913-55222913 on Build GRCh38
rs1009191059 Chr.5: 55223357-55223357 on Build GRCh38
rs369427328 Chr.5: 55223165-55223165 on Build GRCh38
rs766291674 Chr.5: 55223064-55223064 on Build GRCh38
rs908456802 Chr.5: 55223162-55223162 on Build GRCh38
rs889114106 Chr.5: 55223000-55223000 on Build GRCh38
rs952024315 Chr.5: 55223152-55223152 on Build GRCh38
rs927779535 Chr.5: 55223201-55223201 on Build GRCh38
rs749884656 Chr.5: 55222911-55222911 on Build GRCh38
rs868179615 Chr.5: 55223256-55223256 on Build GRCh38
rs902755473 Chr.5: 55223332-55223332 on Build GRCh38
rs960237511 Chr.5: 55223118-55223118 on Build GRCh38
rs983442534 Chr.5: 55223154-55223154 on Build GRCh38
rs193060025 Chr.5: 55223071-55223071 on Build GRCh38
rs573640659 Chr.5: 55223229-55223229 on Build GRCh38
rs778663198 Chr.5: 55223190-55223190 on Build GRCh38
rs75132370 Chr.5: 55223283-55223283 on Build GRCh38
rs910576217 Chr.5: 55223298-55223298 on Build GRCh38
rs1054850586 Chr.5: 55223251-55223251 on Build GRCh38
rs749471827 Chr.5: 55223049-55223049 on Build GRCh38
rs988346163 Chr.5: 55223253-55223253 on Build GRCh38
rs747403512 Chr.5: 55223027-55223027 on Build GRCh38
rs982353685 Chr.5: 55223177-55223177 on Build GRCh38
rs1010587200 Chr.5: 55222972-55222972 on Build GRCh38
rs1026572592 Chr.5: 55223117-55223117 on Build GRCh38
rs939989536 Chr.5: 55223167-55223167 on Build GRCh38
rs537898477 Chr.5: 55223257-55223257 on Build GRCh38
rs375060701 Chr.5: 55223350-55223350 on Build GRCh38

View Full Product Details