Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153968843-153969355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L4
Entrez Gene ID
148327
Gene Name
cAMP responsive element binding protein 3 like 4
Gene Aliases
AIBZIP, ATCE1, CREB3, CREB4, JAL, hJAL
UniGene
Hs.372924
Gene Chromosome Location
Chr.1: 153967679-153974364 on Build GRCh38
Transcripts
NM_001255978.1 NM_001255979.1 NM_001255980.1 NM_001255981.1 NM_130898.3 NR_045658.1 XM_006711172.2 XM_017000372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635317 c.422-8T>C p.? Chr.1: 153969326-153969326 on Build GRCh38
4823149 c.328C>T p.P110S Chr.1: 153969083-153969083 on Build GRCh38
1202266 c.176G>A p.G59D Chr.1: 153968931-153968931 on Build GRCh38
4762911 c.359G>A p.G120E Chr.1: 153969114-153969114 on Build GRCh38
7103690 c.244G>A p.E82K Chr.1: 153968999-153968999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550062829 Chr.1: 153968903-153968904 on Build GRCh38
rs574619822 Chr.1: 153968981-153968981 on Build GRCh38
rs745696050 Chr.1: 153969041-153969041 on Build GRCh38
rs759117656 Chr.1: 153968901-153968901 on Build GRCh38
rs753981486 Chr.1: 153969330-153969330 on Build GRCh38
rs981772956 Chr.1: 153969059-153969059 on Build GRCh38
rs547438158 Chr.1: 153969314-153969314 on Build GRCh38
rs556173822 Chr.1: 153968890-153968890 on Build GRCh38
rs751629488 Chr.1: 153969027-153969027 on Build GRCh38
rs774374216 Chr.1: 153969160-153969160 on Build GRCh38
rs369495321 Chr.1: 153968912-153968913 on Build GRCh38
rs1010300703 Chr.1: 153969015-153969015 on Build GRCh38
rs757762343 Chr.1: 153968921-153968921 on Build GRCh38
rs774035804 Chr.1: 153968955-153968955 on Build GRCh38
rs201475812 Chr.1: 153969089-153969089 on Build GRCh38
rs778769666 Chr.1: 153969109-153969109 on Build GRCh38
rs376194614 Chr.1: 153969163-153969163 on Build GRCh38
rs767577986 Chr.1: 153969278-153969278 on Build GRCh38
rs759948314 Chr.1: 153969302-153969302 on Build GRCh38
rs758616713 Chr.1: 153969013-153969013 on Build GRCh38
rs763524911 Chr.1: 153969176-153969176 on Build GRCh38
rs541913338 Chr.1: 153968987-153968987 on Build GRCh38
rs764693441 Chr.1: 153968975-153968975 on Build GRCh38
rs774723060 Chr.1: 153969184-153969184 on Build GRCh38
rs530328072 Chr.1: 153968870-153968871 on Build GRCh38
rs760964123 Chr.1: 153969318-153969318 on Build GRCh38
rs774673906 Chr.1: 153969296-153969296 on Build GRCh38
rs962412715 Chr.1: 153968916-153968916 on Build GRCh38
rs576653821 Chr.1: 153968868-153968868 on Build GRCh38
rs912223350 Chr.1: 153969192-153969192 on Build GRCh38
rs780188805 Chr.1: 153969021-153969021 on Build GRCh38
rs758061438 Chr.1: 153969124-153969124 on Build GRCh38
rs200562464 Chr.1: 153968914-153968914 on Build GRCh38
rs770695846 Chr.1: 153968947-153968947 on Build GRCh38
rs372290632 Chr.1: 153968907-153968907 on Build GRCh38
rs758429547 Chr.1: 153969228-153969228 on Build GRCh38
rs761976423 Chr.1: 153969190-153969190 on Build GRCh38
rs369936791 Chr.1: 153968871-153968872 on Build GRCh38
rs773664133 Chr.1: 153969174-153969174 on Build GRCh38
rs762342236 Chr.1: 153969064-153969064 on Build GRCh38
rs45600231 Chr.1: 153969091-153969091 on Build GRCh38
rs375085579 Chr.1: 153969216-153969216 on Build GRCh38
rs779957505 Chr.1: 153969242-153969242 on Build GRCh38
rs766675043 Chr.1: 153969181-153969181 on Build GRCh38
rs200303165 Chr.1: 153969116-153969116 on Build GRCh38
rs529365833 Chr.1: 153969207-153969207 on Build GRCh38
rs372931908 Chr.1: 153969147-153969147 on Build GRCh38
rs767772588 Chr.1: 153969188-153969188 on Build GRCh38
rs150082517 Chr.1: 153969058-153969058 on Build GRCh38
rs764238210 Chr.1: 153969214-153969214 on Build GRCh38
rs757192742 Chr.1: 153969105-153969105 on Build GRCh38
rs369304052 Chr.1: 153969129-153969129 on Build GRCh38
rs555731932 Chr.1: 153969313-153969313 on Build GRCh38
rs776305491 Chr.1: 153969062-153969062 on Build GRCh38
rs377029794 Chr.1: 153968927-153968927 on Build GRCh38
rs142562840 Chr.1: 153969087-153969087 on Build GRCh38
rs777750105 Chr.1: 153968931-153968931 on Build GRCh38
rs747131844 Chr.1: 153968930-153968930 on Build GRCh38
rs754655658 Chr.1: 153969254-153969254 on Build GRCh38
rs777500049 Chr.1: 153968925-153968925 on Build GRCh38
rs765590366 Chr.1: 153968990-153968990 on Build GRCh38
rs953766192 Chr.1: 153969213-153969213 on Build GRCh38
rs372716042 Chr.1: 153968972-153968972 on Build GRCh38
rs74369324 Chr.1: 153969126-153969126 on Build GRCh38
rs1039387102 Chr.1: 153969197-153969197 on Build GRCh38
rs762789225 Chr.1: 153968883-153968883 on Build GRCh38
rs113851579 Chr.1: 153969234-153969234 on Build GRCh38
rs374412874 Chr.1: 153969326-153969326 on Build GRCh38
rs61038976 Chr.1: 153969240-153969240 on Build GRCh38
rs770321733 Chr.1: 153969170-153969170 on Build GRCh38
rs771465342 Chr.1: 153969290-153969290 on Build GRCh38
rs759721564 Chr.1: 153969186-153969186 on Build GRCh38
rs375950642 Chr.1: 153969194-153969194 on Build GRCh38
rs146937006 Chr.1: 153969030-153969030 on Build GRCh38
rs764029795 Chr.1: 153969320-153969320 on Build GRCh38
rs996387914 Chr.1: 153968932-153968932 on Build GRCh38
rs747693292 Chr.1: 153969262-153969262 on Build GRCh38
rs752757018 Chr.1: 153969104-153969104 on Build GRCh38
rs766586944 Chr.1: 153969088-153969088 on Build GRCh38
rs758003565 Chr.1: 153968896-153968896 on Build GRCh38
rs137977772 Chr.1: 153969081-153969081 on Build GRCh38
rs866015133 Chr.1: 153969222-153969222 on Build GRCh38
rs142679432 Chr.1: 153968879-153968879 on Build GRCh38
rs752284305 Chr.1: 153968908-153968908 on Build GRCh38
rs368362471 Chr.1: 153968966-153968966 on Build GRCh38
rs758572361 Chr.1: 153968929-153968929 on Build GRCh38
rs1005433438 Chr.1: 153968909-153968909 on Build GRCh38
rs11264743 Chr.1: 153969038-153969038 on Build GRCh38
rs750785366 Chr.1: 153969009-153969009 on Build GRCh38
rs145524837 Chr.1: 153969060-153969060 on Build GRCh38
rs771742358 Chr.1: 153969050-153969050 on Build GRCh38
rs746841106 Chr.1: 153969250-153969250 on Build GRCh38
rs368055346 Chr.1: 153968900-153968900 on Build GRCh38
rs749081011 Chr.1: 153968933-153968933 on Build GRCh38
rs767543282 Chr.1: 153969093-153969093 on Build GRCh38
rs767000961 Chr.1: 153968906-153968906 on Build GRCh38
rs754240817 Chr.1: 153968915-153968915 on Build GRCh38
rs966995997 Chr.1: 153969277-153969277 on Build GRCh38
rs373287482 Chr.1: 153969287-153969287 on Build GRCh38
rs781256369 Chr.1: 153969032-153969032 on Build GRCh38
rs749168871 Chr.1: 153969035-153969035 on Build GRCh38
rs371924090 Chr.1: 153968882-153968882 on Build GRCh38
rs775113088 Chr.1: 153968969-153968969 on Build GRCh38

View Full Product Details