Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 156681189-156681687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMBR1
Entrez Gene ID
64327
Gene Name
limb development membrane protein 1
Gene Aliases
ACHP, C7orf2, DIF14, LSS, PPD2, THYP, TPT, ZRS
UniGene
Hs.209989
Gene Chromosome Location
Chr.7: 156673354-156893208 on Build GRCh38
Transcripts
NM_022458.3 XM_005249555.2 XM_005249558.2 XM_017012507.1 XM_017012509.1 XM_017012514.1 XM_017012515.1 XM_017012517.1 XM_017012518.1

SNP Detail

SNP ID Chromosome Location
rs535905178 Chr.7: 156681646-156681646 on Build GRCh38
rs1028037715 Chr.7: 156681415-156681415 on Build GRCh38
rs566692462 Chr.7: 156681578-156681578 on Build GRCh38
rs934421288 Chr.7: 156681528-156681528 on Build GRCh38
rs778223242 Chr.7: 156681231-156681231 on Build GRCh38
rs775444879 Chr.7: 156681372-156681372 on Build GRCh38
rs182572414 Chr.7: 156681317-156681317 on Build GRCh38
rs983385501 Chr.7: 156681654-156681654 on Build GRCh38
rs772161675 Chr.7: 156681559-156681559 on Build GRCh38
rs1035084510 Chr.7: 156681659-156681659 on Build GRCh38
rs576012030 Chr.7: 156681264-156681264 on Build GRCh38
rs908376896 Chr.7: 156681247-156681247 on Build GRCh38
rs916828732 Chr.7: 156681275-156681275 on Build GRCh38
rs929355715 Chr.7: 156681224-156681224 on Build GRCh38
rs748953678 Chr.7: 156681368-156681368 on Build GRCh38
rs757744585 Chr.7: 156681244-156681244 on Build GRCh38
rs983498368 Chr.7: 156681437-156681437 on Build GRCh38
rs911144502 Chr.7: 156681615-156681615 on Build GRCh38
rs746253016 Chr.7: 156681280-156681280 on Build GRCh38
rs1014236 Chr.7: 156681475-156681475 on Build GRCh38
rs1038448890 Chr.7: 156681641-156681641 on Build GRCh38
rs1012401792 Chr.7: 156681519-156681519 on Build GRCh38
rs920709970 Chr.7: 156681211-156681211 on Build GRCh38
rs551377904 Chr.7: 156681574-156681574 on Build GRCh38
rs1009592190 Chr.7: 156681319-156681319 on Build GRCh38
rs894353723 Chr.7: 156681259-156681259 on Build GRCh38
rs763650373 Chr.7: 156681479-156681479 on Build GRCh38
rs890610050 Chr.7: 156681318-156681318 on Build GRCh38
rs938642022 Chr.7: 156681250-156681250 on Build GRCh38
rs915831166 Chr.7: 156681466-156681466 on Build GRCh38
rs1035154101 Chr.7: 156681233-156681233 on Build GRCh38
rs151167124 Chr.7: 156681601-156681601 on Build GRCh38
rs961032830 Chr.7: 156681666-156681666 on Build GRCh38
rs533101615 Chr.7: 156681534-156681534 on Build GRCh38
rs373702651 Chr.7: 156681633-156681633 on Build GRCh38
rs191993720 Chr.7: 156681589-156681589 on Build GRCh38
rs993045460 Chr.7: 156681457-156681457 on Build GRCh38
rs908409304 Chr.7: 156681444-156681444 on Build GRCh38
rs781634358 Chr.7: 156681276-156681276 on Build GRCh38
rs550075183 Chr.7: 156681258-156681258 on Build GRCh38
rs4716440 Chr.7: 156681227-156681227 on Build GRCh38
rs1052843386 Chr.7: 156681555-156681555 on Build GRCh38
rs562094507 Chr.7: 156681498-156681498 on Build GRCh38
rs114295738 Chr.7: 156681469-156681469 on Build GRCh38
rs756411300 Chr.7: 156681307-156681307 on Build GRCh38
rs994353158 Chr.7: 156681232-156681232 on Build GRCh38
rs1049558063 Chr.7: 156681604-156681604 on Build GRCh38
rs2870 Chr.7: 156681567-156681567 on Build GRCh38
rs904280378 Chr.7: 156681305-156681305 on Build GRCh38
rs938529520 Chr.7: 156681448-156681448 on Build GRCh38
rs181517016 Chr.7: 156681626-156681626 on Build GRCh38
rs1017870786 Chr.7: 156681356-156681356 on Build GRCh38
rs886062111 Chr.7: 156681305-156681305 on Build GRCh38
rs965307754 Chr.7: 156681378-156681378 on Build GRCh38
rs998597268 Chr.7: 156681308-156681308 on Build GRCh38
rs574720242 Chr.7: 156681218-156681218 on Build GRCh38
rs540352987 Chr.7: 156681461-156681461 on Build GRCh38
rs886062112 Chr.7: 156681334-156681334 on Build GRCh38
rs868712224 Chr.7: 156681592-156681592 on Build GRCh38
rs776143065 Chr.7: 156681662-156681662 on Build GRCh38
rs773774639 Chr.7: 156681416-156681416 on Build GRCh38
rs550168152 Chr.7: 156681482-156681482 on Build GRCh38
rs950703982 Chr.7: 156681638-156681638 on Build GRCh38
rs189171531 Chr.7: 156681279-156681279 on Build GRCh38
rs187134101 Chr.7: 156681411-156681411 on Build GRCh38
rs1052891075 Chr.7: 156681359-156681359 on Build GRCh38

View Full Product Details