Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244408738-244409245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSS
Entrez Gene ID
159
Gene Name
adenylosuccinate synthase
Gene Aliases
ADEH, ADSS 2
UniGene
Hs.498313
Gene Chromosome Location
Chr.1: 244408492-244452134 on Build GRCh38
Transcripts
NM_001126.3 XM_011544108.2

SNP Detail

SNP ID Chromosome Location
rs940915185 Chr.1: 244409011-244409011 on Build GRCh38
rs765607331 Chr.1: 244408906-244408906 on Build GRCh38
rs915495189 Chr.1: 244409095-244409095 on Build GRCh38
rs903790016 Chr.1: 244409125-244409125 on Build GRCh38
rs188643354 Chr.1: 244409064-244409064 on Build GRCh38
rs953161502 Chr.1: 244409007-244409007 on Build GRCh38
rs548686962 Chr.1: 244409117-244409117 on Build GRCh38
rs938796038 Chr.1: 244409054-244409054 on Build GRCh38
rs984436332 Chr.1: 244409008-244409008 on Build GRCh38
rs533528247 Chr.1: 244408912-244408912 on Build GRCh38
rs999373806 Chr.1: 244408923-244408923 on Build GRCh38
rs1011993362 Chr.1: 244409083-244409083 on Build GRCh38
rs975817351 Chr.1: 244409000-244409000 on Build GRCh38
rs953552360 Chr.1: 244409126-244409126 on Build GRCh38
rs1036505335 Chr.1: 244409034-244409034 on Build GRCh38
rs986287338 Chr.1: 244409200-244409200 on Build GRCh38
rs1030816931 Chr.1: 244408931-244408931 on Build GRCh38
rs999426115 Chr.1: 244409166-244409166 on Build GRCh38
rs1003565295 Chr.1: 244409042-244409042 on Build GRCh38
rs550440493 Chr.1: 244408922-244408922 on Build GRCh38
rs1012198922 Chr.1: 244408850-244408850 on Build GRCh38
rs909465281 Chr.1: 244409010-244409010 on Build GRCh38
rs1048957791 Chr.1: 244408997-244408997 on Build GRCh38
rs565636331 Chr.1: 244409124-244409124 on Build GRCh38
rs761772668 Chr.1: 244409058-244409058 on Build GRCh38
rs763195509 Chr.1: 244409069-244409069 on Build GRCh38
rs183595622 Chr.1: 244409050-244409050 on Build GRCh38
rs774705210 Chr.1: 244408962-244408962 on Build GRCh38
rs1022631568 Chr.1: 244408856-244408856 on Build GRCh38
rs1030869108 Chr.1: 244409189-244409189 on Build GRCh38
rs965809809 Chr.1: 244408979-244408979 on Build GRCh38
rs35885954 Chr.1: 244408848-244408849 on Build GRCh38
rs762121079 Chr.1: 244408795-244408795 on Build GRCh38
rs928787374 Chr.1: 244409039-244409039 on Build GRCh38
rs1043027809 Chr.1: 244409096-244409096 on Build GRCh38
rs921653783 Chr.1: 244409004-244409004 on Build GRCh38

View Full Product Details