Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 37141705-37142207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30A
Entrez Gene ID
91074
Gene Name
ankyrin repeat domain 30A
Gene Aliases
NY-BR-1
UniGene
Hs.373787
Gene Chromosome Location
Chr.10: 37125754-37235831 on Build GRCh38
Transcripts
NM_052997.2 XM_011519757.2 XM_011519758.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3437510 c.882C>T p.F294F Chr.10: 37141947-37141947 on Build GRCh38
5543716 c.1031A>G p.K344R Chr.10: 37142096-37142096 on Build GRCh38
6873202 c.1092G>A p.E364E Chr.10: 37142157-37142157 on Build GRCh38
6289053 c.744T>C p.D248D Chr.10: 37141809-37141809 on Build GRCh38
1956296 c.917C>T p.T306M Chr.10: 37141982-37141982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764065320 Chr.10: 37141747-37141747 on Build GRCh38
rs777713917 Chr.10: 37142141-37142142 on Build GRCh38
rs780664888 Chr.10: 37142025-37142025 on Build GRCh38
rs753599310 Chr.10: 37141748-37141748 on Build GRCh38
rs78606913 Chr.10: 37141986-37141986 on Build GRCh38
rs757036698 Chr.10: 37141751-37141751 on Build GRCh38
rs759851482 Chr.10: 37141959-37141959 on Build GRCh38
rs368871754 Chr.10: 37142127-37142127 on Build GRCh38
rs553767718 Chr.10: 37142181-37142181 on Build GRCh38
rs764009533 Chr.10: 37142139-37142139 on Build GRCh38
rs754240233 Chr.10: 37142072-37142072 on Build GRCh38
rs776119578 Chr.10: 37141934-37141934 on Build GRCh38
rs879160888 Chr.10: 37141865-37141865 on Build GRCh38
rs771060239 Chr.10: 37142116-37142116 on Build GRCh38
rs763623807 Chr.10: 37141820-37141820 on Build GRCh38
rs759835090 Chr.10: 37142052-37142052 on Build GRCh38
rs779348229 Chr.10: 37141809-37141810 on Build GRCh38
rs372199195 Chr.10: 37141875-37141875 on Build GRCh38
rs369194109 Chr.10: 37141761-37141761 on Build GRCh38
rs756586414 Chr.10: 37141872-37141872 on Build GRCh38
rs377740138 Chr.10: 37141792-37141792 on Build GRCh38
rs1022247968 Chr.10: 37141737-37141737 on Build GRCh38
rs371384886 Chr.10: 37141931-37141931 on Build GRCh38
rs374804721 Chr.10: 37142118-37142118 on Build GRCh38
rs767910551 Chr.10: 37142055-37142055 on Build GRCh38
rs755336380 Chr.10: 37141795-37141796 on Build GRCh38
rs768361506 Chr.10: 37142016-37142016 on Build GRCh38
rs745821773 Chr.10: 37142179-37142179 on Build GRCh38
rs750757638 Chr.10: 37142150-37142150 on Build GRCh38
rs927720500 Chr.10: 37141917-37141917 on Build GRCh38
rs368398639 Chr.10: 37141983-37141983 on Build GRCh38
rs778197641 Chr.10: 37141878-37141878 on Build GRCh38
rs777357067 Chr.10: 37142103-37142103 on Build GRCh38
rs751784076 Chr.10: 37141957-37141957 on Build GRCh38
rs769759631 Chr.10: 37141948-37141948 on Build GRCh38
rs748696914 Chr.10: 37141778-37141778 on Build GRCh38
rs766759978 Chr.10: 37141845-37141845 on Build GRCh38
rs752788472 Chr.10: 37142056-37142056 on Build GRCh38
rs778182851 Chr.10: 37141976-37141976 on Build GRCh38
rs760497000 Chr.10: 37142105-37142105 on Build GRCh38
rs201989974 Chr.10: 37141729-37141729 on Build GRCh38
rs866013853 Chr.10: 37142058-37142058 on Build GRCh38
rs766792327 Chr.10: 37142153-37142153 on Build GRCh38
rs765013969 Chr.10: 37141827-37141827 on Build GRCh38
rs750249237 Chr.10: 37141841-37141841 on Build GRCh38
rs372420008 Chr.10: 37141994-37141994 on Build GRCh38
rs867081731 Chr.10: 37141892-37141892 on Build GRCh38
rs200939078 Chr.10: 37141743-37141743 on Build GRCh38
rs755796064 Chr.10: 37142158-37142158 on Build GRCh38
rs760965708 Chr.10: 37142125-37142125 on Build GRCh38
rs781444410 Chr.10: 37141869-37141869 on Build GRCh38
rs879085220 Chr.10: 37142059-37142059 on Build GRCh38
rs878859148 Chr.10: 37142060-37142060 on Build GRCh38
rs749662238 Chr.10: 37141901-37141901 on Build GRCh38
rs777144209 Chr.10: 37142168-37142168 on Build GRCh38
rs552183541 Chr.10: 37142062-37142062 on Build GRCh38
rs778952605 Chr.10: 37142178-37142178 on Build GRCh38
rs775525773 Chr.10: 37142124-37142124 on Build GRCh38
rs867761806 Chr.10: 37141794-37141795 on Build GRCh38
rs776498766 Chr.10: 37141822-37141822 on Build GRCh38
rs764960024 Chr.10: 37141757-37141757 on Build GRCh38
rs769743263 Chr.10: 37142030-37142030 on Build GRCh38
rs200193852 Chr.10: 37141982-37141982 on Build GRCh38
rs200264724 Chr.10: 37142117-37142117 on Build GRCh38
rs961265103 Chr.10: 37142180-37142180 on Build GRCh38
rs752788101 Chr.10: 37141967-37141967 on Build GRCh38
rs751663501 Chr.10: 37141854-37141854 on Build GRCh38
rs533890211 Chr.10: 37142152-37142152 on Build GRCh38
rs12773436 Chr.10: 37142169-37142169 on Build GRCh38
rs201493184 Chr.10: 37141962-37141962 on Build GRCh38
rs866087611 Chr.10: 37141889-37141889 on Build GRCh38
rs754605525 Chr.10: 37142087-37142087 on Build GRCh38
rs754399162 Chr.10: 37141979-37141979 on Build GRCh38
rs200560040 Chr.10: 37141779-37141779 on Build GRCh38
rs77379875 Chr.10: 37141975-37141975 on Build GRCh38
rs547420743 Chr.10: 37141817-37141817 on Build GRCh38
rs374024060 Chr.10: 37142015-37142015 on Build GRCh38
rs144539033 Chr.10: 37142050-37142050 on Build GRCh38
rs773464055 Chr.10: 37141787-37141787 on Build GRCh38
rs767487436 Chr.10: 37141844-37141844 on Build GRCh38
rs529585679 Chr.10: 37141760-37141760 on Build GRCh38
rs76202571 Chr.10: 37141925-37141925 on Build GRCh38
rs267602478 Chr.10: 37142148-37142148 on Build GRCh38
rs878964411 Chr.10: 37141989-37141989 on Build GRCh38
rs750597468 Chr.10: 37141759-37141759 on Build GRCh38
rs756620752 Chr.10: 37141973-37141973 on Build GRCh38
rs200845385 Chr.10: 37141868-37141868 on Build GRCh38
rs772941971 Chr.10: 37141870-37141870 on Build GRCh38
rs375389390 Chr.10: 37142173-37142173 on Build GRCh38
rs200315495 Chr.10: 37141730-37141730 on Build GRCh38
rs372362957 Chr.10: 37142149-37142149 on Build GRCh38
rs752237951 Chr.10: 37142155-37142155 on Build GRCh38
rs772987097 Chr.10: 37142047-37142047 on Build GRCh38
rs781775542 Chr.10: 37141772-37141772 on Build GRCh38
rs748636471 Chr.10: 37141812-37141844 on Build GRCh38
rs760630514 Chr.10: 37142057-37142057 on Build GRCh38
rs560930371 Chr.10: 37141904-37141904 on Build GRCh38
rs762291547 Chr.10: 37142144-37142144 on Build GRCh38
rs879018987 Chr.10: 37141857-37141857 on Build GRCh38
rs774956263 Chr.10: 37141815-37141815 on Build GRCh38
rs35687031 Chr.10: 37141910-37141911 on Build GRCh38
rs200589399 Chr.10: 37141968-37141968 on Build GRCh38
rs747663870 Chr.10: 37142029-37142029 on Build GRCh38
rs761664646 Chr.10: 37141825-37141825 on Build GRCh38
rs202098264 Chr.10: 37141947-37141947 on Build GRCh38
rs757761864 Chr.10: 37142076-37142076 on Build GRCh38
rs548512897 Chr.10: 37141969-37141969 on Build GRCh38
rs879215737 Chr.10: 37142020-37142020 on Build GRCh38
rs202090351 Chr.10: 37141771-37141771 on Build GRCh38
rs371443557 Chr.10: 37142082-37142082 on Build GRCh38
rs868304038 Chr.10: 37142046-37142046 on Build GRCh38
rs770792408 Chr.10: 37142048-37142048 on Build GRCh38
rs202201583 Chr.10: 37142092-37142092 on Build GRCh38
rs752210218 Chr.10: 37141734-37141734 on Build GRCh38
rs796295970 Chr.10: 37141819-37141819 on Build GRCh38
rs199969776 Chr.10: 37142091-37142091 on Build GRCh38
rs749114201 Chr.10: 37142106-37142106 on Build GRCh38
rs760592156 Chr.10: 37141738-37141738 on Build GRCh38
rs776835391 Chr.10: 37142140-37142140 on Build GRCh38
rs201391167 Chr.10: 37142122-37142122 on Build GRCh38
rs748866515 Chr.10: 37142172-37142172 on Build GRCh38
rs768093815 Chr.10: 37141932-37141932 on Build GRCh38
rs199611649 Chr.10: 37141782-37141782 on Build GRCh38
rs879036982 Chr.10: 37141909-37141909 on Build GRCh38
rs771678462 Chr.10: 37141810-37141810 on Build GRCh38
rs565638896 Chr.10: 37142102-37142102 on Build GRCh38

View Full Product Details