Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 37149573-37150084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30A
Entrez Gene ID
91074
Gene Name
ankyrin repeat domain 30A
Gene Aliases
NY-BR-1
UniGene
Hs.373787
Gene Chromosome Location
Chr.10: 37125754-37235831 on Build GRCh38
Transcripts
NM_052997.2 XM_011519757.2 XM_011519758.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3437550 c.1401C>T p.F467F Chr.10: 37149676-37149676 on Build GRCh38
3437548 c.1386G>A p.K462K Chr.10: 37149661-37149661 on Build GRCh38
3437554 c.1452G>A p.L484L Chr.10: 37149824-37149824 on Build GRCh38
4999897 c.1425C>G p.N475K Chr.10: 37149797-37149797 on Build GRCh38
3978470 c.1376-3T>A p.? Chr.10: 37149648-37149648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200039777 Chr.10: 37149610-37149610 on Build GRCh38
rs780293273 Chr.10: 37149616-37149616 on Build GRCh38
rs376821949 Chr.10: 37149844-37149844 on Build GRCh38
rs760588501 Chr.10: 37149656-37149656 on Build GRCh38
rs775772319 Chr.10: 37150043-37150053 on Build GRCh38
rs199571878 Chr.10: 37149825-37149825 on Build GRCh38
rs753282882 Chr.10: 37149700-37149700 on Build GRCh38
rs763255844 Chr.10: 37149794-37149794 on Build GRCh38
rs763942873 Chr.10: 37149601-37149601 on Build GRCh38
rs572069400 Chr.10: 37149611-37149611 on Build GRCh38
rs980619517 Chr.10: 37150000-37150000 on Build GRCh38
rs777814886 Chr.10: 37149893-37149893 on Build GRCh38
rs758502755 Chr.10: 37149612-37149612 on Build GRCh38
rs753630511 Chr.10: 37149606-37149606 on Build GRCh38
rs779641844 Chr.10: 37149815-37149815 on Build GRCh38
rs557917811 Chr.10: 37149603-37149603 on Build GRCh38
rs200260827 Chr.10: 37149797-37149797 on Build GRCh38
rs761385472 Chr.10: 37149658-37149658 on Build GRCh38
rs761823168 Chr.10: 37149759-37149759 on Build GRCh38
rs756388371 Chr.10: 37149704-37149704 on Build GRCh38
rs142471234 Chr.10: 37149882-37149882 on Build GRCh38
rs776383610 Chr.10: 37149735-37149735 on Build GRCh38
rs559249503 Chr.10: 37150046-37150046 on Build GRCh38
rs761398079 Chr.10: 37149806-37149806 on Build GRCh38
rs770589686 Chr.10: 37149628-37149628 on Build GRCh38
rs1029888297 Chr.10: 37149678-37149678 on Build GRCh38
rs759820225 Chr.10: 37149861-37149861 on Build GRCh38
rs758011391 Chr.10: 37149813-37149813 on Build GRCh38
rs754369971 Chr.10: 37149870-37149870 on Build GRCh38
rs769407441 Chr.10: 37149763-37149763 on Build GRCh38
rs767432608 Chr.10: 37149864-37149864 on Build GRCh38
rs951408458 Chr.10: 37149965-37149965 on Build GRCh38
rs202049333 Chr.10: 37149694-37149694 on Build GRCh38
rs752047598 Chr.10: 37149617-37149617 on Build GRCh38
rs963170222 Chr.10: 37149923-37149923 on Build GRCh38
rs748514445 Chr.10: 37149627-37149627 on Build GRCh38
rs764427256 Chr.10: 37149810-37149810 on Build GRCh38
rs368836727 Chr.10: 37149874-37149874 on Build GRCh38
rs560786304 Chr.10: 37149691-37149691 on Build GRCh38
rs936472130 Chr.10: 37150015-37150015 on Build GRCh38
rs969275424 Chr.10: 37149997-37149997 on Build GRCh38
rs567628202 Chr.10: 37149884-37149884 on Build GRCh38
rs369532435 Chr.10: 37149663-37149663 on Build GRCh38
rs201445322 Chr.10: 37149725-37149726 on Build GRCh38
rs957587615 Chr.10: 37149602-37149602 on Build GRCh38
rs936435275 Chr.10: 37149826-37149826 on Build GRCh38
rs751747625 Chr.10: 37149689-37149689 on Build GRCh38
rs781608245 Chr.10: 37149623-37149623 on Build GRCh38
rs201360743 Chr.10: 37149818-37149818 on Build GRCh38
rs552047613 Chr.10: 37149802-37149802 on Build GRCh38
rs267602480 Chr.10: 37149786-37149786 on Build GRCh38
rs376577847 Chr.10: 37149619-37149619 on Build GRCh38
rs749135492 Chr.10: 37149849-37149849 on Build GRCh38
rs746557883 Chr.10: 37149729-37149729 on Build GRCh38
rs775693170 Chr.10: 37149865-37149865 on Build GRCh38
rs769829463 Chr.10: 37149830-37149830 on Build GRCh38
rs951430659 Chr.10: 37149769-37149769 on Build GRCh38
rs745544497 Chr.10: 37149640-37149640 on Build GRCh38
rs766375213 Chr.10: 37149853-37149853 on Build GRCh38
rs201538199 Chr.10: 37149659-37149659 on Build GRCh38
rs995618655 Chr.10: 37149928-37149928 on Build GRCh38
rs757474847 Chr.10: 37149608-37149608 on Build GRCh38
rs778216167 Chr.10: 37149708-37149708 on Build GRCh38
rs267602481 Chr.10: 37149799-37149799 on Build GRCh38
rs377016392 Chr.10: 37149880-37149880 on Build GRCh38
rs373024622 Chr.10: 37149879-37149879 on Build GRCh38
rs779513345 Chr.10: 37149725-37149725 on Build GRCh38
rs773719390 Chr.10: 37149804-37149805 on Build GRCh38
rs755610695 Chr.10: 37149620-37149620 on Build GRCh38
rs748032163 Chr.10: 37149828-37149828 on Build GRCh38
rs775475111 Chr.10: 37149644-37149644 on Build GRCh38
rs527641248 Chr.10: 37149839-37149839 on Build GRCh38
rs933000802 Chr.10: 37150057-37150057 on Build GRCh38
rs142362140 Chr.10: 37149600-37149600 on Build GRCh38
rs72787532 Chr.10: 37149719-37149719 on Build GRCh38
rs745513666 Chr.10: 37149718-37149718 on Build GRCh38
rs768520588 Chr.10: 37149657-37149657 on Build GRCh38
rs771500416 Chr.10: 37149648-37149648 on Build GRCh38
rs766563948 Chr.10: 37149687-37149687 on Build GRCh38
rs778393411 Chr.10: 37149637-37149637 on Build GRCh38
rs774671198 Chr.10: 37149860-37149860 on Build GRCh38
rs775044197 Chr.10: 37149652-37149652 on Build GRCh38
rs551268572 Chr.10: 37149955-37149955 on Build GRCh38
rs763274185 Chr.10: 37149677-37149677 on Build GRCh38
rs766758057 Chr.10: 37149795-37149795 on Build GRCh38
rs377558375 Chr.10: 37149953-37149953 on Build GRCh38
rs764621582 Chr.10: 37149868-37149868 on Build GRCh38
rs543180343 Chr.10: 37149774-37149774 on Build GRCh38
rs78916295 Chr.10: 37149911-37149911 on Build GRCh38
rs755551682 Chr.10: 37149811-37149811 on Build GRCh38
rs768244171 Chr.10: 37149734-37149734 on Build GRCh38
rs536731413 Chr.10: 37149954-37149954 on Build GRCh38
rs114515059 Chr.10: 37150013-37150013 on Build GRCh38
rs879067354 Chr.10: 37149660-37149660 on Build GRCh38
rs192432374 Chr.10: 37149784-37149784 on Build GRCh38
rs1018399785 Chr.10: 37149701-37149701 on Build GRCh38
rs750613941 Chr.10: 37149669-37149669 on Build GRCh38
rs570254086 Chr.10: 37149982-37149982 on Build GRCh38
rs756707483 Chr.10: 37149808-37149808 on Build GRCh38
rs550040611 Chr.10: 37149979-37149979 on Build GRCh38
rs767007711 Chr.10: 37149805-37149806 on Build GRCh38
rs756088684 Chr.10: 37149891-37149891 on Build GRCh38
rs762952583 Chr.10: 37149682-37149682 on Build GRCh38
rs913620713 Chr.10: 37150048-37150048 on Build GRCh38
rs1056277425 Chr.10: 37149613-37149613 on Build GRCh38

View Full Product Details