Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96191829-96192263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLNK
Entrez Gene ID
29760
Gene Name
B-cell linker
Gene Aliases
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca
UniGene
Hs.665244
Gene Chromosome Location
Chr.10: 96189171-96271576 on Build GRCh38
Transcripts
NM_001114094.1 NM_001258440.1 NM_001258441.1 NM_001258442.1 NM_013314.3 NR_047680.1 NR_047681.1 NR_047682.1 NR_047683.1 XM_011539728.2 XM_017016159.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
428210 c.1275C>T p.I425I Chr.10: 96192069-96192069 on Build GRCh38
4815618 c.1170C>T p.I390I Chr.10: 96192069-96192069 on Build GRCh38
921885 c.1283A>C p.N428T Chr.10: 96192061-96192061 on Build GRCh38
1246134 c.1362A>C p.K454N Chr.10: 96191982-96191982 on Build GRCh38
4870634 c.1228G>T p.D410Y Chr.10: 96192011-96192011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781923122 Chr.10: 96192195-96192195 on Build GRCh38
rs782050505 Chr.10: 96192082-96192082 on Build GRCh38
rs782806042 Chr.10: 96192100-96192100 on Build GRCh38
rs1042063724 Chr.10: 96192181-96192181 on Build GRCh38
rs782063820 Chr.10: 96192113-96192113 on Build GRCh38
rs782515286 Chr.10: 96191959-96191959 on Build GRCh38
rs1057940 Chr.10: 96192005-96192005 on Build GRCh38
rs774507707 Chr.10: 96191883-96191883 on Build GRCh38
rs782296230 Chr.10: 96192053-96192053 on Build GRCh38
rs949259126 Chr.10: 96191965-96191965 on Build GRCh38
rs587730160 Chr.10: 96192207-96192207 on Build GRCh38
rs782147459 Chr.10: 96191944-96191944 on Build GRCh38
rs782280453 Chr.10: 96192025-96192025 on Build GRCh38
rs782547336 Chr.10: 96191993-96191993 on Build GRCh38
rs762002095 Chr.10: 96192030-96192030 on Build GRCh38
rs782462914 Chr.10: 96192144-96192144 on Build GRCh38
rs782422565 Chr.10: 96192192-96192192 on Build GRCh38
rs782445849 Chr.10: 96192028-96192028 on Build GRCh38
rs901452552 Chr.10: 96192123-96192123 on Build GRCh38
rs781881941 Chr.10: 96192109-96192109 on Build GRCh38
rs782736210 Chr.10: 96191969-96191969 on Build GRCh38
rs1032121557 Chr.10: 96192217-96192217 on Build GRCh38
rs782636828 Chr.10: 96192042-96192042 on Build GRCh38
rs782643739 Chr.10: 96191958-96191959 on Build GRCh38
rs782608058 Chr.10: 96192033-96192033 on Build GRCh38
rs781794307 Chr.10: 96191964-96191964 on Build GRCh38
rs918547258 Chr.10: 96191967-96191967 on Build GRCh38
rs202240472 Chr.10: 96192057-96192057 on Build GRCh38
rs781847413 Chr.10: 96191971-96191971 on Build GRCh38
rs931320409 Chr.10: 96192018-96192018 on Build GRCh38
rs782686069 Chr.10: 96192120-96192120 on Build GRCh38
rs781802342 Chr.10: 96192024-96192024 on Build GRCh38
rs782365175 Chr.10: 96192085-96192085 on Build GRCh38
rs587621098 Chr.10: 96192199-96192199 on Build GRCh38
rs782032473 Chr.10: 96191937-96191937 on Build GRCh38
rs587631005 Chr.10: 96191874-96191874 on Build GRCh38
rs917851799 Chr.10: 96191919-96191919 on Build GRCh38
rs587678167 Chr.10: 96192206-96192206 on Build GRCh38
rs200396708 Chr.10: 96191966-96191966 on Build GRCh38
rs149075195 Chr.10: 96192174-96192174 on Build GRCh38
rs145030645 Chr.10: 96192171-96192171 on Build GRCh38
rs959216291 Chr.10: 96191869-96191869 on Build GRCh38
rs781958148 Chr.10: 96192095-96192095 on Build GRCh38
rs376602258 Chr.10: 96191948-96191948 on Build GRCh38
rs781836046 Chr.10: 96192142-96192142 on Build GRCh38
rs781943787 Chr.10: 96192064-96192064 on Build GRCh38
rs782660063 Chr.10: 96192010-96192010 on Build GRCh38
rs782710142 Chr.10: 96191950-96191950 on Build GRCh38
rs372130465 Chr.10: 96192096-96192096 on Build GRCh38

View Full Product Details