Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124696080-124696588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM53B
Entrez Gene ID
9679
Gene Name
family with sequence similarity 53 member B
Gene Aliases
KIAA0140, bA12J10.2, smp
UniGene
Hs.129195
Gene Chromosome Location
Chr.10: 124619294-124744361 on Build GRCh38
Transcripts
NM_014661.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3716378 c.117C>G p.F39L Chr.10: 124696174-124696174 on Build GRCh38
6460978 c.81C>A p.H27Q Chr.10: 124696210-124696210 on Build GRCh38
4683073 c.117C>T p.F39F Chr.10: 124696174-124696174 on Build GRCh38
5739070 c.83C>T p.T28M Chr.10: 124696208-124696208 on Build GRCh38
384726 c.115T>C p.F39L Chr.10: 124696176-124696176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916613758 Chr.10: 124696545-124696545 on Build GRCh38
rs575697509 Chr.10: 124696474-124696474 on Build GRCh38
rs545727556 Chr.10: 124696125-124696126 on Build GRCh38
rs983720978 Chr.10: 124696421-124696421 on Build GRCh38
rs959040646 Chr.10: 124696391-124696391 on Build GRCh38
rs910994917 Chr.10: 124696160-124696160 on Build GRCh38
rs763575534 Chr.10: 124696209-124696209 on Build GRCh38
rs1043505397 Chr.10: 124696489-124696489 on Build GRCh38
rs137857157 Chr.10: 124696267-124696267 on Build GRCh38
rs933339338 Chr.10: 124696466-124696466 on Build GRCh38
rs568233826 Chr.10: 124696300-124696300 on Build GRCh38
rs572291558 Chr.10: 124696368-124696368 on Build GRCh38
rs750654168 Chr.10: 124696260-124696260 on Build GRCh38
rs909061739 Chr.10: 124696428-124696428 on Build GRCh38
rs941817587 Chr.10: 124696454-124696454 on Build GRCh38
rs891898565 Chr.10: 124696477-124696477 on Build GRCh38
rs1038749674 Chr.10: 124696561-124696561 on Build GRCh38
rs946138788 Chr.10: 124696482-124696482 on Build GRCh38
rs1001624731 Chr.10: 124696494-124696494 on Build GRCh38
rs762180379 Chr.10: 124696251-124696251 on Build GRCh38
rs148779480 Chr.10: 124696534-124696534 on Build GRCh38
rs921810277 Chr.10: 124696465-124696465 on Build GRCh38
rs751874221 Chr.10: 124696246-124696246 on Build GRCh38
rs950407633 Chr.10: 124696518-124696518 on Build GRCh38
rs768243782 Chr.10: 124696344-124696344 on Build GRCh38
rs986534482 Chr.10: 124696144-124696144 on Build GRCh38
rs1012660262 Chr.10: 124696397-124696397 on Build GRCh38
rs747887562 Chr.10: 124696510-124696510 on Build GRCh38
rs746783915 Chr.10: 124696139-124696139 on Build GRCh38
rs928464931 Chr.10: 124696296-124696296 on Build GRCh38
rs933267405 Chr.10: 124696272-124696272 on Build GRCh38
rs1013695772 Chr.10: 124696511-124696511 on Build GRCh38
rs974658531 Chr.10: 124696455-124696455 on Build GRCh38
rs561471835 Chr.10: 124696516-124696516 on Build GRCh38
rs777570465 Chr.10: 124696135-124696135 on Build GRCh38
rs770499081 Chr.10: 124696145-124696145 on Build GRCh38
rs372304383 Chr.10: 124696239-124696239 on Build GRCh38
rs557352635 Chr.10: 124696405-124696405 on Build GRCh38
rs1052286965 Chr.10: 124696291-124696291 on Build GRCh38
rs375070498 Chr.10: 124696258-124696258 on Build GRCh38
rs534288128 Chr.10: 124696126-124696126 on Build GRCh38
rs780083126 Chr.10: 124696336-124696336 on Build GRCh38
rs114239281 Chr.10: 124696374-124696374 on Build GRCh38
rs535318158 Chr.10: 124696311-124696311 on Build GRCh38
rs1001753852 Chr.10: 124696359-124696359 on Build GRCh38
rs1008377050 Chr.10: 124696478-124696478 on Build GRCh38
rs950480445 Chr.10: 124696408-124696408 on Build GRCh38
rs1025186719 Chr.10: 124696407-124696407 on Build GRCh38
rs776186824 Chr.10: 124696208-124696208 on Build GRCh38
rs904703448 Chr.10: 124696491-124696491 on Build GRCh38
rs776330495 Chr.10: 124696147-124696147 on Build GRCh38
rs1051767423 Chr.10: 124696470-124696470 on Build GRCh38
rs748049773 Chr.10: 124696113-124696113 on Build GRCh38
rs755235264 Chr.10: 124696390-124696390 on Build GRCh38
rs771906928 Chr.10: 124696123-124696123 on Build GRCh38
rs948172065 Chr.10: 124696549-124696549 on Build GRCh38
rs764654930 Chr.10: 124696232-124696232 on Build GRCh38
rs991378286 Chr.10: 124696539-124696539 on Build GRCh38
rs759031823 Chr.10: 124696169-124696169 on Build GRCh38
rs769394092 Chr.10: 124696180-124696180 on Build GRCh38
rs898719702 Chr.10: 124696453-124696453 on Build GRCh38
rs144371489 Chr.10: 124696503-124696503 on Build GRCh38
rs552111805 Chr.10: 124696341-124696341 on Build GRCh38

View Full Product Details