Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133625873-133626384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRG2B
Entrez Gene ID
441581
Gene Name
FSHD region gene 2 family member B
Gene Aliases
-
UniGene
Hs.690471
Gene Chromosome Location
Chr.10: 133625099-133626795 on Build GRCh38
Transcripts
NM_001080998.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4592336 c.240C>G p.S80R Chr.10: 133626242-133626242 on Build GRCh38
6413346 c.232G>T p.G78W Chr.10: 133626250-133626250 on Build GRCh38
4594184 c.326C>G p.T109R Chr.10: 133625927-133625927 on Build GRCh38
5424389 c.312C>T p.D104D Chr.10: 133625941-133625941 on Build GRCh38
3806823 c.183G>A p.S61S Chr.10: 133626299-133626299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200932273 Chr.10: 133625903-133625903 on Build GRCh38
rs202189720 Chr.10: 133626299-133626299 on Build GRCh38
rs369894857 Chr.10: 133625941-133625941 on Build GRCh38
rs774159950 Chr.10: 133626015-133626015 on Build GRCh38
rs753573385 Chr.10: 133626242-133626242 on Build GRCh38
rs987461933 Chr.10: 133626096-133626096 on Build GRCh38
rs754809295 Chr.10: 133626300-133626300 on Build GRCh38
rs370967667 Chr.10: 133626119-133626119 on Build GRCh38
rs199504252 Chr.10: 133626355-133626355 on Build GRCh38
rs61868732 Chr.10: 133626191-133626191 on Build GRCh38
rs200023384 Chr.10: 133625965-133625965 on Build GRCh38
rs989557370 Chr.10: 133626199-133626199 on Build GRCh38
rs200233832 Chr.10: 133626034-133626034 on Build GRCh38
rs546318039 Chr.10: 133625897-133625897 on Build GRCh38
rs202188827 Chr.10: 133626025-133626025 on Build GRCh38
rs201093409 Chr.10: 133626013-133626013 on Build GRCh38
rs747027346 Chr.10: 133625898-133625898 on Build GRCh38
rs111240927 Chr.10: 133626330-133626330 on Build GRCh38
rs61868730 Chr.10: 133626074-133626074 on Build GRCh38
rs200130584 Chr.10: 133626012-133626012 on Build GRCh38
rs915326231 Chr.10: 133626228-133626228 on Build GRCh38
rs956987992 Chr.10: 133626168-133626168 on Build GRCh38
rs200530375 Chr.10: 133625927-133625927 on Build GRCh38
rs751805370 Chr.10: 133626318-133626318 on Build GRCh38
rs61868731 Chr.10: 133626137-133626137 on Build GRCh38
rs201371219 Chr.10: 133625953-133625953 on Build GRCh38
rs954943168 Chr.10: 133625968-133625968 on Build GRCh38
rs996761044 Chr.10: 133625916-133625916 on Build GRCh38
rs200391649 Chr.10: 133626313-133626313 on Build GRCh38
rs947880983 Chr.10: 133626253-133626253 on Build GRCh38
rs879088694 Chr.10: 133626250-133626250 on Build GRCh38
rs201275211 Chr.10: 133626045-133626045 on Build GRCh38
rs927884763 Chr.10: 133626292-133626292 on Build GRCh38
rs781631189 Chr.10: 133625931-133625931 on Build GRCh38
rs760664927 Chr.10: 133626181-133626181 on Build GRCh38
rs1045305685 Chr.10: 133626265-133626265 on Build GRCh38
rs923968529 Chr.10: 133626160-133626160 on Build GRCh38
rs201387750 Chr.10: 133626325-133626325 on Build GRCh38
rs1029213146 Chr.10: 133625944-133625944 on Build GRCh38
rs796286289 Chr.10: 133626321-133626321 on Build GRCh38

View Full Product Details