Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36227010-36227508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD3
Entrez Gene ID
143458
Gene Name
low density lipoprotein receptor class A domain containing 3
Gene Aliases
LRAD3
UniGene
Hs.745134
Gene Chromosome Location
Chr.11: 35943981-36232136 on Build GRCh38
Transcripts
NM_001304263.1 NM_001304264.1 NM_174902.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4697702 c.735G>T p.Q245H Chr.11: 36227365-36227365 on Build GRCh38
3368356 c.691G>A p.V231I Chr.11: 36227321-36227321 on Build GRCh38
6700323 c.614C>T p.T205M Chr.11: 36227244-36227244 on Build GRCh38
3447029 c.695C>T p.T232I Chr.11: 36227325-36227325 on Build GRCh38
6700322 c.532G>A p.G178S Chr.11: 36227162-36227162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757704221 Chr.11: 36227301-36227301 on Build GRCh38
rs568977599 Chr.11: 36227259-36227259 on Build GRCh38
rs917568676 Chr.11: 36227345-36227345 on Build GRCh38
rs999220610 Chr.11: 36227055-36227055 on Build GRCh38
rs997989453 Chr.11: 36227120-36227120 on Build GRCh38
rs768662503 Chr.11: 36227477-36227477 on Build GRCh38
rs959086054 Chr.11: 36227467-36227467 on Build GRCh38
rs756405388 Chr.11: 36227066-36227066 on Build GRCh38
rs867478663 Chr.11: 36227074-36227074 on Build GRCh38
rs368787104 Chr.11: 36227223-36227223 on Build GRCh38
rs992318799 Chr.11: 36227482-36227482 on Build GRCh38
rs767680853 Chr.11: 36227128-36227129 on Build GRCh38
rs781534942 Chr.11: 36227050-36227050 on Build GRCh38
rs142823958 Chr.11: 36227170-36227170 on Build GRCh38
rs763935519 Chr.11: 36227131-36227131 on Build GRCh38
rs755997476 Chr.11: 36227316-36227317 on Build GRCh38
rs202035397 Chr.11: 36227139-36227139 on Build GRCh38
rs771607204 Chr.11: 36227102-36227102 on Build GRCh38
rs767501890 Chr.11: 36227173-36227173 on Build GRCh38
rs145525386 Chr.11: 36227383-36227383 on Build GRCh38
rs776404499 Chr.11: 36227134-36227134 on Build GRCh38
rs747761677 Chr.11: 36227333-36227333 on Build GRCh38
rs897155198 Chr.11: 36227222-36227222 on Build GRCh38
rs934983721 Chr.11: 36227084-36227084 on Build GRCh38
rs780863881 Chr.11: 36227415-36227415 on Build GRCh38
rs754022854 Chr.11: 36227382-36227382 on Build GRCh38
rs147959076 Chr.11: 36227126-36227126 on Build GRCh38
rs752856483 Chr.11: 36227184-36227184 on Build GRCh38
rs369022315 Chr.11: 36227459-36227459 on Build GRCh38
rs776603047 Chr.11: 36227143-36227143 on Build GRCh38
rs769360706 Chr.11: 36227444-36227444 on Build GRCh38
rs779148159 Chr.11: 36227303-36227303 on Build GRCh38
rs144773833 Chr.11: 36227320-36227320 on Build GRCh38
rs941485441 Chr.11: 36227387-36227387 on Build GRCh38
rs779528487 Chr.11: 36227228-36227228 on Build GRCh38
rs745363307 Chr.11: 36227229-36227229 on Build GRCh38
rs750797339 Chr.11: 36227402-36227402 on Build GRCh38
rs371720202 Chr.11: 36227321-36227321 on Build GRCh38
rs752803367 Chr.11: 36227285-36227285 on Build GRCh38
rs199984146 Chr.11: 36227245-36227245 on Build GRCh38
rs765598827 Chr.11: 36227388-36227388 on Build GRCh38
rs1001731668 Chr.11: 36227268-36227268 on Build GRCh38
rs200303509 Chr.11: 36227331-36227331 on Build GRCh38
rs769576610 Chr.11: 36227365-36227365 on Build GRCh38
rs771541869 Chr.11: 36227234-36227234 on Build GRCh38
rs1018137266 Chr.11: 36227279-36227279 on Build GRCh38
rs1039636652 Chr.11: 36227389-36227389 on Build GRCh38
rs116213177 Chr.11: 36227410-36227410 on Build GRCh38
rs918810030 Chr.11: 36227472-36227472 on Build GRCh38
rs965258527 Chr.11: 36227108-36227108 on Build GRCh38
rs747789364 Chr.11: 36227440-36227440 on Build GRCh38
rs775524868 Chr.11: 36227376-36227376 on Build GRCh38
rs754471821 Chr.11: 36227414-36227414 on Build GRCh38
rs369244883 Chr.11: 36227050-36227052 on Build GRCh38
rs368201136 Chr.11: 36227286-36227286 on Build GRCh38
rs533493135 Chr.11: 36227124-36227124 on Build GRCh38
rs781059733 Chr.11: 36227332-36227332 on Build GRCh38
rs764349508 Chr.11: 36227291-36227291 on Build GRCh38
rs751705397 Chr.11: 36227037-36227037 on Build GRCh38
rs777591884 Chr.11: 36227450-36227450 on Build GRCh38
rs201142444 Chr.11: 36227275-36227275 on Build GRCh38
rs778121415 Chr.11: 36227071-36227071 on Build GRCh38
rs767629424 Chr.11: 36227478-36227478 on Build GRCh38
rs762813324 Chr.11: 36227267-36227267 on Build GRCh38
rs146712517 Chr.11: 36227368-36227368 on Build GRCh38
rs756350036 Chr.11: 36227186-36227186 on Build GRCh38
rs151059364 Chr.11: 36227252-36227252 on Build GRCh38
rs367580930 Chr.11: 36227284-36227284 on Build GRCh38
rs755153900 Chr.11: 36227171-36227171 on Build GRCh38
rs376742134 Chr.11: 36227379-36227379 on Build GRCh38
rs941653541 Chr.11: 36227160-36227160 on Build GRCh38
rs141012443 Chr.11: 36227258-36227258 on Build GRCh38
rs746571027 Chr.11: 36227329-36227329 on Build GRCh38
rs770842001 Chr.11: 36227458-36227458 on Build GRCh38
rs749693649 Chr.11: 36227207-36227207 on Build GRCh38
rs917640699 Chr.11: 36227094-36227094 on Build GRCh38
rs950214056 Chr.11: 36227122-36227122 on Build GRCh38
rs369754473 Chr.11: 36227251-36227251 on Build GRCh38
rs950800771 Chr.11: 36227129-36227129 on Build GRCh38
rs78086369 Chr.11: 36227180-36227180 on Build GRCh38
rs767635829 Chr.11: 36227070-36227070 on Build GRCh38
rs770894941 Chr.11: 36227371-36227371 on Build GRCh38
rs749085601 Chr.11: 36227453-36227453 on Build GRCh38
rs771499207 Chr.11: 36227080-36227080 on Build GRCh38
rs76224006 Chr.11: 36227161-36227161 on Build GRCh38
rs751111119 Chr.11: 36227411-36227411 on Build GRCh38
rs754184080 Chr.11: 36227293-36227293 on Build GRCh38
rs745429372 Chr.11: 36227090-36227090 on Build GRCh38
rs772903246 Chr.11: 36227367-36227367 on Build GRCh38
rs372710536 Chr.11: 36227244-36227244 on Build GRCh38
rs758920048 Chr.11: 36227403-36227403 on Build GRCh38
rs775164207 Chr.11: 36227237-36227237 on Build GRCh38
rs748509604 Chr.11: 36227062-36227062 on Build GRCh38
rs963672509 Chr.11: 36227219-36227219 on Build GRCh38
rs749744878 Chr.11: 36227075-36227075 on Build GRCh38
rs1006843889 Chr.11: 36227096-36227096 on Build GRCh38
rs765269452 Chr.11: 36227162-36227162 on Build GRCh38
rs750570494 Chr.11: 36227214-36227215 on Build GRCh38
rs773702970 Chr.11: 36227089-36227089 on Build GRCh38
rs140347483 Chr.11: 36227380-36227380 on Build GRCh38
rs750896925 Chr.11: 36227304-36227304 on Build GRCh38
rs762163660 Chr.11: 36227104-36227105 on Build GRCh38
rs755134163 Chr.11: 36227047-36227047 on Build GRCh38
rs374256624 Chr.11: 36227101-36227101 on Build GRCh38
rs373306016 Chr.11: 36227473-36227473 on Build GRCh38
rs778070333 Chr.11: 36227198-36227198 on Build GRCh38

View Full Product Details