Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117285790-117286287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACE1
Entrez Gene ID
23621
Gene Name
beta-secretase 1
Gene Aliases
ASP2, BACE, HSPC104
UniGene
Hs.504003
Gene Chromosome Location
Chr.11: 117285686-117316256 on Build GRCh38
Transcripts
NM_001207048.1 NM_001207049.1 NM_012104.4 NM_138971.3 NM_138972.3 NM_138973.3

SNP Detail

SNP ID Chromosome Location
rs771796796 Chr.11: 117285892-117285892 on Build GRCh38
rs961433533 Chr.11: 117286009-117286009 on Build GRCh38
rs906277201 Chr.11: 117286247-117286247 on Build GRCh38
rs565693313 Chr.11: 117286182-117286182 on Build GRCh38
rs561320153 Chr.11: 117285907-117285907 on Build GRCh38
rs7083 Chr.11: 117285836-117285836 on Build GRCh38
rs751950078 Chr.11: 117286085-117286085 on Build GRCh38
rs1053421348 Chr.11: 117285999-117285999 on Build GRCh38
rs141853466 Chr.11: 117286256-117286256 on Build GRCh38
rs146777719 Chr.11: 117286027-117286027 on Build GRCh38
rs943051346 Chr.11: 117285903-117285903 on Build GRCh38
rs551469937 Chr.11: 117286224-117286224 on Build GRCh38
rs1042035402 Chr.11: 117285880-117285880 on Build GRCh38
rs906232510 Chr.11: 117285893-117285893 on Build GRCh38
rs758964120 Chr.11: 117286057-117286057 on Build GRCh38
rs528807974 Chr.11: 117286070-117286070 on Build GRCh38
rs765924282 Chr.11: 117285934-117285934 on Build GRCh38
rs757715251 Chr.11: 117286176-117286176 on Build GRCh38
rs764663717 Chr.11: 117286063-117286063 on Build GRCh38
rs1005533699 Chr.11: 117286178-117286178 on Build GRCh38
rs139297420 Chr.11: 117286204-117286204 on Build GRCh38
rs536505160 Chr.11: 117285825-117285825 on Build GRCh38
rs760359513 Chr.11: 117285928-117285928 on Build GRCh38
rs959569917 Chr.11: 117285925-117285925 on Build GRCh38
rs879032440 Chr.11: 117286163-117286163 on Build GRCh38
rs1009295635 Chr.11: 117285859-117285859 on Build GRCh38
rs753444396 Chr.11: 117285970-117285970 on Build GRCh38
rs1016805268 Chr.11: 117285943-117285943 on Build GRCh38
rs28989517 Chr.11: 117285904-117285904 on Build GRCh38
rs571792562 Chr.11: 117286107-117286107 on Build GRCh38
rs939006613 Chr.11: 117286264-117286264 on Build GRCh38
rs922588061 Chr.11: 117286025-117286025 on Build GRCh38
rs1014467816 Chr.11: 117286023-117286023 on Build GRCh38
rs184140400 Chr.11: 117286140-117286140 on Build GRCh38
rs1047964 Chr.11: 117286177-117286177 on Build GRCh38
rs1030287597 Chr.11: 117286229-117286229 on Build GRCh38
rs544672445 Chr.11: 117286015-117286015 on Build GRCh38
rs756458668 Chr.11: 117286197-117286197 on Build GRCh38
rs775784640 Chr.11: 117286183-117286183 on Build GRCh38

View Full Product Details