Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68303838-68304334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A6OS
Entrez Gene ID
84138
Gene Name
solute carrier family 7 member 6 opposite strand
Gene Aliases
-
UniGene
Hs.729520
Gene Chromosome Location
Chr.16: 68300615-68310965 on Build GRCh38
Transcripts
NM_032178.2 XM_011523372.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5727028 c.487G>A p.V163M Chr.16: 68304217-68304217 on Build GRCh38
5929685 c.672G>A p.W224* Chr.16: 68304032-68304032 on Build GRCh38
972730 c.649G>A p.V217M Chr.16: 68304055-68304055 on Build GRCh38
435575 c.662G>T p.S221I Chr.16: 68304042-68304042 on Build GRCh38
76498 c.587A>G p.Y196C Chr.16: 68304117-68304117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976210250 Chr.16: 68303955-68303955 on Build GRCh38
rs563945083 Chr.16: 68304101-68304101 on Build GRCh38
rs1015148866 Chr.16: 68303943-68303943 on Build GRCh38
rs962617825 Chr.16: 68303947-68303947 on Build GRCh38
rs372590458 Chr.16: 68303993-68303993 on Build GRCh38
rs549790098 Chr.16: 68303882-68303882 on Build GRCh38
rs914520186 Chr.16: 68304303-68304303 on Build GRCh38
rs193289010 Chr.16: 68303939-68303939 on Build GRCh38
rs138428124 Chr.16: 68304055-68304055 on Build GRCh38
rs6499167 Chr.16: 68303895-68303895 on Build GRCh38
rs916771706 Chr.16: 68304244-68304244 on Build GRCh38
rs188304848 Chr.16: 68304056-68304056 on Build GRCh38
rs767583042 Chr.16: 68304149-68304149 on Build GRCh38
rs7200884 Chr.16: 68304004-68304004 on Build GRCh38
rs558919716 Chr.16: 68303958-68303958 on Build GRCh38
rs762583354 Chr.16: 68304279-68304280 on Build GRCh38
rs144208197 Chr.16: 68304003-68304003 on Build GRCh38
rs755124077 Chr.16: 68303945-68303945 on Build GRCh38
rs770051890 Chr.16: 68304144-68304144 on Build GRCh38
rs1046371804 Chr.16: 68304024-68304024 on Build GRCh38
rs767531510 Chr.16: 68304074-68304074 on Build GRCh38
rs202170729 Chr.16: 68304034-68304034 on Build GRCh38
rs768786884 Chr.16: 68304272-68304272 on Build GRCh38
rs747649057 Chr.16: 68304114-68304114 on Build GRCh38
rs751483213 Chr.16: 68303981-68303981 on Build GRCh38
rs149117647 Chr.16: 68304082-68304082 on Build GRCh38
rs376534005 Chr.16: 68304176-68304176 on Build GRCh38
rs543519497 Chr.16: 68304006-68304006 on Build GRCh38
rs377372949 Chr.16: 68304186-68304186 on Build GRCh38
rs746909854 Chr.16: 68304143-68304143 on Build GRCh38
rs143239221 Chr.16: 68304035-68304035 on Build GRCh38
rs778207133 Chr.16: 68304116-68304116 on Build GRCh38
rs935370661 Chr.16: 68304267-68304267 on Build GRCh38
rs34795337 Chr.16: 68304180-68304180 on Build GRCh38
rs764852406 Chr.16: 68304177-68304177 on Build GRCh38
rs370696939 Chr.16: 68304262-68304262 on Build GRCh38
rs757771500 Chr.16: 68304211-68304211 on Build GRCh38
rs139917756 Chr.16: 68304124-68304124 on Build GRCh38
rs753987929 Chr.16: 68303976-68303976 on Build GRCh38
rs983700469 Chr.16: 68304151-68304151 on Build GRCh38
rs149540260 Chr.16: 68304113-68304113 on Build GRCh38
rs1030486489 Chr.16: 68304169-68304169 on Build GRCh38
rs141818719 Chr.16: 68304059-68304059 on Build GRCh38
rs764112242 Chr.16: 68303979-68303979 on Build GRCh38
rs374727375 Chr.16: 68303960-68303960 on Build GRCh38
rs772848364 Chr.16: 68304013-68304023 on Build GRCh38
rs781478250 Chr.16: 68304237-68304237 on Build GRCh38
rs753650859 Chr.16: 68304164-68304164 on Build GRCh38
rs746135718 Chr.16: 68304125-68304125 on Build GRCh38
rs778260157 Chr.16: 68304187-68304187 on Build GRCh38
rs752811984 Chr.16: 68304058-68304058 on Build GRCh38
rs746213685 Chr.16: 68304238-68304238 on Build GRCh38
rs773242217 Chr.16: 68304157-68304157 on Build GRCh38
rs535699453 Chr.16: 68303931-68303931 on Build GRCh38
rs898075161 Chr.16: 68304136-68304136 on Build GRCh38
rs760521229 Chr.16: 68304162-68304162 on Build GRCh38
rs372147827 Chr.16: 68304011-68304011 on Build GRCh38
rs753362393 Chr.16: 68304089-68304089 on Build GRCh38
rs770189571 Chr.16: 68304250-68304250 on Build GRCh38
rs775753104 Chr.16: 68304145-68304145 on Build GRCh38
rs539241288 Chr.16: 68303967-68303967 on Build GRCh38
rs781199487 Chr.16: 68303997-68303997 on Build GRCh38
rs11548855 Chr.16: 68304045-68304045 on Build GRCh38
rs143421140 Chr.16: 68304182-68304182 on Build GRCh38
rs747418833 Chr.16: 68304199-68304199 on Build GRCh38
rs779786318 Chr.16: 68304086-68304086 on Build GRCh38
rs759492044 Chr.16: 68303978-68303978 on Build GRCh38
rs374874080 Chr.16: 68304281-68304281 on Build GRCh38
rs779491490 Chr.16: 68304029-68304029 on Build GRCh38
rs766485012 Chr.16: 68303975-68303975 on Build GRCh38
rs771210829 Chr.16: 68304050-68304050 on Build GRCh38
rs372518649 Chr.16: 68304118-68304118 on Build GRCh38
rs568032665 Chr.16: 68304292-68304292 on Build GRCh38
rs754274503 Chr.16: 68303941-68303941 on Build GRCh38
rs768005272 Chr.16: 68304017-68304017 on Build GRCh38
rs958363610 Chr.16: 68304230-68304230 on Build GRCh38
rs1001224297 Chr.16: 68304062-68304062 on Build GRCh38
rs750291277 Chr.16: 68304080-68304080 on Build GRCh38
rs761308489 Chr.16: 68303861-68303861 on Build GRCh38
rs970810598 Chr.16: 68304258-68304258 on Build GRCh38
rs1017295588 Chr.16: 68304105-68304105 on Build GRCh38
rs766026962 Chr.16: 68304163-68304163 on Build GRCh38
rs549904198 Chr.16: 68304263-68304263 on Build GRCh38
rs139184366 Chr.16: 68304090-68304090 on Build GRCh38
rs760583793 Chr.16: 68304282-68304282 on Build GRCh38
rs907076824 Chr.16: 68304057-68304057 on Build GRCh38
rs146039260 Chr.16: 68304123-68304123 on Build GRCh38
rs531477472 Chr.16: 68303871-68303871 on Build GRCh38

View Full Product Details