Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36887736-36888257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF345
Entrez Gene ID
25850
Gene Name
zinc finger protein 345
Gene Aliases
HZF10
UniGene
Hs.362324
Gene Chromosome Location
Chr.19: 36850350-36893218 on Build GRCh38
Transcripts
NR_038362.1 NM_001037232.3 NM_001171979.1 XM_005258876.4 XM_011526933.2

Gene Symbol
ZNF829
Entrez Gene ID
374899
Gene Name
zinc finger protein 829
Gene Aliases
-
UniGene
Hs.720312
Gene Chromosome Location
Chr.19: 36888124-36916387 on Build GRCh38
Transcripts
NR_038362.1 NM_001037232.3 NM_001171979.1 XM_005258876.4 XM_011526933.2

SNP Detail

SNP ID Chromosome Location
rs968428575 Chr.19: 36887982-36887982 on Build GRCh38
rs752003250 Chr.19: 36888116-36888118 on Build GRCh38
rs192478081 Chr.19: 36887877-36887877 on Build GRCh38
rs979860838 Chr.19: 36888125-36888125 on Build GRCh38
rs16422 Chr.19: 36888202-36888207 on Build GRCh38
rs1020006590 Chr.19: 36888057-36888057 on Build GRCh38
rs781709381 Chr.19: 36888131-36888132 on Build GRCh38
rs150448433 Chr.19: 36887866-36887866 on Build GRCh38
rs184774215 Chr.19: 36888012-36888012 on Build GRCh38
rs758985500 Chr.19: 36887921-36887921 on Build GRCh38
rs868675054 Chr.19: 36887768-36887768 on Build GRCh38
rs1028409540 Chr.19: 36887980-36887980 on Build GRCh38
rs547285707 Chr.19: 36888211-36888211 on Build GRCh38
rs71754010 Chr.19: 36888210-36888210 on Build GRCh38
rs979437978 Chr.19: 36887993-36887993 on Build GRCh38
rs904898005 Chr.19: 36888176-36888176 on Build GRCh38
rs989094081 Chr.19: 36888070-36888070 on Build GRCh38
rs905569223 Chr.19: 36887791-36887791 on Build GRCh38
rs764385982 Chr.19: 36887961-36887961 on Build GRCh38
rs1017036418 Chr.19: 36887948-36887948 on Build GRCh38
rs947139568 Chr.19: 36887779-36887779 on Build GRCh38
rs914416661 Chr.19: 36888074-36888074 on Build GRCh38
rs569659070 Chr.19: 36887784-36887784 on Build GRCh38
rs887013791 Chr.19: 36888136-36888136 on Build GRCh38
rs988315980 Chr.19: 36887968-36887968 on Build GRCh38
rs910654576 Chr.19: 36887805-36887805 on Build GRCh38
rs986807591 Chr.19: 36888038-36888038 on Build GRCh38
rs1035151158 Chr.19: 36887963-36887963 on Build GRCh38
rs1038034577 Chr.19: 36888160-36888160 on Build GRCh38
rs1047021382 Chr.19: 36887932-36887932 on Build GRCh38
rs78323407 Chr.19: 36887967-36887967 on Build GRCh38
rs770085089 Chr.19: 36888069-36888069 on Build GRCh38
rs563757178 Chr.19: 36888055-36888055 on Build GRCh38
rs886747484 Chr.19: 36887938-36887939 on Build GRCh38
rs954052942 Chr.19: 36888010-36888010 on Build GRCh38
rs826285 Chr.19: 36888130-36888130 on Build GRCh38
rs947211915 Chr.19: 36888098-36888098 on Build GRCh38
rs10405878 Chr.19: 36887923-36887923 on Build GRCh38
rs537049524 Chr.19: 36887859-36887859 on Build GRCh38
rs189544679 Chr.19: 36888156-36888156 on Build GRCh38
rs112416579 Chr.19: 36887958-36887958 on Build GRCh38
rs575694530 Chr.19: 36888122-36888122 on Build GRCh38
rs1004841946 Chr.19: 36887925-36887925 on Build GRCh38
rs907493491 Chr.19: 36888221-36888221 on Build GRCh38
rs28416986 Chr.19: 36887956-36887956 on Build GRCh38
rs914364589 Chr.19: 36887761-36887761 on Build GRCh38
rs1015767020 Chr.19: 36887944-36887944 on Build GRCh38
rs938377823 Chr.19: 36888139-36888139 on Build GRCh38
rs564355496 Chr.19: 36888146-36888146 on Build GRCh38

View Full Product Details