Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 109398456-109399001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
XM_017004623.1 NM_001099289.2 XM_011511109.2 XM_011511110.2

Gene Symbol
SH3RF3
Entrez Gene ID
344558
Gene Name
SH3 domain containing ring finger 3
Gene Aliases
POSH2, SH3MD4
UniGene
Hs.171244
Gene Chromosome Location
Chr.2: 109129439-109504636 on Build GRCh38
Transcripts
XM_017004623.1 NM_001099289.2 XM_011511109.2 XM_011511110.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6031712 c.1106G>A p.R369H Chr.2: 109398750-109398750 on Build GRCh38
6777379 c.957C>T p.S319S Chr.2: 109398601-109398601 on Build GRCh38
6777380 c.1210A>G p.S404G Chr.2: 109398854-109398854 on Build GRCh38
3379731 c.1089C>G p.V363V Chr.2: 109398733-109398733 on Build GRCh38
4084132 c.1152G>A p.A384A Chr.2: 109398796-109398796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201743885 Chr.2: 109398601-109398601 on Build GRCh38
rs778176714 Chr.2: 109398888-109398888 on Build GRCh38
rs774770044 Chr.2: 109398656-109398656 on Build GRCh38
rs372778850 Chr.2: 109398769-109398769 on Build GRCh38
rs370742737 Chr.2: 109398934-109398934 on Build GRCh38
rs771925192 Chr.2: 109398937-109398937 on Build GRCh38
rs1056107996 Chr.2: 109398524-109398524 on Build GRCh38
rs779511360 Chr.2: 109398776-109398776 on Build GRCh38
rs56728531 Chr.2: 109398523-109398523 on Build GRCh38
rs990750662 Chr.2: 109398635-109398635 on Build GRCh38
rs775419454 Chr.2: 109398808-109398808 on Build GRCh38
rs1034820696 Chr.2: 109398872-109398872 on Build GRCh38
rs779806232 Chr.2: 109398724-109398724 on Build GRCh38
rs1047066921 Chr.2: 109398780-109398780 on Build GRCh38
rs369452400 Chr.2: 109398668-109398668 on Build GRCh38
rs529655275 Chr.2: 109398756-109398756 on Build GRCh38
rs371341519 Chr.2: 109398542-109398542 on Build GRCh38
rs752556304 Chr.2: 109398615-109398615 on Build GRCh38
rs191354318 Chr.2: 109398570-109398570 on Build GRCh38
rs752070631 Chr.2: 109398973-109398973 on Build GRCh38
rs773536071 Chr.2: 109398663-109398663 on Build GRCh38
rs764770907 Chr.2: 109398969-109398969 on Build GRCh38
rs765935823 Chr.2: 109398878-109398878 on Build GRCh38
rs369969380 Chr.2: 109398693-109398693 on Build GRCh38
rs765952849 Chr.2: 109398956-109398956 on Build GRCh38
rs746617748 Chr.2: 109398641-109398641 on Build GRCh38
rs780675276 Chr.2: 109398642-109398642 on Build GRCh38
rs200172316 Chr.2: 109398893-109398893 on Build GRCh38
rs752631837 Chr.2: 109398681-109398681 on Build GRCh38
rs772982157 Chr.2: 109398954-109398954 on Build GRCh38
rs761616312 Chr.2: 109398844-109398844 on Build GRCh38
rs756843444 Chr.2: 109398690-109398690 on Build GRCh38
rs563047360 Chr.2: 109398801-109398801 on Build GRCh38
rs767185837 Chr.2: 109398768-109398768 on Build GRCh38
rs377256148 Chr.2: 109398911-109398911 on Build GRCh38
rs756503732 Chr.2: 109398978-109398978 on Build GRCh38
rs117154967 Chr.2: 109398682-109398682 on Build GRCh38
rs770064902 Chr.2: 109398807-109398807 on Build GRCh38
rs753301166 Chr.2: 109398880-109398880 on Build GRCh38
rs534497883 Chr.2: 109398955-109398955 on Build GRCh38
rs754445900 Chr.2: 109398881-109398881 on Build GRCh38
rs375333092 Chr.2: 109398953-109398953 on Build GRCh38
rs769161547 Chr.2: 109398654-109398654 on Build GRCh38
rs762105450 Chr.2: 109398578-109398578 on Build GRCh38
rs775607861 Chr.2: 109398750-109398750 on Build GRCh38
rs768024032 Chr.2: 109398955-109398956 on Build GRCh38
rs967951867 Chr.2: 109398670-109398670 on Build GRCh38
rs371355051 Chr.2: 109398795-109398795 on Build GRCh38
rs746149562 Chr.2: 109398894-109398894 on Build GRCh38
rs993194064 Chr.2: 109398576-109398576 on Build GRCh38
rs751321197 Chr.2: 109398766-109398766 on Build GRCh38
rs371934571 Chr.2: 109398679-109398679 on Build GRCh38
rs923750802 Chr.2: 109398487-109398487 on Build GRCh38
rs373779127 Chr.2: 109398645-109398645 on Build GRCh38
rs200752997 Chr.2: 109398581-109398581 on Build GRCh38
rs10181644 Chr.2: 109398853-109398853 on Build GRCh38
rs1000837461 Chr.2: 109398962-109398962 on Build GRCh38
rs925756878 Chr.2: 109398522-109398522 on Build GRCh38
rs780251621 Chr.2: 109398897-109398897 on Build GRCh38
rs573351208 Chr.2: 109398729-109398729 on Build GRCh38
rs889911365 Chr.2: 109398514-109398514 on Build GRCh38
rs765730743 Chr.2: 109398903-109398903 on Build GRCh38
rs770154489 Chr.2: 109398895-109398895 on Build GRCh38
rs746724924 Chr.2: 109398761-109398763 on Build GRCh38
rs745383521 Chr.2: 109398694-109398694 on Build GRCh38
rs200518309 Chr.2: 109398746-109398746 on Build GRCh38
rs542535234 Chr.2: 109398730-109398730 on Build GRCh38
rs779421234 Chr.2: 109398701-109398701 on Build GRCh38
rs949973131 Chr.2: 109398566-109398566 on Build GRCh38
rs765272189 Chr.2: 109398680-109398680 on Build GRCh38
rs748580626 Chr.2: 109398658-109398658 on Build GRCh38
rs755673043 Chr.2: 109398772-109398772 on Build GRCh38
rs760318732 Chr.2: 109398873-109398873 on Build GRCh38
rs376274165 Chr.2: 109398749-109398749 on Build GRCh38
rs757825576 Chr.2: 109398974-109398974 on Build GRCh38
rs748583936 Chr.2: 109398717-109398717 on Build GRCh38
rs538975074 Chr.2: 109398631-109398631 on Build GRCh38
rs369328436 Chr.2: 109398757-109398757 on Build GRCh38
rs201884932 Chr.2: 109398767-109398767 on Build GRCh38
rs774302778 Chr.2: 109398822-109398822 on Build GRCh38
rs760780861 Chr.2: 109398664-109398664 on Build GRCh38
rs375829146 Chr.2: 109398794-109398794 on Build GRCh38
rs767781610 Chr.2: 109398579-109398579 on Build GRCh38
rs767279921 Chr.2: 109398845-109398845 on Build GRCh38
rs776308242 Chr.2: 109398958-109398958 on Build GRCh38
rs776837938 Chr.2: 109398675-109398675 on Build GRCh38
rs768730435 Chr.2: 109398924-109398924 on Build GRCh38
rs565845026 Chr.2: 109398544-109398544 on Build GRCh38
rs755578903 Chr.2: 109398699-109398699 on Build GRCh38
rs367945117 Chr.2: 109398659-109398659 on Build GRCh38
rs774206163 Chr.2: 109398928-109398928 on Build GRCh38
rs753080009 Chr.2: 109398925-109398925 on Build GRCh38
rs763027417 Chr.2: 109398813-109398813 on Build GRCh38
rs753317886 Chr.2: 109398779-109398779 on Build GRCh38
rs569376236 Chr.2: 109398509-109398509 on Build GRCh38
rs376365695 Chr.2: 109398961-109398961 on Build GRCh38
rs759723090 Chr.2: 109398747-109398747 on Build GRCh38
rs915746623 Chr.2: 109398781-109398781 on Build GRCh38
rs372845928 Chr.2: 109398892-109398892 on Build GRCh38
rs866647398 Chr.2: 109398638-109398638 on Build GRCh38
rs537361444 Chr.2: 109398640-109398640 on Build GRCh38
rs559859361 Chr.2: 109398683-109398683 on Build GRCh38
rs765182236 Chr.2: 109398614-109398614 on Build GRCh38
rs532037049 Chr.2: 109398819-109398819 on Build GRCh38
rs11885439 Chr.2: 109398592-109398592 on Build GRCh38
rs762794288 Chr.2: 109398755-109398755 on Build GRCh38
rs771293090 Chr.2: 109398735-109398735 on Build GRCh38
rs942451279 Chr.2: 109398971-109398971 on Build GRCh38
rs377178053 Chr.2: 109398793-109398793 on Build GRCh38
rs925422030 Chr.2: 109398762-109398762 on Build GRCh38
rs865882313 Chr.2: 109398535-109398535 on Build GRCh38
rs1003337474 Chr.2: 109398598-109398598 on Build GRCh38
rs371087893 Chr.2: 109398976-109398976 on Build GRCh38
rs11884441 Chr.2: 109398504-109398504 on Build GRCh38

View Full Product Details