Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1439833-1440339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICALL2
Entrez Gene ID
79778
Gene Name
MICAL like 2
Gene Aliases
JRAB, MICAL-L2
UniGene
Hs.376617
Gene Chromosome Location
Chr.7: 1433639-1459497 on Build GRCh38
Transcripts
NM_182924.3 XM_011515520.2 XM_011515522.2 XM_011515523.2 XM_011515524.2 XM_017012619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7377154 c.1826C>T p.P609L Chr.7: 1440065-1440065 on Build GRCh38
1743786 c.1806-3C>A p.? Chr.7: 1440088-1440088 on Build GRCh38
3381847 c.1904C>T p.T635I Chr.7: 1439987-1439987 on Build GRCh38
1755147 c.1819A>T p.K607* Chr.7: 1440072-1440072 on Build GRCh38
5630180 c.1958G>C p.S653T Chr.7: 1439933-1439933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760890891 Chr.7: 1439905-1439905 on Build GRCh38
rs59904195 Chr.7: 1440316-1440316 on Build GRCh38
rs745628121 Chr.7: 1440266-1440266 on Build GRCh38
rs771147634 Chr.7: 1440190-1440190 on Build GRCh38
rs755140922 Chr.7: 1439970-1439970 on Build GRCh38
rs765012800 Chr.7: 1440027-1440028 on Build GRCh38
rs570236129 Chr.7: 1440179-1440179 on Build GRCh38
rs764535712 Chr.7: 1439941-1439941 on Build GRCh38
rs969640833 Chr.7: 1440301-1440301 on Build GRCh38
rs562017552 Chr.7: 1440308-1440308 on Build GRCh38
rs974250296 Chr.7: 1440148-1440148 on Build GRCh38
rs374682289 Chr.7: 1439981-1439981 on Build GRCh38
rs111747488 Chr.7: 1440253-1440253 on Build GRCh38
rs755636147 Chr.7: 1439856-1439856 on Build GRCh38
rs775142226 Chr.7: 1439993-1439993 on Build GRCh38
rs750325450 Chr.7: 1440110-1440110 on Build GRCh38
rs771580877 Chr.7: 1440132-1440132 on Build GRCh38
rs912816916 Chr.7: 1440192-1440192 on Build GRCh38
rs375763670 Chr.7: 1440118-1440118 on Build GRCh38
rs754083003 Chr.7: 1440037-1440037 on Build GRCh38
rs780920605 Chr.7: 1440165-1440165 on Build GRCh38
rs746901894 Chr.7: 1439978-1439978 on Build GRCh38
rs999257159 Chr.7: 1440281-1440281 on Build GRCh38
rs994858514 Chr.7: 1439889-1439889 on Build GRCh38
rs76110611 Chr.7: 1439977-1439977 on Build GRCh38
rs267601383 Chr.7: 1439944-1439944 on Build GRCh38
rs777169182 Chr.7: 1439912-1439912 on Build GRCh38
rs61287564 Chr.7: 1440023-1440023 on Build GRCh38
rs761305244 Chr.7: 1440068-1440068 on Build GRCh38
rs1033263378 Chr.7: 1440035-1440035 on Build GRCh38
rs748154385 Chr.7: 1439855-1439855 on Build GRCh38
rs59814665 Chr.7: 1440129-1440129 on Build GRCh38
rs758331489 Chr.7: 1440119-1440120 on Build GRCh38
rs967972263 Chr.7: 1439938-1439938 on Build GRCh38
rs749333078 Chr.7: 1439969-1439969 on Build GRCh38
rs61504673 Chr.7: 1439872-1439872 on Build GRCh38
rs990140847 Chr.7: 1439914-1439914 on Build GRCh38
rs752489290 Chr.7: 1440047-1440047 on Build GRCh38
rs774912933 Chr.7: 1439937-1439937 on Build GRCh38
rs369571833 Chr.7: 1439880-1439880 on Build GRCh38
rs770187244 Chr.7: 1439898-1439898 on Build GRCh38
rs143197912 Chr.7: 1440047-1440047 on Build GRCh38
rs371243869 Chr.7: 1440114-1440114 on Build GRCh38
rs1039877040 Chr.7: 1440252-1440252 on Build GRCh38
rs200214323 Chr.7: 1440010-1440010 on Build GRCh38
rs998269509 Chr.7: 1440181-1440181 on Build GRCh38
rs757818737 Chr.7: 1439947-1439947 on Build GRCh38
rs544240623 Chr.7: 1440317-1440317 on Build GRCh38
rs1039704701 Chr.7: 1440169-1440169 on Build GRCh38
rs1042219107 Chr.7: 1440189-1440189 on Build GRCh38
rs201278821 Chr.7: 1439948-1439948 on Build GRCh38
rs368637404 Chr.7: 1440113-1440113 on Build GRCh38
rs370374080 Chr.7: 1440065-1440065 on Build GRCh38
rs769375989 Chr.7: 1439988-1439988 on Build GRCh38
rs372614472 Chr.7: 1440083-1440083 on Build GRCh38
rs984585331 Chr.7: 1440290-1440290 on Build GRCh38
rs756571274 Chr.7: 1440032-1440032 on Build GRCh38
rs759676182 Chr.7: 1440090-1440090 on Build GRCh38
rs376219576 Chr.7: 1439942-1439942 on Build GRCh38
rs796122863 Chr.7: 1440131-1440131 on Build GRCh38
rs762405879 Chr.7: 1439999-1439999 on Build GRCh38
rs770601361 Chr.7: 1439925-1439925 on Build GRCh38
rs139843807 Chr.7: 1440307-1440307 on Build GRCh38
rs758524433 Chr.7: 1440106-1440106 on Build GRCh38
rs768411833 Chr.7: 1440061-1440061 on Build GRCh38
rs112889477 Chr.7: 1440006-1440006 on Build GRCh38
rs772251446 Chr.7: 1439857-1439857 on Build GRCh38
rs1013481375 Chr.7: 1440298-1440298 on Build GRCh38
rs751392623 Chr.7: 1440107-1440107 on Build GRCh38
rs372262317 Chr.7: 1440176-1440176 on Build GRCh38
rs115115094 Chr.7: 1440276-1440276 on Build GRCh38
rs1023944314 Chr.7: 1440277-1440277 on Build GRCh38
rs557099335 Chr.7: 1440213-1440213 on Build GRCh38
rs756092602 Chr.7: 1440111-1440111 on Build GRCh38
rs750697437 Chr.7: 1440028-1440028 on Build GRCh38
rs762462940 Chr.7: 1439940-1439940 on Build GRCh38
rs756397828 Chr.7: 1439954-1439954 on Build GRCh38
rs1033147266 Chr.7: 1440261-1440261 on Build GRCh38
rs745665032 Chr.7: 1439983-1439983 on Build GRCh38
rs544291207 Chr.7: 1439896-1439896 on Build GRCh38
rs919573313 Chr.7: 1440156-1440156 on Build GRCh38
rs370534746 Chr.7: 1439953-1439953 on Build GRCh38
rs765504408 Chr.7: 1440092-1440092 on Build GRCh38
rs552872571 Chr.7: 1440283-1440283 on Build GRCh38
rs755304550 Chr.7: 1440038-1440038 on Build GRCh38
rs1025795647 Chr.7: 1439892-1439892 on Build GRCh38
rs535770399 Chr.7: 1440143-1440143 on Build GRCh38
rs944299614 Chr.7: 1440236-1440236 on Build GRCh38
rs181284121 Chr.7: 1440040-1440040 on Build GRCh38
rs779327645 Chr.7: 1439901-1439901 on Build GRCh38
rs754316219 Chr.7: 1440089-1440089 on Build GRCh38
rs778253235 Chr.7: 1440046-1440046 on Build GRCh38
rs373257576 Chr.7: 1439927-1439927 on Build GRCh38
rs377474066 Chr.7: 1440299-1440299 on Build GRCh38
rs760019051 Chr.7: 1439955-1439955 on Build GRCh38
rs758899963 Chr.7: 1439885-1439885 on Build GRCh38
rs750584606 Chr.7: 1439900-1439901 on Build GRCh38
rs972235273 Chr.7: 1440072-1440072 on Build GRCh38
rs748916625 Chr.7: 1440060-1440060 on Build GRCh38
rs761247110 Chr.7: 1439895-1439895 on Build GRCh38
rs199983192 Chr.7: 1440063-1440063 on Build GRCh38
rs201339113 Chr.7: 1440062-1440062 on Build GRCh38
rs752263690 Chr.7: 1439874-1439874 on Build GRCh38
rs779094059 Chr.7: 1440282-1440282 on Build GRCh38
rs919675316 Chr.7: 1440315-1440315 on Build GRCh38
rs377091059 Chr.7: 1440149-1440149 on Build GRCh38
rs766832708 Chr.7: 1440080-1440080 on Build GRCh38
rs773050309 Chr.7: 1439913-1439913 on Build GRCh38
rs749386444 Chr.7: 1439907-1439907 on Build GRCh38
rs369441287 Chr.7: 1440173-1440173 on Build GRCh38
rs1043539610 Chr.7: 1440265-1440265 on Build GRCh38
rs747572149 Chr.7: 1440127-1440127 on Build GRCh38
rs188093849 Chr.7: 1440033-1440033 on Build GRCh38
rs757844706 Chr.7: 1440056-1440056 on Build GRCh38
rs903350426 Chr.7: 1440247-1440247 on Build GRCh38
rs951849123 Chr.7: 1440289-1440289 on Build GRCh38
rs764938392 Chr.7: 1439936-1439936 on Build GRCh38
rs916326696 Chr.7: 1439971-1439971 on Build GRCh38
rs766801281 Chr.7: 1439906-1439906 on Build GRCh38
rs112998114 Chr.7: 1439964-1439964 on Build GRCh38
rs1047776502 Chr.7: 1440031-1440031 on Build GRCh38
rs557804391 Chr.7: 1440228-1440228 on Build GRCh38
rs759177756 Chr.7: 1439929-1439929 on Build GRCh38
rs745609983 Chr.7: 1440043-1440043 on Build GRCh38
rs1049035786 Chr.7: 1439960-1439960 on Build GRCh38
rs1047271718 Chr.7: 1440184-1440184 on Build GRCh38
rs555453972 Chr.7: 1439858-1439858 on Build GRCh38
rs183766085 Chr.7: 1439919-1439919 on Build GRCh38
rs752659144 Chr.7: 1440105-1440105 on Build GRCh38
rs991197846 Chr.7: 1439877-1439877 on Build GRCh38
rs375638403 Chr.7: 1440041-1440041 on Build GRCh38
rs1011632822 Chr.7: 1440257-1440257 on Build GRCh38
rs898742278 Chr.7: 1439873-1439873 on Build GRCh38
rs960600785 Chr.7: 1440294-1440294 on Build GRCh38

View Full Product Details