Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 9030074-9030591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERI1
Entrez Gene ID
90459
Gene Name
exoribonuclease 1
Gene Aliases
3'HEXO, HEXO, THEX1
UniGene
Hs.20000
Gene Chromosome Location
Chr.8: 8892225-9100262 on Build GRCh38
Transcripts
NM_153332.3 XM_005272401.2 XM_011543848.2 XM_011543849.1 XM_011543850.2 XM_011543851.1 XM_017013949.1 XM_017013950.1 XM_017013951.1 XM_017013952.1 XM_017013953.1 XM_017013954.1 XM_017013955.1

SNP Detail

SNP ID Chromosome Location
rs190213778 Chr.8: 9030350-9030350 on Build GRCh38
rs1029355060 Chr.8: 9030451-9030451 on Build GRCh38
rs1053787843 Chr.8: 9030172-9030172 on Build GRCh38
rs1047707968 Chr.8: 9030205-9030205 on Build GRCh38
rs989531996 Chr.8: 9030288-9030288 on Build GRCh38
rs888186790 Chr.8: 9030215-9030215 on Build GRCh38
rs762573854 Chr.8: 9030466-9030466 on Build GRCh38
rs1023384639 Chr.8: 9030336-9030336 on Build GRCh38
rs930245915 Chr.8: 9030405-9030405 on Build GRCh38
rs114209463 Chr.8: 9030545-9030545 on Build GRCh38
rs775009059 Chr.8: 9030158-9030159 on Build GRCh38
rs769732423 Chr.8: 9030255-9030255 on Build GRCh38
rs778054140 Chr.8: 9030306-9030306 on Build GRCh38
rs868076876 Chr.8: 9030250-9030250 on Build GRCh38
rs1023794740 Chr.8: 9030181-9030181 on Build GRCh38
rs550685702 Chr.8: 9030419-9030419 on Build GRCh38
rs565344197 Chr.8: 9030529-9030529 on Build GRCh38
rs1005269076 Chr.8: 9030424-9030424 on Build GRCh38
rs967726646 Chr.8: 9030246-9030246 on Build GRCh38
rs1039477005 Chr.8: 9030439-9030439 on Build GRCh38
rs796906568 Chr.8: 9030245-9030245 on Build GRCh38
rs960176567 Chr.8: 9030370-9030370 on Build GRCh38
rs771343556 Chr.8: 9030364-9030364 on Build GRCh38
rs1032810705 Chr.8: 9030506-9030506 on Build GRCh38
rs552910546 Chr.8: 9030187-9030187 on Build GRCh38
rs922397489 Chr.8: 9030131-9030131 on Build GRCh38
rs997835897 Chr.8: 9030441-9030441 on Build GRCh38
rs936772611 Chr.8: 9030169-9030169 on Build GRCh38
rs76165386 Chr.8: 9030109-9030109 on Build GRCh38
rs911344427 Chr.8: 9030436-9030436 on Build GRCh38
rs370741190 Chr.8: 9030528-9030531 on Build GRCh38
rs540233991 Chr.8: 9030449-9030449 on Build GRCh38
rs999199066 Chr.8: 9030161-9030166 on Build GRCh38
rs200010608 Chr.8: 9030365-9030365 on Build GRCh38
rs976702262 Chr.8: 9030130-9030130 on Build GRCh38
rs989499854 Chr.8: 9030515-9030515 on Build GRCh38
rs957434957 Chr.8: 9030507-9030507 on Build GRCh38
rs1017948113 Chr.8: 9030222-9030222 on Build GRCh38
rs945916615 Chr.8: 9030142-9030142 on Build GRCh38
rs566628384 Chr.8: 9030218-9030218 on Build GRCh38
rs149209082 Chr.8: 9030546-9030546 on Build GRCh38
rs770550082 Chr.8: 9030214-9030214 on Build GRCh38
rs564533646 Chr.8: 9030435-9030435 on Build GRCh38
rs911258860 Chr.8: 9030240-9030240 on Build GRCh38
rs775385164 Chr.8: 9030313-9030313 on Build GRCh38
rs575832868 Chr.8: 9030345-9030345 on Build GRCh38
rs375315959 Chr.8: 9030175-9030175 on Build GRCh38
rs960558892 Chr.8: 9030204-9030204 on Build GRCh38
rs749821457 Chr.8: 9030149-9030149 on Build GRCh38
rs763023692 Chr.8: 9030457-9030457 on Build GRCh38
rs145398773 Chr.8: 9030414-9030414 on Build GRCh38
rs893487950 Chr.8: 9030282-9030282 on Build GRCh38
rs776344254 Chr.8: 9030235-9030235 on Build GRCh38
rs924034585 Chr.8: 9030161-9030161 on Build GRCh38
rs555267048 Chr.8: 9030384-9030384 on Build GRCh38
rs899468590 Chr.8: 9030440-9030440 on Build GRCh38
rs994360467 Chr.8: 9030193-9030193 on Build GRCh38
rs78090829 Chr.8: 9030112-9030112 on Build GRCh38
rs958139766 Chr.8: 9030278-9030278 on Build GRCh38
rs150068853 Chr.8: 9030249-9030249 on Build GRCh38
rs2953807 Chr.8: 9030178-9030178 on Build GRCh38
rs984178819 Chr.8: 9030232-9030232 on Build GRCh38
rs1029595863 Chr.8: 9030243-9030243 on Build GRCh38
rs183629220 Chr.8: 9030182-9030182 on Build GRCh38
rs555287382 Chr.8: 9030275-9030275 on Build GRCh38
rs115377185 Chr.8: 9030327-9030327 on Build GRCh38
rs1041005308 Chr.8: 9030110-9030110 on Build GRCh38
rs747489262 Chr.8: 9030309-9030309 on Build GRCh38
rs751644609 Chr.8: 9030562-9030562 on Build GRCh38
rs1035951339 Chr.8: 9030190-9030190 on Build GRCh38
rs922445518 Chr.8: 9030514-9030514 on Build GRCh38
rs931286832 Chr.8: 9030223-9030223 on Build GRCh38
rs998066251 Chr.8: 9030242-9030242 on Build GRCh38
rs548490956 Chr.8: 9030153-9030153 on Build GRCh38
rs928225497 Chr.8: 9030396-9030396 on Build GRCh38
rs1047225773 Chr.8: 9030408-9030408 on Build GRCh38
rs775098574 Chr.8: 9030422-9030422 on Build GRCh38
rs948272064 Chr.8: 9030180-9030180 on Build GRCh38
rs972974143 Chr.8: 9030381-9030381 on Build GRCh38
rs942588169 Chr.8: 9030241-9030241 on Build GRCh38
rs982061297 Chr.8: 9030561-9030561 on Build GRCh38
rs113462617 Chr.8: 9030461-9030461 on Build GRCh38
rs1036252528 Chr.8: 9030363-9030363 on Build GRCh38

View Full Product Details