Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229452447-229452950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP133
Entrez Gene ID
55746
Gene Name
nucleoporin 133
Gene Aliases
hNUP133
UniGene
Hs.12457
Gene Chromosome Location
Chr.1: 229440995-229508343 on Build GRCh38
Transcripts
NM_018230.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
283429 c.2981-1G>T p.? Chr.1: 229452644-229452644 on Build GRCh38
1997466 c.3087A>G p.P1029P Chr.1: 229452537-229452537 on Build GRCh38
7316745 c.2989G>A p.E997K Chr.1: 229452635-229452635 on Build GRCh38
679179 c.2999G>T p.R1000L Chr.1: 229452625-229452625 on Build GRCh38
6209002 c.3098G>A p.G1033D Chr.1: 229452526-229452526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961059661 Chr.1: 229452740-229452740 on Build GRCh38
rs952809653 Chr.1: 229452570-229452570 on Build GRCh38
rs890906490 Chr.1: 229452906-229452906 on Build GRCh38
rs1008352252 Chr.1: 229452724-229452724 on Build GRCh38
rs557870934 Chr.1: 229452662-229452662 on Build GRCh38
rs967605662 Chr.1: 229452903-229452903 on Build GRCh38
rs992681296 Chr.1: 229452873-229452873 on Build GRCh38
rs144837509 Chr.1: 229452582-229452582 on Build GRCh38
rs529080246 Chr.1: 229452484-229452484 on Build GRCh38
rs751402996 Chr.1: 229452559-229452559 on Build GRCh38
rs913157076 Chr.1: 229452602-229452602 on Build GRCh38
rs978728334 Chr.1: 229452910-229452910 on Build GRCh38
rs371151940 Chr.1: 229452500-229452500 on Build GRCh38
rs771153151 Chr.1: 229452603-229452603 on Build GRCh38
rs932356959 Chr.1: 229452880-229452880 on Build GRCh38
rs1034442863 Chr.1: 229452839-229452839 on Build GRCh38
rs771289756 Chr.1: 229452494-229452494 on Build GRCh38
rs926768921 Chr.1: 229452700-229452700 on Build GRCh38
rs759109247 Chr.1: 229452517-229452517 on Build GRCh38
rs183776587 Chr.1: 229452477-229452477 on Build GRCh38
rs374502193 Chr.1: 229452579-229452579 on Build GRCh38
rs574368417 Chr.1: 229452817-229452817 on Build GRCh38
rs537919966 Chr.1: 229452585-229452585 on Build GRCh38
rs367562007 Chr.1: 229452552-229452552 on Build GRCh38
rs762714822 Chr.1: 229452682-229452682 on Build GRCh38
rs960073777 Chr.1: 229452846-229452846 on Build GRCh38
rs35550140 Chr.1: 229452526-229452526 on Build GRCh38
rs746331640 Chr.1: 229452625-229452625 on Build GRCh38
rs774831705 Chr.1: 229452611-229452611 on Build GRCh38
rs1008340934 Chr.1: 229452502-229452502 on Build GRCh38
rs188115342 Chr.1: 229452778-229452778 on Build GRCh38
rs931635127 Chr.1: 229452663-229452663 on Build GRCh38
rs764840695 Chr.1: 229452666-229452666 on Build GRCh38
rs79575284 Chr.1: 229452481-229452481 on Build GRCh38
rs530370592 Chr.1: 229452815-229452815 on Build GRCh38
rs370430320 Chr.1: 229452746-229452746 on Build GRCh38
rs959896358 Chr.1: 229452588-229452588 on Build GRCh38
rs754959122 Chr.1: 229452622-229452622 on Build GRCh38
rs749544779 Chr.1: 229452599-229452599 on Build GRCh38
rs765125062 Chr.1: 229452832-229452832 on Build GRCh38
rs945964129 Chr.1: 229452617-229452617 on Build GRCh38
rs554497309 Chr.1: 229452789-229452789 on Build GRCh38
rs992224720 Chr.1: 229452757-229452757 on Build GRCh38
rs879177161 Chr.1: 229452655-229452662 on Build GRCh38
rs776271058 Chr.1: 229452639-229452639 on Build GRCh38
rs765043363 Chr.1: 229452545-229452545 on Build GRCh38
rs575313692 Chr.1: 229452816-229452816 on Build GRCh38
rs201150016 Chr.1: 229452525-229452525 on Build GRCh38
rs112502534 Chr.1: 229452824-229452824 on Build GRCh38
rs754990281 Chr.1: 229452563-229452563 on Build GRCh38
rs370240365 Chr.1: 229452558-229452558 on Build GRCh38
rs137900776 Chr.1: 229452501-229452501 on Build GRCh38
rs761556746 Chr.1: 229452538-229452538 on Build GRCh38
rs897423934 Chr.1: 229452763-229452763 on Build GRCh38
rs571576157 Chr.1: 229452679-229452679 on Build GRCh38
rs772706798 Chr.1: 229452626-229452626 on Build GRCh38
rs1041171996 Chr.1: 229452744-229452744 on Build GRCh38
rs756200848 Chr.1: 229452583-229452583 on Build GRCh38
rs904291980 Chr.1: 229452479-229452479 on Build GRCh38
rs992627686 Chr.1: 229452598-229452598 on Build GRCh38
rs913042749 Chr.1: 229452874-229452874 on Build GRCh38
rs542481332 Chr.1: 229452883-229452883 on Build GRCh38
rs758197941 Chr.1: 229452556-229452556 on Build GRCh38
rs1001934254 Chr.1: 229452829-229452829 on Build GRCh38

View Full Product Details