Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 8612281-8612789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM66
Entrez Gene ID
9866
Gene Name
tripartite motif containing 66
Gene Aliases
C11orf29, TIF1D, TIF1DELTA
UniGene
Hs.130836
Gene Chromosome Location
Chr.11: 8612037-8683217 on Build GRCh38
Transcripts
NM_014818.1 XM_006718397.2 XM_006718398.3 XM_011520504.1 XM_011520507.2 XM_011520508.1 XM_011520509.1 XM_011520510.1 XM_011520511.1 XM_011520512.1 XM_011520513.1 XM_011520514.2 XM_011520515.1 XM_011520516.1 XM_011520517.2 XM_011520518.1 XM_011520519.1 XM_011520522.2 XM_011520523.2 XM_011520526.2 XM_011520527.1 XM_017018629.1 XM_017018630.1 XM_017018631.1

SNP Detail

SNP ID Chromosome Location
rs897558603 Chr.11: 8612730-8612730 on Build GRCh38
rs1055385052 Chr.11: 8612342-8612342 on Build GRCh38
rs1046981462 Chr.11: 8612514-8612514 on Build GRCh38
rs1014713290 Chr.11: 8612352-8612352 on Build GRCh38
rs755284791 Chr.11: 8612311-8612311 on Build GRCh38
rs534355261 Chr.11: 8612500-8612500 on Build GRCh38
rs1015006348 Chr.11: 8612760-8612760 on Build GRCh38
rs147915507 Chr.11: 8612341-8612341 on Build GRCh38
rs1030397816 Chr.11: 8612429-8612429 on Build GRCh38
rs765227928 Chr.11: 8612428-8612428 on Build GRCh38
rs539623540 Chr.11: 8612734-8612734 on Build GRCh38
rs887562379 Chr.11: 8612346-8612346 on Build GRCh38
rs537152303 Chr.11: 8612630-8612630 on Build GRCh38
rs986172581 Chr.11: 8612484-8612484 on Build GRCh38
rs1005336630 Chr.11: 8612743-8612743 on Build GRCh38
rs1042564470 Chr.11: 8612698-8612698 on Build GRCh38
rs372637366 Chr.11: 8612515-8612515 on Build GRCh38
rs556629411 Chr.11: 8612703-8612703 on Build GRCh38
rs1005893667 Chr.11: 8612351-8612351 on Build GRCh38
rs548183833 Chr.11: 8612503-8612503 on Build GRCh38
rs949606688 Chr.11: 8612508-8612508 on Build GRCh38
rs752857910 Chr.11: 8612507-8612507 on Build GRCh38
rs958109845 Chr.11: 8612501-8612501 on Build GRCh38
rs997856747 Chr.11: 8612413-8612413 on Build GRCh38
rs112589900 Chr.11: 8612354-8612355 on Build GRCh38
rs961882361 Chr.11: 8612389-8612389 on Build GRCh38
rs953401812 Chr.11: 8612466-8612466 on Build GRCh38
rs945755845 Chr.11: 8612661-8612661 on Build GRCh38
rs926934337 Chr.11: 8612518-8612518 on Build GRCh38
rs1036084304 Chr.11: 8612713-8612713 on Build GRCh38
rs902721314 Chr.11: 8612700-8612700 on Build GRCh38
rs577070777 Chr.11: 8612712-8612712 on Build GRCh38
rs567934998 Chr.11: 8612542-8612542 on Build GRCh38
rs878884429 Chr.11: 8612620-8612620 on Build GRCh38
rs992474104 Chr.11: 8612560-8612560 on Build GRCh38
rs757117323 Chr.11: 8612608-8612608 on Build GRCh38
rs143494265 Chr.11: 8612742-8612744 on Build GRCh38
rs866464679 Chr.11: 8612663-8612663 on Build GRCh38
rs954935316 Chr.11: 8612444-8612444 on Build GRCh38
rs936836518 Chr.11: 8612701-8612701 on Build GRCh38
rs184490667 Chr.11: 8612735-8612735 on Build GRCh38
rs996152923 Chr.11: 8612720-8612720 on Build GRCh38
rs374227328 Chr.11: 8612743-8612745 on Build GRCh38
rs937016705 Chr.11: 8612557-8612557 on Build GRCh38
rs565939351 Chr.11: 8612474-8612474 on Build GRCh38
rs1015136130 Chr.11: 8612490-8612490 on Build GRCh38
rs938258122 Chr.11: 8612577-8612577 on Build GRCh38

View Full Product Details