Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68974824-68975394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA9
Entrez Gene ID
10350
Gene Name
ATP binding cassette subfamily A member 9
Gene Aliases
EST640918
UniGene
Hs.131686
Gene Chromosome Location
Chr.17: 68974632-69078978 on Build GRCh38
Transcripts
NM_080283.3 XM_017024011.1 XM_017024012.1 XM_017024013.1 XM_017024014.1 XM_017024015.1

SNP Detail

SNP ID Chromosome Location
rs763168553 Chr.17: 68974944-68974944 on Build GRCh38
rs910292417 Chr.17: 68974897-68974897 on Build GRCh38
rs1034476722 Chr.17: 68975071-68975071 on Build GRCh38
rs36036253 Chr.17: 68975117-68975118 on Build GRCh38
rs980048788 Chr.17: 68975002-68975002 on Build GRCh38
rs1046279196 Chr.17: 68975032-68975032 on Build GRCh38
rs928959156 Chr.17: 68975256-68975256 on Build GRCh38
rs894572636 Chr.17: 68975325-68975325 on Build GRCh38
rs879593220 Chr.17: 68975187-68975187 on Build GRCh38
rs922773385 Chr.17: 68975292-68975293 on Build GRCh38
rs1024137099 Chr.17: 68975193-68975193 on Build GRCh38
rs941709572 Chr.17: 68974930-68974930 on Build GRCh38
rs910311334 Chr.17: 68975230-68975230 on Build GRCh38
rs754477539 Chr.17: 68975177-68975177 on Build GRCh38
rs977698711 Chr.17: 68975161-68975161 on Build GRCh38
rs370008140 Chr.17: 68975211-68975211 on Build GRCh38
rs796518516 Chr.17: 68975302-68975302 on Build GRCh38
rs937861141 Chr.17: 68975315-68975315 on Build GRCh38
rs959364324 Chr.17: 68975012-68975012 on Build GRCh38
rs188677912 Chr.17: 68975074-68975074 on Build GRCh38
rs1033676737 Chr.17: 68975011-68975012 on Build GRCh38
rs757677440 Chr.17: 68975096-68975096 on Build GRCh38
rs553283738 Chr.17: 68975081-68975081 on Build GRCh38
rs989907110 Chr.17: 68975247-68975247 on Build GRCh38
rs566856667 Chr.17: 68975306-68975306 on Build GRCh38
rs528105974 Chr.17: 68975001-68975001 on Build GRCh38
rs1050007565 Chr.17: 68975099-68975099 on Build GRCh38
rs142571313 Chr.17: 68975100-68975100 on Build GRCh38
rs552129818 Chr.17: 68975299-68975299 on Build GRCh38
rs377659629 Chr.17: 68975036-68975036 on Build GRCh38
rs373644193 Chr.17: 68974851-68974854 on Build GRCh38
rs528391597 Chr.17: 68975016-68975016 on Build GRCh38
rs528936449 Chr.17: 68975243-68975243 on Build GRCh38
rs1043149419 Chr.17: 68975348-68975348 on Build GRCh38
rs539739811 Chr.17: 68975251-68975251 on Build GRCh38
rs546277608 Chr.17: 68975020-68975020 on Build GRCh38
rs778568541 Chr.17: 68975195-68975195 on Build GRCh38
rs994422990 Chr.17: 68974883-68974883 on Build GRCh38
rs906393068 Chr.17: 68975050-68975050 on Build GRCh38
rs967640537 Chr.17: 68975119-68975119 on Build GRCh38
rs550401936 Chr.17: 68975244-68975244 on Build GRCh38
rs934239319 Chr.17: 68975083-68975083 on Build GRCh38
rs986666701 Chr.17: 68975203-68975203 on Build GRCh38
rs180791150 Chr.17: 68975088-68975088 on Build GRCh38
rs185833264 Chr.17: 68975108-68975108 on Build GRCh38
rs1002438007 Chr.17: 68975069-68975069 on Build GRCh38
rs978237115 Chr.17: 68975281-68975281 on Build GRCh38
rs941035452 Chr.17: 68975103-68975103 on Build GRCh38
rs1055331016 Chr.17: 68975318-68975318 on Build GRCh38
rs985933151 Chr.17: 68974872-68974872 on Build GRCh38
rs1011643649 Chr.17: 68975337-68975337 on Build GRCh38
rs967186752 Chr.17: 68975276-68975276 on Build GRCh38

View Full Product Details