Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135479662-135480225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R26
Entrez Gene ID
9858
Gene Name
protein phosphatase 1 regulatory subunit 26
Gene Aliases
KIAA0649
UniGene
Hs.533260
Gene Chromosome Location
Chr.9: 135477650-135488894 on Build GRCh38
Transcripts
NM_014811.3 XM_005263411.2 XM_017015357.1 XM_017015358.1

SNP Detail

SNP ID Chromosome Location
rs921122879 Chr.9: 135480079-135480079 on Build GRCh38
rs1004719597 Chr.9: 135480121-135480121 on Build GRCh38
rs563511493 Chr.9: 135480125-135480125 on Build GRCh38
rs926229380 Chr.9: 135479957-135479957 on Build GRCh38
rs1016474787 Chr.9: 135480170-135480170 on Build GRCh38
rs1051700848 Chr.9: 135479880-135479880 on Build GRCh38
rs1005405639 Chr.9: 135479766-135479767 on Build GRCh38
rs921176683 Chr.9: 135479873-135479873 on Build GRCh38
rs1036856132 Chr.9: 135479899-135479899 on Build GRCh38
rs975248878 Chr.9: 135479862-135479862 on Build GRCh38
rs987140432 Chr.9: 135480117-135480117 on Build GRCh38
rs932959728 Chr.9: 135480032-135480032 on Build GRCh38
rs1051545953 Chr.9: 135480044-135480044 on Build GRCh38
rs112016231 Chr.9: 135479751-135479751 on Build GRCh38
rs899297252 Chr.9: 135479904-135479904 on Build GRCh38
rs1036501588 Chr.9: 135479772-135479772 on Build GRCh38
rs942913820 Chr.9: 135479963-135479963 on Build GRCh38
rs886186023 Chr.9: 135480077-135480077 on Build GRCh38
rs900143650 Chr.9: 135480024-135480024 on Build GRCh38
rs933931745 Chr.9: 135479876-135479876 on Build GRCh38
rs1035125332 Chr.9: 135479839-135479846 on Build GRCh38
rs956594433 Chr.9: 135480012-135480013 on Build GRCh38
rs1009281200 Chr.9: 135480036-135480041 on Build GRCh38
rs994997244 Chr.9: 135479942-135479942 on Build GRCh38
rs921899761 Chr.9: 135479746-135479746 on Build GRCh38
rs915058817 Chr.9: 135479855-135479855 on Build GRCh38
rs956469193 Chr.9: 135479842-135479842 on Build GRCh38
rs969664280 Chr.9: 135479831-135479831 on Build GRCh38
rs980133180 Chr.9: 135479759-135479759 on Build GRCh38
rs913307425 Chr.9: 135479856-135479856 on Build GRCh38
rs1003826696 Chr.9: 135479987-135479987 on Build GRCh38
rs1038710361 Chr.9: 135479991-135479991 on Build GRCh38
rs972125243 Chr.9: 135480160-135480160 on Build GRCh38
rs992898798 Chr.9: 135479853-135479853 on Build GRCh38
rs978899294 Chr.9: 135479951-135479951 on Build GRCh38
rs1023902176 Chr.9: 135479822-135479823 on Build GRCh38
rs866212363 Chr.9: 135479811-135479811 on Build GRCh38
rs911893531 Chr.9: 135479883-135479883 on Build GRCh38
rs1022473283 Chr.9: 135480068-135480068 on Build GRCh38
rs975196426 Chr.9: 135480074-135480074 on Build GRCh38
rs946254304 Chr.9: 135479888-135479888 on Build GRCh38
rs915088758 Chr.9: 135479700-135479700 on Build GRCh38
rs899345408 Chr.9: 135479807-135479807 on Build GRCh38
rs1045298126 Chr.9: 135479958-135479958 on Build GRCh38
rs1034664719 Chr.9: 135480001-135480001 on Build GRCh38
rs940762074 Chr.9: 135479887-135479887 on Build GRCh38
rs967809356 Chr.9: 135480071-135480071 on Build GRCh38
rs1038867242 Chr.9: 135480109-135480131 on Build GRCh38
rs967858625 Chr.9: 135479857-135479857 on Build GRCh38
rs988309958 Chr.9: 135479851-135479851 on Build GRCh38
rs1034733883 Chr.9: 135479764-135479764 on Build GRCh38
rs960167448 Chr.9: 135479771-135479771 on Build GRCh38
rs931975548 Chr.9: 135479747-135479748 on Build GRCh38
rs890527108 Chr.9: 135479762-135479762 on Build GRCh38
rs946626152 Chr.9: 135479701-135479701 on Build GRCh38
rs1039615980 Chr.9: 135479731-135479731 on Build GRCh38
rs940693966 Chr.9: 135480129-135480129 on Build GRCh38
rs1003209633 Chr.9: 135479836-135479837 on Build GRCh38
rs963585213 Chr.9: 135479738-135479738 on Build GRCh38
rs920674562 Chr.9: 135480194-135480194 on Build GRCh38
rs944581113 Chr.9: 135479757-135479758 on Build GRCh38

View Full Product Details