Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27735484-27735980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
UniGene
Hs.445512
Gene Chromosome Location
Chr.8: 27733316-27772656 on Build GRCh38
Transcripts
NM_001304529.1 NM_001304530.1 NM_001304532.1 NM_018246.2 NR_130761.1 NR_130762.1 XM_011544570.2 XM_011544571.2

SNP Detail

SNP ID Chromosome Location
rs138856582 Chr.8: 27735732-27735732 on Build GRCh38
rs1050082380 Chr.8: 27735543-27735543 on Build GRCh38
rs960979144 Chr.8: 27735718-27735718 on Build GRCh38
rs898298189 Chr.8: 27735527-27735527 on Build GRCh38
rs143309163 Chr.8: 27735684-27735684 on Build GRCh38
rs4732601 Chr.8: 27735698-27735698 on Build GRCh38
rs189074287 Chr.8: 27735608-27735608 on Build GRCh38
rs3832544 Chr.8: 27735923-27735924 on Build GRCh38
rs1008597682 Chr.8: 27735571-27735571 on Build GRCh38
rs531118563 Chr.8: 27735585-27735585 on Build GRCh38
rs908162558 Chr.8: 27735738-27735738 on Build GRCh38
rs892443988 Chr.8: 27735941-27735941 on Build GRCh38
rs565102916 Chr.8: 27735537-27735537 on Build GRCh38
rs1048887 Chr.8: 27735633-27735633 on Build GRCh38
rs891033611 Chr.8: 27735560-27735560 on Build GRCh38
rs560665667 Chr.8: 27735716-27735716 on Build GRCh38
rs1049575745 Chr.8: 27735868-27735868 on Build GRCh38
rs753230224 Chr.8: 27735697-27735697 on Build GRCh38
rs193049795 Chr.8: 27735799-27735799 on Build GRCh38
rs552664085 Chr.8: 27735889-27735889 on Build GRCh38
rs545603932 Chr.8: 27735532-27735532 on Build GRCh38
rs113447699 Chr.8: 27735787-27735787 on Build GRCh38
rs943932752 Chr.8: 27735885-27735885 on Build GRCh38
rs1038085729 Chr.8: 27735514-27735514 on Build GRCh38
rs932425487 Chr.8: 27735829-27735829 on Build GRCh38
rs536271602 Chr.8: 27735884-27735884 on Build GRCh38
rs892476228 Chr.8: 27735586-27735586 on Build GRCh38
rs551572091 Chr.8: 27735745-27735745 on Build GRCh38
rs537236479 Chr.8: 27735784-27735784 on Build GRCh38
rs201295407 Chr.8: 27735506-27735507 on Build GRCh38
rs149136421 Chr.8: 27735678-27735678 on Build GRCh38
rs571418027 Chr.8: 27735750-27735750 on Build GRCh38

View Full Product Details