Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 31064278-31064801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
UniGene
Hs.632050
Gene Chromosome Location
Chr.8: 31033262-31173761 on Build GRCh38
Transcripts
NM_000553.4 XM_011544639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
750649 c.254G>T p.W85L Chr.8: 31064333-31064333 on Build GRCh38
5063161 c.320G>A p.S107N Chr.8: 31064399-31064399 on Build GRCh38
5611167 c.344C>T p.S115F Chr.8: 31064423-31064423 on Build GRCh38
486388 c.301C>G p.Q101E Chr.8: 31064380-31064380 on Build GRCh38
33194 c.275G>T p.G92V Chr.8: 31064354-31064354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142346721 Chr.8: 31064405-31064405 on Build GRCh38
rs1012707449 Chr.8: 31064761-31064761 on Build GRCh38
rs768216398 Chr.8: 31064428-31064428 on Build GRCh38
rs776287613 Chr.8: 31064447-31064447 on Build GRCh38
rs148919437 Chr.8: 31064683-31064683 on Build GRCh38
rs1004139509 Chr.8: 31064647-31064647 on Build GRCh38
rs374898432 Chr.8: 31064418-31064418 on Build GRCh38
rs11574198 Chr.8: 31064525-31064525 on Build GRCh38
rs1041953851 Chr.8: 31064708-31064708 on Build GRCh38
rs555043639 Chr.8: 31064748-31064748 on Build GRCh38
rs906831867 Chr.8: 31064642-31064642 on Build GRCh38
rs773717744 Chr.8: 31064338-31064338 on Build GRCh38
rs766733097 Chr.8: 31064407-31064408 on Build GRCh38
rs904031191 Chr.8: 31064496-31064496 on Build GRCh38
rs180951501 Chr.8: 31064585-31064585 on Build GRCh38
rs919144391 Chr.8: 31064543-31064543 on Build GRCh38
rs758873256 Chr.8: 31064394-31064394 on Build GRCh38
rs894127358 Chr.8: 31064754-31064754 on Build GRCh38
rs922826464 Chr.8: 31064684-31064684 on Build GRCh38
rs979134587 Chr.8: 31064417-31064417 on Build GRCh38
rs945667050 Chr.8: 31064470-31064470 on Build GRCh38
rs772594712 Chr.8: 31064330-31064344 on Build GRCh38
rs1043004968 Chr.8: 31064475-31064475 on Build GRCh38
rs749053792 Chr.8: 31064409-31064409 on Build GRCh38
rs760991758 Chr.8: 31064392-31064393 on Build GRCh38
rs371538747 Chr.8: 31064406-31064406 on Build GRCh38
rs4987239 Chr.8: 31064454-31064454 on Build GRCh38
rs962654960 Chr.8: 31064747-31064747 on Build GRCh38
rs763509734 Chr.8: 31064480-31064480 on Build GRCh38
rs775196937 Chr.8: 31064435-31064435 on Build GRCh38
rs951364909 Chr.8: 31064562-31064562 on Build GRCh38
rs768425708 Chr.8: 31064446-31064446 on Build GRCh38
rs757573403 Chr.8: 31064368-31064368 on Build GRCh38
rs903421722 Chr.8: 31064725-31064727 on Build GRCh38
rs780157148 Chr.8: 31064401-31064401 on Build GRCh38
rs1024166954 Chr.8: 31064495-31064495 on Build GRCh38
rs2230009 Chr.8: 31064419-31064419 on Build GRCh38
rs962825268 Chr.8: 31064366-31064366 on Build GRCh38
rs747587698 Chr.8: 31064304-31064305 on Build GRCh38
rs758563330 Chr.8: 31064697-31064697 on Build GRCh38
rs760452488 Chr.8: 31064320-31064320 on Build GRCh38
rs528811015 Chr.8: 31064609-31064609 on Build GRCh38
rs772108759 Chr.8: 31064429-31064429 on Build GRCh38
rs909870349 Chr.8: 31064630-31064630 on Build GRCh38
rs542896005 Chr.8: 31064526-31064526 on Build GRCh38
rs148758902 Chr.8: 31064349-31064349 on Build GRCh38
rs3213197 Chr.8: 31064610-31064610 on Build GRCh38
rs750732168 Chr.8: 31064478-31064478 on Build GRCh38
rs754221635 Chr.8: 31064355-31064355 on Build GRCh38
rs145764920 Chr.8: 31064438-31064438 on Build GRCh38
rs1040187966 Chr.8: 31064653-31064653 on Build GRCh38
rs144670883 Chr.8: 31064308-31064308 on Build GRCh38
rs765687541 Chr.8: 31064382-31064382 on Build GRCh38
rs765674933 Chr.8: 31064459-31064459 on Build GRCh38
rs936805450 Chr.8: 31064522-31064522 on Build GRCh38
rs750903457 Chr.8: 31064383-31064383 on Build GRCh38

View Full Product Details