Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32443465-32443983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRA
Entrez Gene ID
3122
Gene Name
major histocompatibility complex, class II, DR alpha
Gene Aliases
HLA-DRA1, MLRW
UniGene
Hs.520048
Gene Chromosome Location
Chr.6: 32439842-32445046 on Build GRCh38
Transcripts
NM_019111.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6680828 c.611-9_611-8insC p.? Chr.6: 32443747-32443748 on Build GRCh38
5512233 c.613_614delTT p.F205fs*1 Chr.6: 32443758-32443759 on Build GRCh38
1254267 c.709T>G p.F237V Chr.6: 32443854-32443854 on Build GRCh38
224924 c.632T>C p.L211P Chr.6: 32443777-32443777 on Build GRCh38
126737 c.646G>T p.E216* Chr.6: 32443791-32443791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755226372 Chr.6: 32443925-32443925 on Build GRCh38
rs755315159 Chr.6: 32443831-32443831 on Build GRCh38
rs16822632 Chr.6: 32443935-32443935 on Build GRCh38
rs960870983 Chr.6: 32443871-32443871 on Build GRCh38
rs13218331 Chr.6: 32443585-32443585 on Build GRCh38
rs879185925 Chr.6: 32443754-32443754 on Build GRCh38
rs142044688 Chr.6: 32443903-32443903 on Build GRCh38
rs917732578 Chr.6: 32443659-32443659 on Build GRCh38
rs777243715 Chr.6: 32443848-32443848 on Build GRCh38
rs780623107 Chr.6: 32443958-32443958 on Build GRCh38
rs879058172 Chr.6: 32443926-32443926 on Build GRCh38
rs11544315 Chr.6: 32443796-32443796 on Build GRCh38
rs201633626 Chr.6: 32443687-32443687 on Build GRCh38
rs548267071 Chr.6: 32443550-32443550 on Build GRCh38
rs139874048 Chr.6: 32443808-32443808 on Build GRCh38
rs916798925 Chr.6: 32443657-32443657 on Build GRCh38
rs375630609 Chr.6: 32443704-32443704 on Build GRCh38
rs1001627549 Chr.6: 32443916-32443916 on Build GRCh38
rs371441506 Chr.6: 32443512-32443512 on Build GRCh38
rs778808460 Chr.6: 32443675-32443675 on Build GRCh38
rs557232047 Chr.6: 32443583-32443583 on Build GRCh38
rs117531802 Chr.6: 32443613-32443613 on Build GRCh38
rs766104968 Chr.6: 32443742-32443742 on Build GRCh38
rs762948804 Chr.6: 32443872-32443872 on Build GRCh38
rs747520598 Chr.6: 32443950-32443950 on Build GRCh38
rs960905364 Chr.6: 32443602-32443602 on Build GRCh38
rs576887908 Chr.6: 32443691-32443691 on Build GRCh38
rs879014311 Chr.6: 32443537-32443540 on Build GRCh38
rs754080261 Chr.6: 32443907-32443907 on Build GRCh38
rs770380561 Chr.6: 32443853-32443853 on Build GRCh38
rs771561521 Chr.6: 32443511-32443511 on Build GRCh38
rs916851071 Chr.6: 32443920-32443920 on Build GRCh38
rs553117732 Chr.6: 32443523-32443523 on Build GRCh38
rs3129888 Chr.6: 32443949-32443949 on Build GRCh38
rs368601450 Chr.6: 32443778-32443778 on Build GRCh38
rs757405321 Chr.6: 32443615-32443615 on Build GRCh38
rs759121846 Chr.6: 32443867-32443867 on Build GRCh38
rs111492440 Chr.6: 32443707-32443707 on Build GRCh38
rs139677040 Chr.6: 32443749-32443749 on Build GRCh38
rs758466353 Chr.6: 32443759-32443759 on Build GRCh38
rs746125136 Chr.6: 32443733-32443733 on Build GRCh38
rs370989285 Chr.6: 32443735-32443735 on Build GRCh38
rs77288165 Chr.6: 32443753-32443753 on Build GRCh38
rs946733940 Chr.6: 32443666-32443666 on Build GRCh38
rs530789959 Chr.6: 32443893-32443893 on Build GRCh38
rs7192 Chr.6: 32443869-32443869 on Build GRCh38
rs776279402 Chr.6: 32443863-32443863 on Build GRCh38
rs754750691 Chr.6: 32443569-32443569 on Build GRCh38
rs149917607 Chr.6: 32443847-32443847 on Build GRCh38
rs764172368 Chr.6: 32443873-32443873 on Build GRCh38
rs552421989 Chr.6: 32443894-32443894 on Build GRCh38
rs959510329 Chr.6: 32443600-32443600 on Build GRCh38
rs541798926 Chr.6: 32443868-32443868 on Build GRCh38
rs878981056 Chr.6: 32443927-32443927 on Build GRCh38
rs60110771 Chr.6: 32443509-32443509 on Build GRCh38
rs991037547 Chr.6: 32443608-32443608 on Build GRCh38
rs995975632 Chr.6: 32443766-32443766 on Build GRCh38
rs1006122813 Chr.6: 32443799-32443799 on Build GRCh38
rs374588694 Chr.6: 32443740-32443740 on Build GRCh38
rs373187293 Chr.6: 32443954-32443954 on Build GRCh38
rs762199845 Chr.6: 32443747-32443748 on Build GRCh38
rs541984284 Chr.6: 32443899-32443899 on Build GRCh38
rs780271812 Chr.6: 32443819-32443819 on Build GRCh38
rs749480059 Chr.6: 32443826-32443826 on Build GRCh38
rs371727409 Chr.6: 32443748-32443748 on Build GRCh38
rs574757583 Chr.6: 32443528-32443528 on Build GRCh38
rs751562870 Chr.6: 32443875-32443875 on Build GRCh38
rs962124507 Chr.6: 32443531-32443531 on Build GRCh38
rs776171876 Chr.6: 32443744-32443744 on Build GRCh38
rs779308837 Chr.6: 32443929-32443929 on Build GRCh38
rs375429096 Chr.6: 32443948-32443948 on Build GRCh38
rs779207648 Chr.6: 32443838-32443838 on Build GRCh38
rs977938714 Chr.6: 32443728-32443728 on Build GRCh38
rs368192225 Chr.6: 32443732-32443732 on Build GRCh38
rs748653096 Chr.6: 32443843-32443843 on Build GRCh38
rs746475812 Chr.6: 32443852-32443852 on Build GRCh38
rs2239804 Chr.6: 32443746-32443746 on Build GRCh38
rs2239806 Chr.6: 32443530-32443530 on Build GRCh38
rs2239805 Chr.6: 32443599-32443599 on Build GRCh38
rs576697175 Chr.6: 32443561-32443561 on Build GRCh38

View Full Product Details