Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61187427-61188002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSA2
Entrez Gene ID
130872
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Gene Aliases
Hch1
UniGene
Hs.655602
Gene Chromosome Location
Chr.2: 61177418-61191210 on Build GRCh38
Transcripts
NM_001321300.1 NM_001321301.1 NM_001321302.1 NM_001321303.1 NM_001321304.1 NM_001321305.1 NM_152392.4 XM_011532541.2 XM_011532543.2 XM_011532545.2 XM_017003377.1 XM_017003378.1 NM_014709.3

Gene Symbol
USP34
Entrez Gene ID
9736
Gene Name
ubiquitin specific peptidase 34
Gene Aliases
-
UniGene
Hs.644708
Gene Chromosome Location
Chr.2: 61187455-61470714 on Build GRCh38
Transcripts
NM_001321300.1 NM_001321301.1 NM_001321302.1 NM_001321303.1 NM_001321304.1 NM_001321305.1 NM_152392.4 XM_011532541.2 XM_011532543.2 XM_011532545.2 XM_017003377.1 XM_017003378.1 NM_014709.3

SNP Detail

SNP ID Chromosome Location
rs945242949 Chr.2: 61187774-61187774 on Build GRCh38
rs911696688 Chr.2: 61187814-61187814 on Build GRCh38
rs1016121677 Chr.2: 61187966-61187966 on Build GRCh38
rs955449229 Chr.2: 61187705-61187705 on Build GRCh38
rs780131018 Chr.2: 61187840-61187840 on Build GRCh38
rs1053696917 Chr.2: 61187477-61187477 on Build GRCh38
rs1045561133 Chr.2: 61187512-61187512 on Build GRCh38
rs1024388930 Chr.2: 61187695-61187695 on Build GRCh38
rs865847560 Chr.2: 61187456-61187456 on Build GRCh38
rs925954298 Chr.2: 61187469-61187471 on Build GRCh38
rs973943645 Chr.2: 61187730-61187730 on Build GRCh38
rs3841388 Chr.2: 61187761-61187762 on Build GRCh38
rs1045507749 Chr.2: 61187760-61187760 on Build GRCh38
rs529132736 Chr.2: 61187467-61187467 on Build GRCh38
rs187721106 Chr.2: 61187674-61187674 on Build GRCh38
rs547710189 Chr.2: 61187469-61187469 on Build GRCh38
rs1011156445 Chr.2: 61187665-61187665 on Build GRCh38
rs1047351330 Chr.2: 61187879-61187879 on Build GRCh38
rs1038288947 Chr.2: 61187450-61187450 on Build GRCh38
rs192206142 Chr.2: 61187928-61187928 on Build GRCh38
rs752014377 Chr.2: 61187696-61187696 on Build GRCh38
rs1051547677 Chr.2: 61187554-61187554 on Build GRCh38
rs1029847559 Chr.2: 61187688-61187688 on Build GRCh38
rs892792537 Chr.2: 61187832-61187832 on Build GRCh38
rs915806362 Chr.2: 61187739-61187739 on Build GRCh38
rs3217620 Chr.2: 61187515-61187518 on Build GRCh38
rs762005879 Chr.2: 61187476-61187476 on Build GRCh38
rs762965552 Chr.2: 61187585-61187586 on Build GRCh38
rs780616057 Chr.2: 61187905-61187905 on Build GRCh38
rs928463409 Chr.2: 61187765-61187765 on Build GRCh38
rs983273784 Chr.2: 61187713-61187713 on Build GRCh38
rs201709566 Chr.2: 61187508-61187508 on Build GRCh38
rs936598865 Chr.2: 61187471-61187475 on Build GRCh38
rs565664995 Chr.2: 61187505-61187512 on Build GRCh38
rs994504512 Chr.2: 61187872-61187872 on Build GRCh38
rs765484115 Chr.2: 61187524-61187524 on Build GRCh38
rs866503877 Chr.2: 61187679-61187679 on Build GRCh38
rs528239073 Chr.2: 61187509-61187512 on Build GRCh38
rs530209873 Chr.2: 61187829-61187829 on Build GRCh38
rs900054395 Chr.2: 61187867-61187867 on Build GRCh38
rs1051613636 Chr.2: 61187800-61187800 on Build GRCh38
rs906922430 Chr.2: 61187514-61187514 on Build GRCh38
rs890219635 Chr.2: 61187560-61187560 on Build GRCh38
rs1038737194 Chr.2: 61187856-61187856 on Build GRCh38
rs527712653 Chr.2: 61187655-61187655 on Build GRCh38
rs747693313 Chr.2: 61187907-61187907 on Build GRCh38
rs996992972 Chr.2: 61187686-61187686 on Build GRCh38
rs995519038 Chr.2: 61187668-61187668 on Build GRCh38
rs1053315610 Chr.2: 61187831-61187831 on Build GRCh38
rs566749020 Chr.2: 61187602-61187602 on Build GRCh38
rs750801604 Chr.2: 61187631-61187631 on Build GRCh38
rs371160915 Chr.2: 61187724-61187724 on Build GRCh38
rs945191784 Chr.2: 61187534-61187534 on Build GRCh38
rs989845128 Chr.2: 61187746-61187746 on Build GRCh38
rs556754974 Chr.2: 61187878-61187878 on Build GRCh38
rs891326992 Chr.2: 61187911-61187911 on Build GRCh38
rs758576699 Chr.2: 61187722-61187722 on Build GRCh38
rs948580263 Chr.2: 61187478-61187478 on Build GRCh38
rs754500703 Chr.2: 61187792-61187792 on Build GRCh38
rs934427488 Chr.2: 61187770-61187770 on Build GRCh38
rs561466742 Chr.2: 61187669-61187669 on Build GRCh38
rs948651941 Chr.2: 61187747-61187747 on Build GRCh38
rs913814386 Chr.2: 61187762-61187762 on Build GRCh38
rs541132321 Chr.2: 61187663-61187663 on Build GRCh38
rs769393544 Chr.2: 61187932-61187932 on Build GRCh38
rs997107575 Chr.2: 61187886-61187886 on Build GRCh38
rs538346630 Chr.2: 61187734-61187734 on Build GRCh38
rs564888708 Chr.2: 61187459-61187459 on Build GRCh38
rs908536712 Chr.2: 61187720-61187720 on Build GRCh38
rs767751640 Chr.2: 61187672-61187677 on Build GRCh38
rs879237901 Chr.2: 61187725-61187726 on Build GRCh38
rs201913510 Chr.2: 61187515-61187515 on Build GRCh38
rs917323895 Chr.2: 61187450-61187451 on Build GRCh38
rs16348 Chr.2: 61187505-61187508 on Build GRCh38
rs1002050275 Chr.2: 61187948-61187948 on Build GRCh38
rs980514074 Chr.2: 61187464-61187465 on Build GRCh38
rs1004291637 Chr.2: 61187936-61187936 on Build GRCh38
rs898816087 Chr.2: 61187666-61187666 on Build GRCh38
rs747705896 Chr.2: 61187484-61187487 on Build GRCh38
rs970239950 Chr.2: 61187925-61187925 on Build GRCh38
rs35833610 Chr.2: 61187686-61187687 on Build GRCh38

View Full Product Details