Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 83981189-83981764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA3A
Entrez Gene ID
10371
Gene Name
semaphorin 3A
Gene Aliases
COLL1, HH16, Hsema-I, Hsema-III, SEMA1, SEMAD, SEMAIII, SEMAL, SemD, coll-1
UniGene
Hs.252451
Gene Chromosome Location
Chr.7: 83956846-84492724 on Build GRCh38
Transcripts
NM_006080.2 XM_005250110.3 XM_005250111.4 XM_006715839.3 XM_011515734.2 XM_017011673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6774492 c.1560C>T p.Y520Y Chr.7: 83981413-83981413 on Build GRCh38
747804 c.1517C>A p.T506K Chr.7: 83981456-83981456 on Build GRCh38
461529 c.1588G>A p.A530T Chr.7: 83981385-83981385 on Build GRCh38
5408134 c.1587C>T p.L529L Chr.7: 83981386-83981386 on Build GRCh38
6193509 c.1546C>T p.R516W Chr.7: 83981427-83981427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111491741 Chr.7: 83981309-83981309 on Build GRCh38
rs902319777 Chr.7: 83981688-83981688 on Build GRCh38
rs781780991 Chr.7: 83981386-83981386 on Build GRCh38
rs542021082 Chr.7: 83981650-83981650 on Build GRCh38
rs564500996 Chr.7: 83981279-83981279 on Build GRCh38
rs199943253 Chr.7: 83981449-83981449 on Build GRCh38
rs764946516 Chr.7: 83981437-83981437 on Build GRCh38
rs558111586 Chr.7: 83981533-83981533 on Build GRCh38
rs17158435 Chr.7: 83981224-83981224 on Build GRCh38
rs949279427 Chr.7: 83981654-83981654 on Build GRCh38
rs201612526 Chr.7: 83981483-83981483 on Build GRCh38
rs942160612 Chr.7: 83981561-83981561 on Build GRCh38
rs940179933 Chr.7: 83981583-83981583 on Build GRCh38
rs895216469 Chr.7: 83981595-83981595 on Build GRCh38
rs919666991 Chr.7: 83981590-83981590 on Build GRCh38
rs748598600 Chr.7: 83981456-83981456 on Build GRCh38
rs912709128 Chr.7: 83981564-83981564 on Build GRCh38
rs553752767 Chr.7: 83981704-83981704 on Build GRCh38
rs1012365903 Chr.7: 83981608-83981608 on Build GRCh38
rs529494220 Chr.7: 83981440-83981440 on Build GRCh38
rs1031563824 Chr.7: 83981719-83981719 on Build GRCh38
rs751075384 Chr.7: 83981450-83981450 on Build GRCh38
rs748508588 Chr.7: 83981388-83981388 on Build GRCh38
rs10487865 Chr.7: 83981410-83981410 on Build GRCh38
rs908012186 Chr.7: 83981555-83981556 on Build GRCh38
rs764858876 Chr.7: 83981342-83981342 on Build GRCh38
rs574504688 Chr.7: 83981622-83981622 on Build GRCh38
rs758022840 Chr.7: 83981447-83981447 on Build GRCh38
rs972439526 Chr.7: 83981586-83981586 on Build GRCh38
rs560402131 Chr.7: 83981230-83981230 on Build GRCh38
rs770224463 Chr.7: 83981404-83981404 on Build GRCh38
rs1046310169 Chr.7: 83981675-83981675 on Build GRCh38
rs745420951 Chr.7: 83981353-83981353 on Build GRCh38
rs1012642284 Chr.7: 83981287-83981287 on Build GRCh38
rs774464552 Chr.7: 83981417-83981417 on Build GRCh38
rs546200170 Chr.7: 83981261-83981261 on Build GRCh38
rs374162228 Chr.7: 83981381-83981381 on Build GRCh38
rs1038202700 Chr.7: 83981264-83981264 on Build GRCh38
rs574692442 Chr.7: 83981547-83981547 on Build GRCh38
rs1051254078 Chr.7: 83981254-83981254 on Build GRCh38
rs745950134 Chr.7: 83981515-83981515 on Build GRCh38
rs771077834 Chr.7: 83981413-83981413 on Build GRCh38
rs746665974 Chr.7: 83981641-83981641 on Build GRCh38
rs370307743 Chr.7: 83981494-83981494 on Build GRCh38
rs750024161 Chr.7: 83981343-83981343 on Build GRCh38
rs778230837 Chr.7: 83981462-83981462 on Build GRCh38
rs779744872 Chr.7: 83981454-83981454 on Build GRCh38
rs369709964 Chr.7: 83981350-83981350 on Build GRCh38
rs148900275 Chr.7: 83981427-83981427 on Build GRCh38
rs377259138 Chr.7: 83981405-83981405 on Build GRCh38
rs369182739 Chr.7: 83981457-83981457 on Build GRCh38
rs537540376 Chr.7: 83981600-83981600 on Build GRCh38
rs374226073 Chr.7: 83981275-83981275 on Build GRCh38
rs376837957 Chr.7: 83981329-83981329 on Build GRCh38
rs772086583 Chr.7: 83981528-83981528 on Build GRCh38
rs555842012 Chr.7: 83981609-83981609 on Build GRCh38
rs527649336 Chr.7: 83981234-83981234 on Build GRCh38
rs1024744950 Chr.7: 83981430-83981430 on Build GRCh38
rs780861957 Chr.7: 83981455-83981455 on Build GRCh38
rs574334384 Chr.7: 83981218-83981218 on Build GRCh38
rs760607396 Chr.7: 83981290-83981290 on Build GRCh38
rs759578109 Chr.7: 83981421-83981421 on Build GRCh38
rs552964434 Chr.7: 83981491-83981491 on Build GRCh38
rs971711679 Chr.7: 83981377-83981377 on Build GRCh38
rs753603368 Chr.7: 83981306-83981306 on Build GRCh38
rs755489133 Chr.7: 83981385-83981385 on Build GRCh38
rs999315610 Chr.7: 83981737-83981737 on Build GRCh38
rs776611896 Chr.7: 83981426-83981426 on Build GRCh38
rs763983106 Chr.7: 83981291-83981291 on Build GRCh38
rs545192996 Chr.7: 83981425-83981425 on Build GRCh38
rs746525414 Chr.7: 83981382-83981382 on Build GRCh38
rs930976605 Chr.7: 83981407-83981407 on Build GRCh38

View Full Product Details