Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111913313-111913912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPB2
Entrez Gene ID
3316
Gene Name
heat shock protein family B (small) member 2
Gene Aliases
HSP27, Hs.78846, LOH11CR1K, MKBP
UniGene
Hs.709660
Gene Chromosome Location
Chr.11: 111912736-111914093 on Build GRCh38
Transcripts
NM_001541.3 NR_037651.1

Gene Symbol
HSPB2-C11orf52
Entrez Gene ID
100528019
Gene Name
HSPB2-C11orf52 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.97013
Gene Chromosome Location
Chr.11: 111912736-111926871 on Build GRCh38
Transcripts
NM_001541.3 NR_037651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6552666 c.175G>T p.G59C Chr.11: 111913521-111913521 on Build GRCh38
261341 c.196G>A p.E66K Chr.11: 111913542-111913542 on Build GRCh38
6251722 c.356G>T p.R119L Chr.11: 111913702-111913702 on Build GRCh38
2111838 c.500C>T p.A167V Chr.11: 111913846-111913846 on Build GRCh38
6053788 c.455A>C p.H152P Chr.11: 111913801-111913801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367609134 Chr.11: 111913392-111913392 on Build GRCh38
rs781825092 Chr.11: 111913480-111913480 on Build GRCh38
rs782167833 Chr.11: 111913768-111913768 on Build GRCh38
rs146155375 Chr.11: 111913878-111913878 on Build GRCh38
rs887591018 Chr.11: 111913535-111913535 on Build GRCh38
rs587698938 Chr.11: 111913585-111913585 on Build GRCh38
rs782471186 Chr.11: 111913556-111913556 on Build GRCh38
rs782456306 Chr.11: 111913703-111913703 on Build GRCh38
rs782777418 Chr.11: 111913534-111913534 on Build GRCh38
rs782496139 Chr.11: 111913882-111913882 on Build GRCh38
rs145827670 Chr.11: 111913457-111913457 on Build GRCh38
rs141000255 Chr.11: 111913791-111913791 on Build GRCh38
rs782144055 Chr.11: 111913521-111913521 on Build GRCh38
rs587647075 Chr.11: 111913461-111913461 on Build GRCh38
rs587616093 Chr.11: 111913760-111913760 on Build GRCh38
rs1050526 Chr.11: 111913826-111913826 on Build GRCh38
rs782149177 Chr.11: 111913830-111913830 on Build GRCh38
rs782682010 Chr.11: 111913503-111913503 on Build GRCh38
rs754003263 Chr.11: 111913509-111913519 on Build GRCh38
rs782512419 Chr.11: 111913706-111913706 on Build GRCh38
rs983384295 Chr.11: 111913690-111913690 on Build GRCh38
rs782340446 Chr.11: 111913646-111913646 on Build GRCh38
rs782030412 Chr.11: 111913637-111913637 on Build GRCh38
rs781975640 Chr.11: 111913647-111913647 on Build GRCh38
rs4252588 Chr.11: 111913505-111913505 on Build GRCh38
rs782583689 Chr.11: 111913489-111913489 on Build GRCh38
rs985530456 Chr.11: 111913445-111913445 on Build GRCh38
rs545557968 Chr.11: 111913416-111913416 on Build GRCh38
rs587644541 Chr.11: 111913889-111913889 on Build GRCh38
rs782533003 Chr.11: 111913798-111913798 on Build GRCh38
rs587699399 Chr.11: 111913495-111913495 on Build GRCh38
rs587764844 Chr.11: 111913885-111913885 on Build GRCh38
rs781938446 Chr.11: 111913513-111913513 on Build GRCh38
rs200672845 Chr.11: 111913733-111913733 on Build GRCh38
rs1054895107 Chr.11: 111913559-111913559 on Build GRCh38
rs146268210 Chr.11: 111913546-111913546 on Build GRCh38
rs781799087 Chr.11: 111913797-111913797 on Build GRCh38
rs782751614 Chr.11: 111913682-111913682 on Build GRCh38
rs76737820 Chr.11: 111913669-111913669 on Build GRCh38
rs1025054358 Chr.11: 111913587-111913587 on Build GRCh38
rs4252589 Chr.11: 111913677-111913677 on Build GRCh38
rs950685918 Chr.11: 111913689-111913689 on Build GRCh38
rs782303314 Chr.11: 111913727-111913727 on Build GRCh38
rs782052133 Chr.11: 111913750-111913750 on Build GRCh38
rs377506407 Chr.11: 111913482-111913482 on Build GRCh38
rs199640609 Chr.11: 111913875-111913875 on Build GRCh38
rs782763815 Chr.11: 111913769-111913769 on Build GRCh38
rs782553149 Chr.11: 111913500-111913500 on Build GRCh38
rs202111419 Chr.11: 111913520-111913520 on Build GRCh38
rs535736907 Chr.11: 111913364-111913364 on Build GRCh38
rs201135642 Chr.11: 111913718-111913718 on Build GRCh38
rs782202385 Chr.11: 111913672-111913672 on Build GRCh38
rs556929117 Chr.11: 111913381-111913381 on Build GRCh38
rs781830805 Chr.11: 111913880-111913880 on Build GRCh38
rs587693757 Chr.11: 111913789-111913789 on Build GRCh38
rs1053974807 Chr.11: 111913762-111913762 on Build GRCh38
rs568298059 Chr.11: 111913339-111913339 on Build GRCh38
rs376090834 Chr.11: 111913577-111913577 on Build GRCh38
rs782136947 Chr.11: 111913742-111913742 on Build GRCh38
rs782384792 Chr.11: 111913804-111913804 on Build GRCh38
rs782676473 Chr.11: 111913799-111913799 on Build GRCh38
rs782198420 Chr.11: 111913891-111913891 on Build GRCh38
rs782086590 Chr.11: 111913538-111913538 on Build GRCh38
rs142266981 Chr.11: 111913507-111913507 on Build GRCh38
rs149492778 Chr.11: 111913680-111913680 on Build GRCh38
rs587639270 Chr.11: 111913511-111913511 on Build GRCh38
rs782498365 Chr.11: 111913369-111913369 on Build GRCh38
rs782049019 Chr.11: 111913447-111913447 on Build GRCh38
rs587739341 Chr.11: 111913738-111913738 on Build GRCh38
rs918568668 Chr.11: 111913688-111913688 on Build GRCh38
rs782421371 Chr.11: 111913831-111913831 on Build GRCh38
rs587679193 Chr.11: 111913737-111913737 on Build GRCh38
rs147812082 Chr.11: 111913589-111913589 on Build GRCh38
rs781799151 Chr.11: 111913553-111913553 on Build GRCh38
rs782646187 Chr.11: 111913406-111913406 on Build GRCh38
rs782498567 Chr.11: 111913876-111913876 on Build GRCh38
rs781982989 Chr.11: 111913845-111913845 on Build GRCh38
rs782617445 Chr.11: 111913819-111913819 on Build GRCh38
rs111836778 Chr.11: 111913373-111913373 on Build GRCh38
rs587627710 Chr.11: 111913610-111913610 on Build GRCh38
rs916367766 Chr.11: 111913728-111913728 on Build GRCh38
rs192172142 Chr.11: 111913846-111913846 on Build GRCh38
rs45602839 Chr.11: 111913811-111913811 on Build GRCh38
rs782060242 Chr.11: 111913870-111913870 on Build GRCh38
rs782415821 Chr.11: 111913635-111913635 on Build GRCh38
rs782315130 Chr.11: 111913825-111913825 on Build GRCh38
rs781834478 Chr.11: 111913446-111913446 on Build GRCh38
rs782506463 Chr.11: 111913467-111913467 on Build GRCh38
rs782803706 Chr.11: 111913873-111913873 on Build GRCh38
rs782717759 Chr.11: 111913650-111913650 on Build GRCh38
rs575404020 Chr.11: 111913412-111913412 on Build GRCh38
rs782740585 Chr.11: 111913442-111913442 on Build GRCh38
rs781794984 Chr.11: 111913683-111913683 on Build GRCh38
rs967362137 Chr.11: 111913361-111913361 on Build GRCh38
rs782767495 Chr.11: 111913435-111913436 on Build GRCh38
rs978319137 Chr.11: 111913378-111913378 on Build GRCh38
rs782241905 Chr.11: 111913801-111913801 on Build GRCh38
rs782609126 Chr.11: 111913716-111913716 on Build GRCh38
rs764919055 Chr.11: 111913702-111913702 on Build GRCh38
rs758224029 Chr.11: 111913823-111913823 on Build GRCh38
rs587724960 Chr.11: 111913537-111913537 on Build GRCh38
rs202071359 Chr.11: 111913849-111913849 on Build GRCh38
rs143011638 Chr.11: 111913836-111913836 on Build GRCh38
rs781787278 Chr.11: 111913851-111913851 on Build GRCh38
rs782393308 Chr.11: 111913788-111913788 on Build GRCh38
rs587694964 Chr.11: 111913649-111913649 on Build GRCh38
rs782651807 Chr.11: 111913470-111913470 on Build GRCh38
rs146569701 Chr.11: 111913459-111913459 on Build GRCh38
rs782260577 Chr.11: 111913641-111913641 on Build GRCh38
rs782230322 Chr.11: 111913824-111913824 on Build GRCh38
rs782718651 Chr.11: 111913752-111913752 on Build GRCh38
rs150314549 Chr.11: 111913656-111913656 on Build GRCh38
rs1032460520 Chr.11: 111913399-111913399 on Build GRCh38
rs587754107 Chr.11: 111913586-111913586 on Build GRCh38
rs782564961 Chr.11: 111913605-111913605 on Build GRCh38
rs782242246 Chr.11: 111913708-111913708 on Build GRCh38
rs375687384 Chr.11: 111913847-111913847 on Build GRCh38
rs782494423 Chr.11: 111913545-111913545 on Build GRCh38

View Full Product Details