Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46612887-46613482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOB3C
Entrez Gene ID
148932
Gene Name
MOB kinase activator 3C
Gene Aliases
MOB1E, MOBKL2C
UniGene
Hs.632401
Gene Chromosome Location
Chr.1: 46607715-46616891 on Build GRCh38
Transcripts
NM_145279.4 NM_201403.2 XM_017000407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4165225 c.28C>T p.Q10* Chr.1: 46613363-46613363 on Build GRCh38
4499492 c.543C>T p.I181I Chr.1: 46612935-46612935 on Build GRCh38
6714088 c.67G>A p.A23T Chr.1: 46613324-46613324 on Build GRCh38
7401748 c.390G>A p.M130I Chr.1: 46613088-46613088 on Build GRCh38
4647778 c.325A>G p.S109G Chr.1: 46613066-46613066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769417720 Chr.1: 46613234-46613254 on Build GRCh38
rs375302001 Chr.1: 46613141-46613141 on Build GRCh38
rs1034535136 Chr.1: 46613018-46613018 on Build GRCh38
rs749258127 Chr.1: 46613300-46613300 on Build GRCh38
rs748781587 Chr.1: 46613024-46613024 on Build GRCh38
rs764208170 Chr.1: 46613119-46613119 on Build GRCh38
rs768093778 Chr.1: 46613078-46613078 on Build GRCh38
rs765420980 Chr.1: 46613044-46613044 on Build GRCh38
rs765320718 Chr.1: 46613111-46613111 on Build GRCh38
rs200539864 Chr.1: 46612995-46612995 on Build GRCh38
rs778095678 Chr.1: 46613054-46613054 on Build GRCh38
rs755541058 Chr.1: 46613368-46613368 on Build GRCh38
rs771605084 Chr.1: 46613087-46613087 on Build GRCh38
rs201224467 Chr.1: 46613294-46613294 on Build GRCh38
rs756563654 Chr.1: 46613233-46613233 on Build GRCh38
rs766445532 Chr.1: 46613281-46613281 on Build GRCh38
rs753948020 Chr.1: 46613000-46613000 on Build GRCh38
rs780667819 Chr.1: 46613031-46613032 on Build GRCh38
rs772855014 Chr.1: 46613035-46613035 on Build GRCh38
rs748454556 Chr.1: 46612947-46612947 on Build GRCh38
rs377232112 Chr.1: 46613053-46613053 on Build GRCh38
rs71638014 Chr.1: 46612984-46612984 on Build GRCh38
rs574352913 Chr.1: 46613038-46613038 on Build GRCh38
rs760885130 Chr.1: 46613344-46613344 on Build GRCh38
rs1006451264 Chr.1: 46613027-46613027 on Build GRCh38
rs768339116 Chr.1: 46612991-46613007 on Build GRCh38
rs746675262 Chr.1: 46613160-46613160 on Build GRCh38
rs758733206 Chr.1: 46613013-46613013 on Build GRCh38
rs139968018 Chr.1: 46613421-46613421 on Build GRCh38
rs780986230 Chr.1: 46613382-46613382 on Build GRCh38
rs371678179 Chr.1: 46613140-46613140 on Build GRCh38
rs566315966 Chr.1: 46613378-46613378 on Build GRCh38
rs750312246 Chr.1: 46612994-46612994 on Build GRCh38
rs61753780 Chr.1: 46613117-46613117 on Build GRCh38
rs566258784 Chr.1: 46612954-46612954 on Build GRCh38
rs776600787 Chr.1: 46613171-46613171 on Build GRCh38
rs1026421185 Chr.1: 46613258-46613258 on Build GRCh38
rs770974479 Chr.1: 46613391-46613392 on Build GRCh38
rs1015597550 Chr.1: 46613439-46613439 on Build GRCh38
rs768603868 Chr.1: 46613403-46613403 on Build GRCh38
rs774496217 Chr.1: 46613206-46613206 on Build GRCh38
rs756353510 Chr.1: 46613135-46613135 on Build GRCh38
rs115135917 Chr.1: 46613231-46613231 on Build GRCh38
rs771395935 Chr.1: 46613020-46613020 on Build GRCh38
rs777170203 Chr.1: 46613023-46613023 on Build GRCh38
rs764236187 Chr.1: 46613205-46613205 on Build GRCh38
rs763711743 Chr.1: 46612975-46612975 on Build GRCh38
rs775553966 Chr.1: 46613203-46613203 on Build GRCh38
rs773493416 Chr.1: 46613325-46613325 on Build GRCh38
rs1055309020 Chr.1: 46613444-46613444 on Build GRCh38
rs756586971 Chr.1: 46613138-46613138 on Build GRCh38
rs776654062 Chr.1: 46613100-46613100 on Build GRCh38
rs768567480 Chr.1: 46613306-46613306 on Build GRCh38
rs747995999 Chr.1: 46613231-46613232 on Build GRCh38
rs748534010 Chr.1: 46612928-46612928 on Build GRCh38
rs751846975 Chr.1: 46613058-46613058 on Build GRCh38
rs779275572 Chr.1: 46613003-46613003 on Build GRCh38
rs752687868 Chr.1: 46613114-46613114 on Build GRCh38
rs779919343 Chr.1: 46613392-46613392 on Build GRCh38
rs781660781 Chr.1: 46613060-46613060 on Build GRCh38
rs754376722 Chr.1: 46613252-46613252 on Build GRCh38
rs745644252 Chr.1: 46613278-46613278 on Build GRCh38
rs767825672 Chr.1: 46613227-46613227 on Build GRCh38
rs904953441 Chr.1: 46613272-46613272 on Build GRCh38
rs775011378 Chr.1: 46612970-46612970 on Build GRCh38
rs757789714 Chr.1: 46613254-46613254 on Build GRCh38
rs766793952 Chr.1: 46613243-46613243 on Build GRCh38
rs773930180 Chr.1: 46612980-46612980 on Build GRCh38
rs773204541 Chr.1: 46613280-46613281 on Build GRCh38
rs529251316 Chr.1: 46613317-46613317 on Build GRCh38
rs759444040 Chr.1: 46613174-46613174 on Build GRCh38
rs766409369 Chr.1: 46613039-46613039 on Build GRCh38
rs768220375 Chr.1: 46613025-46613025 on Build GRCh38
rs568493531 Chr.1: 46613302-46613302 on Build GRCh38
rs779328921 Chr.1: 46612925-46612925 on Build GRCh38
rs936571990 Chr.1: 46613133-46613133 on Build GRCh38
rs757674170 Chr.1: 46613059-46613059 on Build GRCh38
rs547494627 Chr.1: 46613324-46613324 on Build GRCh38
rs112622804 Chr.1: 46613081-46613081 on Build GRCh38
rs756885909 Chr.1: 46613380-46613380 on Build GRCh38
rs373731733 Chr.1: 46613371-46613371 on Build GRCh38
rs770855909 Chr.1: 46613097-46613097 on Build GRCh38
rs915348490 Chr.1: 46613357-46613357 on Build GRCh38
rs541497333 Chr.1: 46613042-46613042 on Build GRCh38
rs767217807 Chr.1: 46612987-46612987 on Build GRCh38
rs893771080 Chr.1: 46612977-46612977 on Build GRCh38
rs141351633 Chr.1: 46613033-46613033 on Build GRCh38
rs778021599 Chr.1: 46613015-46613015 on Build GRCh38
rs115764884 Chr.1: 46613034-46613034 on Build GRCh38
rs751844185 Chr.1: 46613129-46613129 on Build GRCh38
rs757549206 Chr.1: 46613132-46613132 on Build GRCh38
rs138258229 Chr.1: 46612982-46612982 on Build GRCh38
rs780307724 Chr.1: 46612909-46612909 on Build GRCh38
rs763141334 Chr.1: 46613115-46613115 on Build GRCh38
rs1033804000 Chr.1: 46613048-46613048 on Build GRCh38
rs778883520 Chr.1: 46613409-46613409 on Build GRCh38
rs111448646 Chr.1: 46613026-46613026 on Build GRCh38
rs748481466 Chr.1: 46613016-46613016 on Build GRCh38
rs749136671 Chr.1: 46613395-46613395 on Build GRCh38
rs777449352 Chr.1: 46613264-46613264 on Build GRCh38
rs867445391 Chr.1: 46613086-46613086 on Build GRCh38
rs770800766 Chr.1: 46613168-46613168 on Build GRCh38
rs748324401 Chr.1: 46613010-46613010 on Build GRCh38
rs762935989 Chr.1: 46613204-46613204 on Build GRCh38
rs560363036 Chr.1: 46613043-46613043 on Build GRCh38
rs752915168 Chr.1: 46613049-46613049 on Build GRCh38
rs4660947 Chr.1: 46612965-46612965 on Build GRCh38
rs771683063 Chr.1: 46613150-46613150 on Build GRCh38
rs766781944 Chr.1: 46613356-46613356 on Build GRCh38
rs745537437 Chr.1: 46613387-46613387 on Build GRCh38
rs539615481 Chr.1: 46612956-46612956 on Build GRCh38
rs747738229 Chr.1: 46613085-46613085 on Build GRCh38
rs754252369 Chr.1: 46613367-46613367 on Build GRCh38
rs774620178 Chr.1: 46613394-46613395 on Build GRCh38
rs781039558 Chr.1: 46613271-46613271 on Build GRCh38
rs769596245 Chr.1: 46613200-46613200 on Build GRCh38
rs755169615 Chr.1: 46613002-46613002 on Build GRCh38
rs774441553 Chr.1: 46613316-46613316 on Build GRCh38
rs112472629 Chr.1: 46613090-46613090 on Build GRCh38
rs531580883 Chr.1: 46613283-46613283 on Build GRCh38
rs755804772 Chr.1: 46613391-46613391 on Build GRCh38
rs527607971 Chr.1: 46613070-46613070 on Build GRCh38
rs763048034 Chr.1: 46613282-46613283 on Build GRCh38
rs576489578 Chr.1: 46612974-46612974 on Build GRCh38
rs762041431 Chr.1: 46613210-46613210 on Build GRCh38
rs756939338 Chr.1: 46613269-46613269 on Build GRCh38
rs1006418561 Chr.1: 46613437-46613437 on Build GRCh38
rs372911972 Chr.1: 46612929-46612929 on Build GRCh38
rs750876270 Chr.1: 46613137-46613137 on Build GRCh38
rs772146205 Chr.1: 46613438-46613438 on Build GRCh38
rs538105910 Chr.1: 46613242-46613242 on Build GRCh38
rs760544426 Chr.1: 46613091-46613091 on Build GRCh38
rs759569065 Chr.1: 46613102-46613102 on Build GRCh38

View Full Product Details