Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80437061-80437660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENDOV
Entrez Gene ID
284131
Gene Name
endonuclease V
Gene Aliases
-
UniGene
Hs.389678
Gene Chromosome Location
Chr.17: 80415117-80438086 on Build GRCh38
Transcripts
NM_001164637.2 NM_173627.4 XM_011524658.2 XM_011524669.2 XM_011524670.1 XM_011524671.2 XM_017024511.1 XM_017024512.1 XM_017024514.1 XM_017024515.1 XM_017024516.1 XM_017024517.1 XM_017024518.1 XM_017024519.1 XM_017024520.1

SNP Detail

SNP ID Chromosome Location
rs372134225 Chr.17: 80437304-80437304 on Build GRCh38
rs41303574 Chr.17: 80437620-80437620 on Build GRCh38
rs566990942 Chr.17: 80437442-80437442 on Build GRCh38
rs377218883 Chr.17: 80437531-80437531 on Build GRCh38
rs901651279 Chr.17: 80437372-80437372 on Build GRCh38
rs11870849 Chr.17: 80437273-80437273 on Build GRCh38
rs920535411 Chr.17: 80437584-80437584 on Build GRCh38
rs372922880 Chr.17: 80437333-80437333 on Build GRCh38
rs940072650 Chr.17: 80437407-80437407 on Build GRCh38
rs1000378664 Chr.17: 80437498-80437498 on Build GRCh38
rs574847891 Chr.17: 80437569-80437569 on Build GRCh38
rs920517006 Chr.17: 80437389-80437389 on Build GRCh38
rs747871712 Chr.17: 80437616-80437616 on Build GRCh38
rs746590415 Chr.17: 80437544-80437544 on Build GRCh38
rs898956193 Chr.17: 80437145-80437145 on Build GRCh38
rs368088358 Chr.17: 80437303-80437303 on Build GRCh38
rs1034561208 Chr.17: 80437513-80437514 on Build GRCh38
rs878921822 Chr.17: 80437279-80437279 on Build GRCh38
rs980466871 Chr.17: 80437387-80437387 on Build GRCh38
rs567759625 Chr.17: 80437322-80437323 on Build GRCh38
rs997465990 Chr.17: 80437285-80437285 on Build GRCh38
rs907680841 Chr.17: 80437628-80437628 on Build GRCh38
rs546769582 Chr.17: 80437332-80437332 on Build GRCh38
rs955751542 Chr.17: 80437330-80437330 on Build GRCh38
rs549382908 Chr.17: 80437612-80437612 on Build GRCh38
rs887203562 Chr.17: 80437400-80437400 on Build GRCh38
rs776279919 Chr.17: 80437583-80437583 on Build GRCh38
rs8066800 Chr.17: 80437130-80437130 on Build GRCh38
rs567163532 Chr.17: 80437428-80437428 on Build GRCh38
rs200909407 Chr.17: 80437243-80437243 on Build GRCh38
rs28641025 Chr.17: 80437199-80437199 on Build GRCh38
rs9909067 Chr.17: 80437408-80437408 on Build GRCh38
rs191128945 Chr.17: 80437423-80437423 on Build GRCh38
rs1048497419 Chr.17: 80437153-80437153 on Build GRCh38
rs41303572 Chr.17: 80437545-80437545 on Build GRCh38
rs1031448550 Chr.17: 80437316-80437316 on Build GRCh38
rs534443838 Chr.17: 80437469-80437469 on Build GRCh38
rs530437237 Chr.17: 80437410-80437410 on Build GRCh38
rs41303570 Chr.17: 80437134-80437134 on Build GRCh38
rs528229376 Chr.17: 80437254-80437254 on Build GRCh38
rs866597302 Chr.17: 80437582-80437582 on Build GRCh38
rs994679157 Chr.17: 80437148-80437148 on Build GRCh38
rs930530433 Chr.17: 80437608-80437608 on Build GRCh38
rs564672951 Chr.17: 80437125-80437125 on Build GRCh38
rs116748335 Chr.17: 80437530-80437530 on Build GRCh38
rs1047538598 Chr.17: 80437394-80437394 on Build GRCh38
rs956144328 Chr.17: 80437203-80437203 on Build GRCh38
rs907621745 Chr.17: 80437327-80437327 on Build GRCh38
rs148451337 Chr.17: 80437436-80437437 on Build GRCh38
rs778105483 Chr.17: 80437294-80437294 on Build GRCh38
rs971991537 Chr.17: 80437266-80437266 on Build GRCh38
rs916868643 Chr.17: 80437383-80437383 on Build GRCh38
rs758918125 Chr.17: 80437190-80437190 on Build GRCh38
rs558864955 Chr.17: 80437502-80437502 on Build GRCh38
rs560750468 Chr.17: 80437249-80437249 on Build GRCh38
rs989373494 Chr.17: 80437242-80437242 on Build GRCh38
rs930509031 Chr.17: 80437391-80437391 on Build GRCh38
rs887182298 Chr.17: 80437197-80437197 on Build GRCh38
rs904760860 Chr.17: 80437443-80437443 on Build GRCh38
rs757756634 Chr.17: 80437384-80437384 on Build GRCh38
rs550984968 Chr.17: 80437317-80437317 on Build GRCh38
rs931834026 Chr.17: 80437585-80437585 on Build GRCh38
rs147575863 Chr.17: 80437622-80437622 on Build GRCh38
rs1031820004 Chr.17: 80437511-80437511 on Build GRCh38
rs1004195634 Chr.17: 80437198-80437198 on Build GRCh38
rs952331583 Chr.17: 80437515-80437515 on Build GRCh38
rs538321155 Chr.17: 80437543-80437543 on Build GRCh38

View Full Product Details