Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 116655688-116656227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3H
Entrez Gene ID
8667
Gene Name
eukaryotic translation initiation factor 3 subunit H
Gene Aliases
EIF3S3, eIF3-gamma, eIF3-p40
UniGene
Hs.492599
Gene Chromosome Location
Chr.8: 116644816-116755823 on Build GRCh38
Transcripts
NM_003756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7203701 c.595G>T p.E199* Chr.8: 116655968-116655968 on Build GRCh38
5644471 c.567delA p.A190fs*15 Chr.8: 116655996-116655996 on Build GRCh38
7200608 c.649G>A p.E217K Chr.8: 116655914-116655914 on Build GRCh38
6862051 c.615T>G p.I205M Chr.8: 116655948-116655948 on Build GRCh38
6652862 c.615T>C p.I205I Chr.8: 116655948-116655948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054991523 Chr.8: 116656091-116656091 on Build GRCh38
rs761373388 Chr.8: 116655873-116655873 on Build GRCh38
rs1021182788 Chr.8: 116655748-116655748 on Build GRCh38
rs200317577 Chr.8: 116655957-116655957 on Build GRCh38
rs543605604 Chr.8: 116655774-116655774 on Build GRCh38
rs771648274 Chr.8: 116655986-116655986 on Build GRCh38
rs770132800 Chr.8: 116655974-116655974 on Build GRCh38
rs1021571184 Chr.8: 116656126-116656126 on Build GRCh38
rs529322944 Chr.8: 116656058-116656061 on Build GRCh38
rs6469657 Chr.8: 116656150-116656150 on Build GRCh38
rs766220038 Chr.8: 116655893-116655893 on Build GRCh38
rs763423573 Chr.8: 116655984-116655984 on Build GRCh38
rs943860758 Chr.8: 116655840-116655840 on Build GRCh38
rs746070849 Chr.8: 116656052-116656052 on Build GRCh38
rs767583263 Chr.8: 116656028-116656030 on Build GRCh38
rs776029537 Chr.8: 116655981-116655981 on Build GRCh38
rs369010403 Chr.8: 116656030-116656030 on Build GRCh38
rs772548324 Chr.8: 116655987-116655987 on Build GRCh38
rs753526576 Chr.8: 116655999-116655999 on Build GRCh38
rs1020237484 Chr.8: 116655779-116655779 on Build GRCh38
rs753521440 Chr.8: 116655912-116655912 on Build GRCh38
rs768685365 Chr.8: 116656039-116656039 on Build GRCh38
rs139268505 Chr.8: 116655879-116655879 on Build GRCh38
rs759452229 Chr.8: 116656000-116656000 on Build GRCh38
rs1032005473 Chr.8: 116656163-116656163 on Build GRCh38
rs985535088 Chr.8: 116656177-116656177 on Build GRCh38
rs367678431 Chr.8: 116656148-116656148 on Build GRCh38
rs985243193 Chr.8: 116655803-116655803 on Build GRCh38
rs765881862 Chr.8: 116655995-116655995 on Build GRCh38
rs183745245 Chr.8: 116655758-116655758 on Build GRCh38
rs918496213 Chr.8: 116655898-116655898 on Build GRCh38
rs771153235 Chr.8: 116656126-116656131 on Build GRCh38
rs779048876 Chr.8: 116655917-116655917 on Build GRCh38
rs757351878 Chr.8: 116656038-116656038 on Build GRCh38
rs893260413 Chr.8: 116656108-116656108 on Build GRCh38
rs946125051 Chr.8: 116656122-116656122 on Build GRCh38
rs953662128 Chr.8: 116656175-116656175 on Build GRCh38
rs553537490 Chr.8: 116656098-116656098 on Build GRCh38
rs927507220 Chr.8: 116656059-116656059 on Build GRCh38
rs532675105 Chr.8: 116655886-116655886 on Build GRCh38
rs750514295 Chr.8: 116656030-116656034 on Build GRCh38
rs751523693 Chr.8: 116656034-116656034 on Build GRCh38
rs375917291 Chr.8: 116656019-116656019 on Build GRCh38
rs760046708 Chr.8: 116655836-116655836 on Build GRCh38
rs747273145 Chr.8: 116655940-116655940 on Build GRCh38
rs1041728159 Chr.8: 116656126-116656126 on Build GRCh38
rs781329328 Chr.8: 116656047-116656047 on Build GRCh38
rs57306258 Chr.8: 116656189-116656189 on Build GRCh38
rs1001159448 Chr.8: 116656161-116656161 on Build GRCh38
rs372658117 Chr.8: 116656016-116656016 on Build GRCh38
rs570161833 Chr.8: 116656194-116656194 on Build GRCh38
rs758589853 Chr.8: 116655928-116655928 on Build GRCh38
rs983150378 Chr.8: 116655935-116655935 on Build GRCh38
rs770187852 Chr.8: 116656057-116656057 on Build GRCh38
rs756370847 Chr.8: 116656042-116656043 on Build GRCh38
rs964590456 Chr.8: 116655852-116655852 on Build GRCh38
rs748075330 Chr.8: 116655925-116655925 on Build GRCh38
rs754971614 Chr.8: 116655915-116655915 on Build GRCh38
rs912310614 Chr.8: 116655813-116655813 on Build GRCh38
rs565608381 Chr.8: 116655848-116655848 on Build GRCh38
rs924965913 Chr.8: 116655773-116655773 on Build GRCh38
rs1008157758 Chr.8: 116656155-116656155 on Build GRCh38
rs755682334 Chr.8: 116655890-116655890 on Build GRCh38
rs746296103 Chr.8: 116655958-116655958 on Build GRCh38
rs369635660 Chr.8: 116655870-116655870 on Build GRCh38
rs760390298 Chr.8: 116655991-116655991 on Build GRCh38
rs951886355 Chr.8: 116655903-116655903 on Build GRCh38
rs777674913 Chr.8: 116655929-116655929 on Build GRCh38
rs1054558310 Chr.8: 116655751-116655751 on Build GRCh38
rs866274354 Chr.8: 116655953-116655953 on Build GRCh38
rs377471784 Chr.8: 116656043-116656043 on Build GRCh38
rs765092349 Chr.8: 116656004-116656004 on Build GRCh38
rs1018755622 Chr.8: 116655816-116655816 on Build GRCh38
rs966629857 Chr.8: 116655790-116655790 on Build GRCh38
rs201605498 Chr.8: 116656017-116656017 on Build GRCh38
rs1008754329 Chr.8: 116655796-116655796 on Build GRCh38
rs937532233 Chr.8: 116656071-116656071 on Build GRCh38
rs770997701 Chr.8: 116655947-116655947 on Build GRCh38
rs530063034 Chr.8: 116656184-116656184 on Build GRCh38

View Full Product Details