Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229460677-229461262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP133
Entrez Gene ID
55746
Gene Name
nucleoporin 133
Gene Aliases
hNUP133
UniGene
Hs.12457
Gene Chromosome Location
Chr.1: 229440995-229508343 on Build GRCh38
Transcripts
NM_018230.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6234108 c.2736C>A p.G912G Chr.1: 229460719-229460719 on Build GRCh38
679178 c.2745A>T p.L915F Chr.1: 229460710-229460710 on Build GRCh38
679177 c.2686-1G>C p.? Chr.1: 229460770-229460770 on Build GRCh38
5993458 c.2708G>A p.R903H Chr.1: 229460747-229460747 on Build GRCh38
4434942 c.2763G>C p.Q921H Chr.1: 229460692-229460692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047519885 Chr.1: 229461047-229461047 on Build GRCh38
rs970794172 Chr.1: 229460989-229460989 on Build GRCh38
rs773201846 Chr.1: 229460803-229460803 on Build GRCh38
rs375137512 Chr.1: 229460749-229460749 on Build GRCh38
rs1005740210 Chr.1: 229461058-229461058 on Build GRCh38
rs1024939548 Chr.1: 229461130-229461130 on Build GRCh38
rs77564806 Chr.1: 229460785-229460785 on Build GRCh38
rs760829745 Chr.1: 229460884-229460884 on Build GRCh38
rs1025215512 Chr.1: 229461232-229461232 on Build GRCh38
rs746752161 Chr.1: 229460768-229460768 on Build GRCh38
rs895342432 Chr.1: 229461206-229461206 on Build GRCh38
rs1010298229 Chr.1: 229460836-229460836 on Build GRCh38
rs151070299 Chr.1: 229461114-229461114 on Build GRCh38
rs781740868 Chr.1: 229460723-229460723 on Build GRCh38
rs74971830 Chr.1: 229460818-229460818 on Build GRCh38
rs201287976 Chr.1: 229460874-229460874 on Build GRCh38
rs971394199 Chr.1: 229461233-229461233 on Build GRCh38
rs548135501 Chr.1: 229461010-229461010 on Build GRCh38
rs370456239 Chr.1: 229460816-229460816 on Build GRCh38
rs145861862 Chr.1: 229460744-229460744 on Build GRCh38
rs531662081 Chr.1: 229460883-229460883 on Build GRCh38
rs959503049 Chr.1: 229460942-229460942 on Build GRCh38
rs1012368503 Chr.1: 229461221-229461221 on Build GRCh38
rs115803241 Chr.1: 229461037-229461037 on Build GRCh38
rs879411980 Chr.1: 229460956-229460956 on Build GRCh38
rs1026064692 Chr.1: 229460772-229460772 on Build GRCh38
rs764575185 Chr.1: 229460709-229460711 on Build GRCh38
rs959120144 Chr.1: 229461100-229461100 on Build GRCh38
rs577248195 Chr.1: 229461231-229461231 on Build GRCh38
rs943191856 Chr.1: 229461237-229461237 on Build GRCh38
rs1034049346 Chr.1: 229461193-229461193 on Build GRCh38
rs765335036 Chr.1: 229460802-229460802 on Build GRCh38
rs766818317 Chr.1: 229460712-229460712 on Build GRCh38
rs763261914 Chr.1: 229460804-229460804 on Build GRCh38
rs951528592 Chr.1: 229460800-229460800 on Build GRCh38
rs537512193 Chr.1: 229461116-229461116 on Build GRCh38
rs887496234 Chr.1: 229461054-229461054 on Build GRCh38
rs771782173 Chr.1: 229460748-229460748 on Build GRCh38
rs140279934 Chr.1: 229461148-229461148 on Build GRCh38
rs993122019 Chr.1: 229461038-229461038 on Build GRCh38
rs559994210 Chr.1: 229460722-229460722 on Build GRCh38
rs7545077 Chr.1: 229461004-229461004 on Build GRCh38
rs527670118 Chr.1: 229461007-229461007 on Build GRCh38
rs918150837 Chr.1: 229460985-229460985 on Build GRCh38
rs556437237 Chr.1: 229461086-229461086 on Build GRCh38
rs776588126 Chr.1: 229460787-229460787 on Build GRCh38
rs924719041 Chr.1: 229461157-229461157 on Build GRCh38
rs145595619 Chr.1: 229460706-229460706 on Build GRCh38
rs971801741 Chr.1: 229460894-229460894 on Build GRCh38
rs990178612 Chr.1: 229461204-229461204 on Build GRCh38
rs977478649 Chr.1: 229460920-229460920 on Build GRCh38
rs144996756 Chr.1: 229460714-229460714 on Build GRCh38
rs182773980 Chr.1: 229460996-229460996 on Build GRCh38
rs901086287 Chr.1: 229461089-229461089 on Build GRCh38
rs924666621 Chr.1: 229460934-229460934 on Build GRCh38
rs762406885 Chr.1: 229460747-229460747 on Build GRCh38
rs957476798 Chr.1: 229461158-229461158 on Build GRCh38
rs754240190 Chr.1: 229460815-229460816 on Build GRCh38
rs1041005534 Chr.1: 229461084-229461084 on Build GRCh38
rs1017132368 Chr.1: 229461067-229461067 on Build GRCh38
rs750741808 Chr.1: 229460846-229460846 on Build GRCh38
rs748634904 Chr.1: 229460724-229460724 on Build GRCh38
rs1040952757 Chr.1: 229460808-229460809 on Build GRCh38
rs761674637 Chr.1: 229460789-229460789 on Build GRCh38
rs563009439 Chr.1: 229460847-229460847 on Build GRCh38
rs187104042 Chr.1: 229461011-229461011 on Build GRCh38
rs766764132 Chr.1: 229460805-229460805 on Build GRCh38
rs375959042 Chr.1: 229460997-229460997 on Build GRCh38
rs778947219 Chr.1: 229461139-229461139 on Build GRCh38

View Full Product Details