Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105512543-105512970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSD1L
Entrez Gene ID
83856
Gene Name
fibronectin type III and SPRY domain containing 1 like
Gene Aliases
CCDC10, CSDUFD1, FSD1CL, FSD1NL, MIR1
UniGene
Hs.136901
Gene Chromosome Location
Chr.9: 105442183-105552433 on Build GRCh38
Transcripts
NM_001145313.2 NM_001287191.1 NM_001287192.1 XM_005252254.2 XM_005252257.3 XM_005252260.3 XM_011519077.2 XM_011519078.2 XM_011519079.2 XM_011519080.2 XM_011519081.2 XM_017015182.1 XM_017015183.1 XM_017015184.1 XM_017015185.1 XM_017015186.1 XM_017015187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4759956 c.941A>C p.K314T Chr.9: 105512852-105512852 on Build GRCh38
6876960 c.936C>T p.N312N Chr.9: 105512847-105512847 on Build GRCh38
6472806 c.471A>C p.K157N Chr.9: 105512853-105512853 on Build GRCh38
6898645 c.943G>A p.V315I Chr.9: 105512854-105512854 on Build GRCh38
5700947 c.442G>T p.D148Y Chr.9: 105512824-105512824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038756390 Chr.9: 105512909-105512913 on Build GRCh38
rs902652792 Chr.9: 105512590-105512590 on Build GRCh38
rs796543803 Chr.9: 105512733-105512735 on Build GRCh38
rs973266317 Chr.9: 105512797-105512797 on Build GRCh38
rs182209648 Chr.9: 105512892-105512892 on Build GRCh38
rs991831541 Chr.9: 105512912-105512912 on Build GRCh38
rs1052478377 Chr.9: 105512872-105512872 on Build GRCh38
rs920493182 Chr.9: 105512881-105512881 on Build GRCh38
rs1015018787 Chr.9: 105512742-105512742 on Build GRCh38
rs148753022 Chr.9: 105512714-105512714 on Build GRCh38
rs980866995 Chr.9: 105512736-105512736 on Build GRCh38
rs199966291 Chr.9: 105512732-105512733 on Build GRCh38
rs999649284 Chr.9: 105512622-105512622 on Build GRCh38
rs989488713 Chr.9: 105512784-105512784 on Build GRCh38
rs370320917 Chr.9: 105512615-105512615 on Build GRCh38
rs527550486 Chr.9: 105512770-105512770 on Build GRCh38
rs142680826 Chr.9: 105512736-105512738 on Build GRCh38
rs961871783 Chr.9: 105512776-105512776 on Build GRCh38
rs550652063 Chr.9: 105512865-105512865 on Build GRCh38
rs914117096 Chr.9: 105512844-105512844 on Build GRCh38
rs113815098 Chr.9: 105512649-105512649 on Build GRCh38
rs954888008 Chr.9: 105512685-105512685 on Build GRCh38
rs10820812 Chr.9: 105512650-105512650 on Build GRCh38
rs958672864 Chr.9: 105512905-105512905 on Build GRCh38
rs1008953180 Chr.9: 105512718-105512718 on Build GRCh38
rs994479292 Chr.9: 105512578-105512578 on Build GRCh38
rs1054440824 Chr.9: 105512643-105512643 on Build GRCh38
rs898814876 Chr.9: 105512571-105512571 on Build GRCh38
rs147270052 Chr.9: 105512678-105512678 on Build GRCh38

View Full Product Details