Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 31808471-31808948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP12
Entrez Gene ID
94134
Gene Name
Rho GTPase activating protein 12
Gene Aliases
-
UniGene
Hs.499264
Gene Chromosome Location
Chr.10: 31805398-31928876 on Build GRCh38
Transcripts
NM_001270695.1 NM_001270696.1 NM_001270697.1 NM_001270698.1 NM_001270699.1 NM_018287.6 XM_005252644.1 XM_011519761.1 XM_017016954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5927571 c.2366+8G>A p.? Chr.10: 31808641-31808641 on Build GRCh38
5836803 c.2281C>T p.R761* Chr.10: 31808734-31808734 on Build GRCh38
5616450 c.2352C>T p.F784F Chr.10: 31808663-31808663 on Build GRCh38
3414982 c.2347C>A p.L783I Chr.10: 31808668-31808668 on Build GRCh38
3790752 c.2309G>T p.R770I Chr.10: 31808706-31808706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374142977 Chr.10: 31808874-31808874 on Build GRCh38
rs763583114 Chr.10: 31808716-31808716 on Build GRCh38
rs2183959 Chr.10: 31808554-31808554 on Build GRCh38
rs775764085 Chr.10: 31808645-31808645 on Build GRCh38
rs112005286 Chr.10: 31808922-31808923 on Build GRCh38
rs143564246 Chr.10: 31808681-31808681 on Build GRCh38
rs923800626 Chr.10: 31808806-31808806 on Build GRCh38
rs565744387 Chr.10: 31808555-31808555 on Build GRCh38
rs747014050 Chr.10: 31808651-31808651 on Build GRCh38
rs369784094 Chr.10: 31808904-31808904 on Build GRCh38
rs1039299138 Chr.10: 31808727-31808727 on Build GRCh38
rs566230206 Chr.10: 31808837-31808837 on Build GRCh38
rs796754040 Chr.10: 31808549-31808549 on Build GRCh38
rs752212763 Chr.10: 31808611-31808611 on Build GRCh38
rs750180294 Chr.10: 31808734-31808734 on Build GRCh38
rs753706224 Chr.10: 31808629-31808629 on Build GRCh38
rs370969355 Chr.10: 31808622-31808622 on Build GRCh38
rs758356681 Chr.10: 31808762-31808762 on Build GRCh38
rs770584251 Chr.10: 31808643-31808643 on Build GRCh38
rs759296561 Chr.10: 31808769-31808769 on Build GRCh38
rs1026423086 Chr.10: 31808866-31808866 on Build GRCh38
rs895851933 Chr.10: 31808594-31808594 on Build GRCh38
rs974859051 Chr.10: 31808741-31808741 on Build GRCh38
rs948753731 Chr.10: 31808891-31808891 on Build GRCh38
rs763492032 Chr.10: 31808608-31808608 on Build GRCh38
rs970426481 Chr.10: 31808917-31808918 on Build GRCh38
rs923872649 Chr.10: 31808513-31808513 on Build GRCh38
rs1029077 Chr.10: 31808878-31808878 on Build GRCh38
rs199703275 Chr.10: 31808776-31808776 on Build GRCh38
rs554499461 Chr.10: 31808577-31808577 on Build GRCh38
rs200866580 Chr.10: 31808634-31808634 on Build GRCh38
rs982436845 Chr.10: 31808498-31808498 on Build GRCh38
rs112095318 Chr.10: 31808858-31808858 on Build GRCh38
rs899292963 Chr.10: 31808626-31808626 on Build GRCh38
rs939260072 Chr.10: 31808829-31808829 on Build GRCh38
rs1029078 Chr.10: 31808923-31808923 on Build GRCh38
rs34750454 Chr.10: 31808722-31808722 on Build GRCh38
rs557723340 Chr.10: 31808541-31808541 on Build GRCh38
rs780140801 Chr.10: 31808646-31808646 on Build GRCh38
rs560222995 Chr.10: 31808733-31808733 on Build GRCh38
rs760070815 Chr.10: 31808726-31808726 on Build GRCh38
rs368128317 Chr.10: 31808696-31808696 on Build GRCh38
rs915005712 Chr.10: 31808742-31808742 on Build GRCh38
rs771513913 Chr.10: 31808718-31808718 on Build GRCh38
rs763630795 Chr.10: 31808624-31808624 on Build GRCh38
rs182167975 Chr.10: 31808918-31808918 on Build GRCh38
rs995055508 Chr.10: 31808839-31808839 on Build GRCh38
rs770467452 Chr.10: 31808600-31808600 on Build GRCh38
rs926496474 Chr.10: 31808590-31808590 on Build GRCh38
rs946573605 Chr.10: 31808500-31808500 on Build GRCh38
rs774153319 Chr.10: 31808701-31808701 on Build GRCh38
rs768974025 Chr.10: 31808661-31808661 on Build GRCh38
rs779947110 Chr.10: 31808770-31808770 on Build GRCh38
rs377648203 Chr.10: 31808601-31808601 on Build GRCh38
rs561375398 Chr.10: 31808890-31808890 on Build GRCh38
rs754275026 Chr.10: 31808728-31808728 on Build GRCh38
rs773641302 Chr.10: 31808713-31808713 on Build GRCh38
rs768672269 Chr.10: 31808862-31808863 on Build GRCh38
rs748319958 Chr.10: 31808777-31808777 on Build GRCh38
rs576654881 Chr.10: 31808673-31808673 on Build GRCh38
rs374026928 Chr.10: 31808679-31808679 on Build GRCh38
rs371681711 Chr.10: 31808773-31808773 on Build GRCh38
rs970771782 Chr.10: 31808510-31808510 on Build GRCh38
rs536260447 Chr.10: 31808568-31808568 on Build GRCh38
rs11008654 Chr.10: 31808506-31808506 on Build GRCh38
rs1024640747 Chr.10: 31808894-31808894 on Build GRCh38
rs558555642 Chr.10: 31808644-31808644 on Build GRCh38

View Full Product Details