Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178612524-178612982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK4
Entrez Gene ID
57396
Gene Name
CDC like kinase 4
Gene Aliases
-
UniGene
Hs.406557
Gene Chromosome Location
Chr.5: 178602664-178627053 on Build GRCh38
Transcripts
NM_020666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6417359 c.933A>T p.E311D Chr.5: 178612534-178612534 on Build GRCh38
4763674 c.931G>A p.E311K Chr.5: 178612536-178612536 on Build GRCh38
239346 c.919A>C p.M307L Chr.5: 178612798-178612798 on Build GRCh38
5632018 c.903C>T p.V301V Chr.5: 178612814-178612814 on Build GRCh38
1436398 c.856G>A p.D286N Chr.5: 178612861-178612861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765519971 Chr.5: 178612589-178612589 on Build GRCh38
rs749028378 Chr.5: 178612812-178612812 on Build GRCh38
rs993207536 Chr.5: 178612905-178612905 on Build GRCh38
rs142116327 Chr.5: 178612729-178612729 on Build GRCh38
rs996660041 Chr.5: 178612675-178612675 on Build GRCh38
rs553798661 Chr.5: 178612715-178612715 on Build GRCh38
rs753131469 Chr.5: 178612574-178612574 on Build GRCh38
rs770737225 Chr.5: 178612710-178612710 on Build GRCh38
rs780656831 Chr.5: 178612780-178612780 on Build GRCh38
rs1003901035 Chr.5: 178612690-178612690 on Build GRCh38
rs961418546 Chr.5: 178612885-178612885 on Build GRCh38
rs889131793 Chr.5: 178612880-178612880 on Build GRCh38
rs775964888 Chr.5: 178612872-178612872 on Build GRCh38
rs757326227 Chr.5: 178612933-178612934 on Build GRCh38
rs537354001 Chr.5: 178612786-178612786 on Build GRCh38
rs371635907 Chr.5: 178612918-178612918 on Build GRCh38
rs550008874 Chr.5: 178612651-178612651 on Build GRCh38
rs368574938 Chr.5: 178612597-178612597 on Build GRCh38
rs776517203 Chr.5: 178612886-178612886 on Build GRCh38
rs187298625 Chr.5: 178612664-178612664 on Build GRCh38
rs754821226 Chr.5: 178612548-178612549 on Build GRCh38
rs772872790 Chr.5: 178612558-178612558 on Build GRCh38
rs747152188 Chr.5: 178612912-178612913 on Build GRCh38
rs761312502 Chr.5: 178612577-178612577 on Build GRCh38
rs752716593 Chr.5: 178612833-178612836 on Build GRCh38
rs764650590 Chr.5: 178612877-178612877 on Build GRCh38
rs761067456 Chr.5: 178612874-178612874 on Build GRCh38
rs1045464835 Chr.5: 178612668-178612668 on Build GRCh38
rs915597328 Chr.5: 178612647-178612647 on Build GRCh38
rs374707856 Chr.5: 178612595-178612595 on Build GRCh38
rs372278919 Chr.5: 178612555-178612555 on Build GRCh38
rs747606535 Chr.5: 178612783-178612783 on Build GRCh38
rs140479491 Chr.5: 178612626-178612628 on Build GRCh38
rs754981854 Chr.5: 178612775-178612775 on Build GRCh38
rs777747156 Chr.5: 178612911-178612913 on Build GRCh38
rs758067420 Chr.5: 178612754-178612754 on Build GRCh38
rs899301247 Chr.5: 178612723-178612723 on Build GRCh38
rs777312370 Chr.5: 178612908-178612908 on Build GRCh38
rs1008938535 Chr.5: 178612893-178612893 on Build GRCh38
rs886892167 Chr.5: 178612687-178612687 on Build GRCh38
rs769324789 Chr.5: 178612784-178612784 on Build GRCh38
rs531874880 Chr.5: 178612622-178612622 on Build GRCh38
rs746913861 Chr.5: 178612762-178612762 on Build GRCh38
rs1029622635 Chr.5: 178612742-178612742 on Build GRCh38
rs758310931 Chr.5: 178612901-178612902 on Build GRCh38
rs767939283 Chr.5: 178612570-178612570 on Build GRCh38
rs769484276 Chr.5: 178612553-178612553 on Build GRCh38
rs575941668 Chr.5: 178612587-178612587 on Build GRCh38
rs758716998 Chr.5: 178612939-178612939 on Build GRCh38
rs984230405 Chr.5: 178612827-178612827 on Build GRCh38
rs764052144 Chr.5: 178612581-178612581 on Build GRCh38
rs535208624 Chr.5: 178612678-178612678 on Build GRCh38
rs759779799 Chr.5: 178612563-178612563 on Build GRCh38
rs952832917 Chr.5: 178612704-178612704 on Build GRCh38
rs111966205 Chr.5: 178612943-178612943 on Build GRCh38
rs772320387 Chr.5: 178612835-178612835 on Build GRCh38
rs764878312 Chr.5: 178612579-178612579 on Build GRCh38
rs779954047 Chr.5: 178612760-178612760 on Build GRCh38
rs765416672 Chr.5: 178612916-178612916 on Build GRCh38

View Full Product Details