Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25861436-25862030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A3
Entrez Gene ID
10786
Gene Name
solute carrier family 17 member 3
Gene Aliases
GOUT4, NPT4, UAQTL4
UniGene
Hs.327179
Gene Chromosome Location
Chr.6: 25845100-25874243 on Build GRCh38
Transcripts
NM_001098486.1 NM_006632.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3777377 c.378C>A p.S126R Chr.6: 25861637-25861637 on Build GRCh38
6197228 c.308C>A p.S103* Chr.6: 25861707-25861707 on Build GRCh38
6197227 c.542C>A p.S181* Chr.6: 25861707-25861707 on Build GRCh38
1685995 c.596G>A p.R199Q Chr.6: 25861653-25861653 on Build GRCh38
6208496 c.316G>T p.G106W Chr.6: 25861699-25861699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750151513 Chr.6: 25861879-25861879 on Build GRCh38
rs371848996 Chr.6: 25861729-25861729 on Build GRCh38
rs922632384 Chr.6: 25861480-25861480 on Build GRCh38
rs550429416 Chr.6: 25861994-25861994 on Build GRCh38
rs771182088 Chr.6: 25861575-25861575 on Build GRCh38
rs778539524 Chr.6: 25861657-25861657 on Build GRCh38
rs915616119 Chr.6: 25861791-25861791 on Build GRCh38
rs761390290 Chr.6: 25861976-25861976 on Build GRCh38
rs770866165 Chr.6: 25861710-25861729 on Build GRCh38
rs774878955 Chr.6: 25861809-25861809 on Build GRCh38
rs764515211 Chr.6: 25861695-25861695 on Build GRCh38
rs527389249 Chr.6: 25861750-25861750 on Build GRCh38
rs531315084 Chr.6: 25861696-25861696 on Build GRCh38
rs759603958 Chr.6: 25861579-25861579 on Build GRCh38
rs764920343 Chr.6: 25861842-25861842 on Build GRCh38
rs773437633 Chr.6: 25861831-25861831 on Build GRCh38
rs746751670 Chr.6: 25861668-25861668 on Build GRCh38
rs370924957 Chr.6: 25861999-25861999 on Build GRCh38
rs767800626 Chr.6: 25861699-25861699 on Build GRCh38
rs772108912 Chr.6: 25861660-25861660 on Build GRCh38
rs368693445 Chr.6: 25861725-25861725 on Build GRCh38
rs375918157 Chr.6: 25861839-25861839 on Build GRCh38
rs752413057 Chr.6: 25861844-25861844 on Build GRCh38
rs751026114 Chr.6: 25861973-25861973 on Build GRCh38
rs762186639 Chr.6: 25861697-25861697 on Build GRCh38
rs769999214 Chr.6: 25861762-25861762 on Build GRCh38
rs750810898 Chr.6: 25861971-25861971 on Build GRCh38
rs200669181 Chr.6: 25861722-25861722 on Build GRCh38
rs768740492 Chr.6: 25861806-25861806 on Build GRCh38
rs755859454 Chr.6: 25861989-25861989 on Build GRCh38
rs868307643 Chr.6: 25861942-25861942 on Build GRCh38
rs972648112 Chr.6: 25861718-25861718 on Build GRCh38
rs779696749 Chr.6: 25861649-25861649 on Build GRCh38
rs374516952 Chr.6: 25861858-25861858 on Build GRCh38
rs750378719 Chr.6: 25861988-25861988 on Build GRCh38
rs763545856 Chr.6: 25861853-25861853 on Build GRCh38
rs754494201 Chr.6: 25861654-25861654 on Build GRCh38
rs754657678 Chr.6: 25861711-25861711 on Build GRCh38
rs764641864 Chr.6: 25861608-25861608 on Build GRCh38
rs368452184 Chr.6: 25861686-25861686 on Build GRCh38
rs775692570 Chr.6: 25861765-25861765 on Build GRCh38
rs938931942 Chr.6: 25861772-25861772 on Build GRCh38
rs865972698 Chr.6: 25861965-25861965 on Build GRCh38
rs770048973 Chr.6: 25861674-25861674 on Build GRCh38
rs541465684 Chr.6: 25861715-25861715 on Build GRCh38
rs375777292 Chr.6: 25861818-25861818 on Build GRCh38
rs369858810 Chr.6: 25861852-25861852 on Build GRCh38
rs770646517 Chr.6: 25861669-25861669 on Build GRCh38
rs115841196 Chr.6: 25861667-25861667 on Build GRCh38
rs941535379 Chr.6: 25861640-25861640 on Build GRCh38
rs759495316 Chr.6: 25861614-25861614 on Build GRCh38
rs771444524 Chr.6: 25861955-25861955 on Build GRCh38
rs370989265 Chr.6: 25861872-25861872 on Build GRCh38
rs956028200 Chr.6: 25861826-25861826 on Build GRCh38
rs750842238 Chr.6: 25861701-25861701 on Build GRCh38
rs547545784 Chr.6: 25861819-25861819 on Build GRCh38
rs561444551 Chr.6: 25861723-25861723 on Build GRCh38
rs998672017 Chr.6: 25861907-25861907 on Build GRCh38
rs745718327 Chr.6: 25861753-25861753 on Build GRCh38
rs776724707 Chr.6: 25861578-25861578 on Build GRCh38
rs753679881 Chr.6: 25861995-25861995 on Build GRCh38
rs139166770 Chr.6: 25861635-25861635 on Build GRCh38
rs755144743 Chr.6: 25861996-25861996 on Build GRCh38
rs554009999 Chr.6: 25861533-25861533 on Build GRCh38
rs139396357 Chr.6: 25861684-25861684 on Build GRCh38
rs772456900 Chr.6: 25861932-25861932 on Build GRCh38
rs757038753 Chr.6: 25861733-25861733 on Build GRCh38
rs765547232 Chr.6: 25861951-25861951 on Build GRCh38
rs377627217 Chr.6: 25861860-25861860 on Build GRCh38
rs765323828 Chr.6: 25861591-25861591 on Build GRCh38
rs1049853296 Chr.6: 25861535-25861535 on Build GRCh38
rs374433559 Chr.6: 25861602-25861602 on Build GRCh38
rs749540605 Chr.6: 25861908-25861908 on Build GRCh38
rs755492216 Chr.6: 25861912-25861912 on Build GRCh38
rs1003804442 Chr.6: 25861655-25861655 on Build GRCh38
rs752813464 Chr.6: 25861599-25861599 on Build GRCh38
rs373256050 Chr.6: 25861788-25861788 on Build GRCh38
rs757564537 Chr.6: 25861629-25861629 on Build GRCh38
rs780197221 Chr.6: 25861906-25861906 on Build GRCh38
rs376946211 Chr.6: 25861719-25861719 on Build GRCh38
rs758400774 Chr.6: 25861665-25861665 on Build GRCh38
rs747711332 Chr.6: 25861658-25861658 on Build GRCh38
rs775245671 Chr.6: 25861961-25861961 on Build GRCh38
rs999098155 Chr.6: 25861510-25861510 on Build GRCh38
rs904397982 Chr.6: 25861475-25861475 on Build GRCh38
rs77220283 Chr.6: 25861473-25861473 on Build GRCh38
rs546093722 Chr.6: 25861671-25861671 on Build GRCh38
rs377515468 Chr.6: 25861653-25861653 on Build GRCh38
rs376483140 Chr.6: 25861983-25861983 on Build GRCh38
rs533432418 Chr.6: 25861924-25861924 on Build GRCh38
rs144108103 Chr.6: 25861911-25861911 on Build GRCh38

View Full Product Details