Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23523199-23523611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EARS2
Entrez Gene ID
124454
Gene Name
glutamyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD12, MSE1, gluRS
UniGene
Hs.620541
Gene Chromosome Location
Chr.16: 23522013-23557375 on Build GRCh38
Transcripts
NM_001083614.1 NR_003501.1 XM_011545738.1 XM_011545739.1

SNP Detail

SNP ID Chromosome Location
rs572499573 Chr.16: 23523263-23523263 on Build GRCh38
rs74014928 Chr.16: 23523272-23523272 on Build GRCh38
rs545493711 Chr.16: 23523218-23523218 on Build GRCh38
rs999311424 Chr.16: 23523472-23523472 on Build GRCh38
rs1051615451 Chr.16: 23523317-23523317 on Build GRCh38
rs765036019 Chr.16: 23523505-23523505 on Build GRCh38
rs769170999 Chr.16: 23523228-23523228 on Build GRCh38
rs536146161 Chr.16: 23523588-23523588 on Build GRCh38
rs1055101556 Chr.16: 23523408-23523408 on Build GRCh38
rs921565892 Chr.16: 23523568-23523568 on Build GRCh38
rs750087699 Chr.16: 23523323-23523323 on Build GRCh38
rs547094685 Chr.16: 23523535-23523535 on Build GRCh38
rs974391343 Chr.16: 23523565-23523565 on Build GRCh38
rs530431066 Chr.16: 23523311-23523311 on Build GRCh38
rs1051212183 Chr.16: 23523345-23523345 on Build GRCh38
rs899239616 Chr.16: 23523348-23523348 on Build GRCh38
rs990440410 Chr.16: 23523302-23523302 on Build GRCh38
rs868673991 Chr.16: 23523412-23523412 on Build GRCh38
rs940155820 Chr.16: 23523351-23523351 on Build GRCh38
rs994876338 Chr.16: 23523382-23523382 on Build GRCh38
rs1013470517 Chr.16: 23523439-23523439 on Build GRCh38
rs532554451 Chr.16: 23523435-23523435 on Build GRCh38
rs932873198 Chr.16: 23523344-23523344 on Build GRCh38
rs921535842 Chr.16: 23523307-23523307 on Build GRCh38
rs978242473 Chr.16: 23523226-23523226 on Build GRCh38
rs185800148 Chr.16: 23523457-23523457 on Build GRCh38
rs890257212 Chr.16: 23523319-23523319 on Build GRCh38
rs1037512117 Chr.16: 23523364-23523364 on Build GRCh38
rs762102458 Chr.16: 23523301-23523301 on Build GRCh38
rs957785655 Chr.16: 23523554-23523554 on Build GRCh38
rs143336824 Chr.16: 23523266-23523266 on Build GRCh38
rs1053856571 Chr.16: 23523242-23523242 on Build GRCh38
rs566896041 Chr.16: 23523547-23523547 on Build GRCh38
rs756961049 Chr.16: 23523454-23523454 on Build GRCh38
rs931875451 Chr.16: 23523312-23523312 on Build GRCh38
rs752712603 Chr.16: 23523589-23523589 on Build GRCh38
rs912688939 Chr.16: 23523350-23523350 on Build GRCh38
rs151312469 Chr.16: 23523405-23523405 on Build GRCh38
rs760812762 Chr.16: 23523470-23523470 on Build GRCh38
rs10239 Chr.16: 23523233-23523233 on Build GRCh38
rs767908220 Chr.16: 23523447-23523447 on Build GRCh38
rs140666015 Chr.16: 23523440-23523440 on Build GRCh38
rs544098976 Chr.16: 23523397-23523397 on Build GRCh38
rs1014892256 Chr.16: 23523427-23523427 on Build GRCh38
rs527366751 Chr.16: 23523458-23523458 on Build GRCh38

View Full Product Details