Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67317836-67318329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100130987
Entrez Gene ID
100130987
Gene Name
uncharacterized LOC100130987
Gene Aliases
-
UniGene
Hs.664585
Gene Chromosome Location
Chr.11: 67317839-67391687 on Build GRCh38
Transcripts
NR_024469.1

SNP Detail

SNP ID Chromosome Location
rs754502241 Chr.11: 67317922-67317922 on Build GRCh38
rs996236472 Chr.11: 67318118-67318118 on Build GRCh38
rs951670583 Chr.11: 67318019-67318019 on Build GRCh38
rs912771801 Chr.11: 67318065-67318065 on Build GRCh38
rs940698385 Chr.11: 67318070-67318070 on Build GRCh38
rs796518557 Chr.11: 67318287-67318288 on Build GRCh38
rs944192658 Chr.11: 67318077-67318077 on Build GRCh38
rs969100278 Chr.11: 67318282-67318282 on Build GRCh38
rs1054165315 Chr.11: 67317861-67317861 on Build GRCh38
rs994962175 Chr.11: 67318287-67318287 on Build GRCh38
rs1033409996 Chr.11: 67317936-67317936 on Build GRCh38
rs913856211 Chr.11: 67318143-67318143 on Build GRCh38
rs1049326048 Chr.11: 67318052-67318052 on Build GRCh38
rs369786018 Chr.11: 67318310-67318311 on Build GRCh38
rs140324789 Chr.11: 67318054-67318054 on Build GRCh38
rs980527669 Chr.11: 67318285-67318285 on Build GRCh38
rs1024814595 Chr.11: 67317908-67317908 on Build GRCh38
rs927419247 Chr.11: 67318310-67318310 on Build GRCh38
rs963067988 Chr.11: 67317971-67317971 on Build GRCh38
rs541870866 Chr.11: 67318013-67318013 on Build GRCh38
rs1026362160 Chr.11: 67318005-67318005 on Build GRCh38
rs1030917805 Chr.11: 67318151-67318151 on Build GRCh38
rs1054941715 Chr.11: 67318281-67318281 on Build GRCh38
rs552335184 Chr.11: 67318076-67318076 on Build GRCh38
rs368657649 Chr.11: 67318130-67318130 on Build GRCh38
rs1013375650 Chr.11: 67318194-67318194 on Build GRCh38
rs936732122 Chr.11: 67318104-67318104 on Build GRCh38
rs979387155 Chr.11: 67317880-67317880 on Build GRCh38
rs992275971 Chr.11: 67317900-67317900 on Build GRCh38
rs956403756 Chr.11: 67317894-67317894 on Build GRCh38
rs921363351 Chr.11: 67318089-67318089 on Build GRCh38
rs927461221 Chr.11: 67317941-67317941 on Build GRCh38
rs1009856972 Chr.11: 67317901-67317903 on Build GRCh38
rs1024668843 Chr.11: 67318203-67318203 on Build GRCh38
rs991812829 Chr.11: 67318198-67318198 on Build GRCh38
rs562664499 Chr.11: 67317873-67317873 on Build GRCh38
rs113203860 Chr.11: 67318176-67318176 on Build GRCh38
rs776558575 Chr.11: 67317962-67317962 on Build GRCh38
rs956871630 Chr.11: 67318196-67318196 on Build GRCh38
rs1001153654 Chr.11: 67318141-67318141 on Build GRCh38
rs903693654 Chr.11: 67318138-67318138 on Build GRCh38
rs575604025 Chr.11: 67317882-67317882 on Build GRCh38
rs564366630 Chr.11: 67318084-67318084 on Build GRCh38
rs560207373 Chr.11: 67318018-67318018 on Build GRCh38
rs1026014953 Chr.11: 67318292-67318292 on Build GRCh38
rs901918520 Chr.11: 67318110-67318110 on Build GRCh38
rs947812917 Chr.11: 67317934-67317934 on Build GRCh38
rs906387528 Chr.11: 67318225-67318225 on Build GRCh38
rs945347025 Chr.11: 67318182-67318182 on Build GRCh38
rs950433479 Chr.11: 67318297-67318297 on Build GRCh38
rs796887855 Chr.11: 67317988-67317988 on Build GRCh38
rs1002024666 Chr.11: 67318230-67318230 on Build GRCh38
rs968345780 Chr.11: 67317874-67317874 on Build GRCh38
rs1028755262 Chr.11: 67318137-67318137 on Build GRCh38
rs759274013 Chr.11: 67318029-67318029 on Build GRCh38
rs375725431 Chr.11: 67317937-67317937 on Build GRCh38

View Full Product Details