Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200713495-200714006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX3P-AOX2P
Entrez Gene ID
107161151
Gene Name
AOX3P-AOX2P readthrough
Gene Aliases
AOX3/AOX3L1
UniGene
Hs.148269
Gene Chromosome Location
Chr.2: 200695723-200794915 on Build GRCh38
Transcripts
NR_135011.1 NR_135012.1 NR_037886.1

Gene Symbol
LOC100507140
Entrez Gene ID
100507140
Gene Name
uncharacterized LOC100507140
Gene Aliases
-
UniGene
Hs.382116
Gene Chromosome Location
Chr.2: 200712305-200735177 on Build GRCh38
Transcripts
NR_135011.1 NR_135012.1 NR_037886.1

SNP Detail

SNP ID Chromosome Location
rs554432164 Chr.2: 200713727-200713727 on Build GRCh38
rs1022932174 Chr.2: 200713786-200713787 on Build GRCh38
rs117673341 Chr.2: 200713947-200713947 on Build GRCh38
rs1002949739 Chr.2: 200713799-200713800 on Build GRCh38
rs956214909 Chr.2: 200713820-200713820 on Build GRCh38
rs562782106 Chr.2: 200713634-200713634 on Build GRCh38
rs990698665 Chr.2: 200713833-200713833 on Build GRCh38
rs555854651 Chr.2: 200713746-200713746 on Build GRCh38
rs1013102979 Chr.2: 200713880-200713880 on Build GRCh38
rs897978510 Chr.2: 200713563-200713563 on Build GRCh38
rs981933459 Chr.2: 200713575-200713575 on Build GRCh38
rs943640907 Chr.2: 200713682-200713682 on Build GRCh38
rs989332932 Chr.2: 200713738-200713738 on Build GRCh38
rs893240523 Chr.2: 200713980-200713980 on Build GRCh38
rs746392557 Chr.2: 200713967-200713967 on Build GRCh38
rs1047970482 Chr.2: 200713761-200713761 on Build GRCh38
rs944433022 Chr.2: 200713554-200713554 on Build GRCh38
rs139255203 Chr.2: 200713696-200713696 on Build GRCh38
rs751401803 Chr.2: 200713644-200713644 on Build GRCh38
rs994971684 Chr.2: 200713576-200713576 on Build GRCh38
rs6707459 Chr.2: 200713756-200713756 on Build GRCh38
rs971462442 Chr.2: 200713681-200713681 on Build GRCh38
rs1012176611 Chr.2: 200713786-200713786 on Build GRCh38
rs188206721 Chr.2: 200713911-200713911 on Build GRCh38
rs149574422 Chr.2: 200713895-200713895 on Build GRCh38
rs77975636 Chr.2: 200713651-200713651 on Build GRCh38
rs1036150399 Chr.2: 200713558-200713558 on Build GRCh38
rs964892945 Chr.2: 200713903-200713903 on Build GRCh38
rs914754445 Chr.2: 200713875-200713875 on Build GRCh38
rs778033352 Chr.2: 200713667-200713667 on Build GRCh38
rs530115845 Chr.2: 200713670-200713670 on Build GRCh38
rs1032151173 Chr.2: 200713927-200713927 on Build GRCh38
rs866694197 Chr.2: 200713774-200713774 on Build GRCh38
rs1049231138 Chr.2: 200713800-200713800 on Build GRCh38
rs919014563 Chr.2: 200713791-200713791 on Build GRCh38
rs1031764817 Chr.2: 200713717-200713717 on Build GRCh38
rs571188214 Chr.2: 200713764-200713764 on Build GRCh38
rs930788600 Chr.2: 200713792-200713792 on Build GRCh38
rs550544199 Chr.2: 200713870-200713870 on Build GRCh38
rs911541821 Chr.2: 200713769-200713769 on Build GRCh38
rs754734047 Chr.2: 200713655-200713655 on Build GRCh38
rs558820239 Chr.2: 200713773-200713773 on Build GRCh38
rs998871589 Chr.2: 200713918-200713918 on Build GRCh38
rs1024713334 Chr.2: 200713664-200713664 on Build GRCh38
rs1032201856 Chr.2: 200713806-200713806 on Build GRCh38
rs150842333 Chr.2: 200713602-200713602 on Build GRCh38
rs911592730 Chr.2: 200713534-200713534 on Build GRCh38
rs894675318 Chr.2: 200713826-200713826 on Build GRCh38
rs772581855 Chr.2: 200713976-200713976 on Build GRCh38
rs1006493305 Chr.2: 200713765-200713765 on Build GRCh38

View Full Product Details