Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34182722-34183262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00310
Entrez Gene ID
114036
Gene Name
long intergenic non-protein coding RNA 310
Gene Aliases
C21orf82, NCRNA00310
UniGene
Hs.720230
Gene Chromosome Location
Chr.21: 34180677-34189919 on Build GRCh38
Transcripts
NR_027266.1 NR_027267.1

SNP Detail

SNP ID Chromosome Location
rs900772947 Chr.21: 34183208-34183208 on Build GRCh38
rs773167302 Chr.21: 34183094-34183094 on Build GRCh38
rs534233693 Chr.21: 34183215-34183215 on Build GRCh38
rs368419294 Chr.21: 34183216-34183216 on Build GRCh38
rs963230765 Chr.21: 34183019-34183019 on Build GRCh38
rs965318594 Chr.21: 34182928-34182928 on Build GRCh38
rs997807313 Chr.21: 34182929-34182929 on Build GRCh38
rs186169548 Chr.21: 34182978-34182978 on Build GRCh38
rs1016007371 Chr.21: 34182965-34182965 on Build GRCh38
rs989555061 Chr.21: 34183109-34183109 on Build GRCh38
rs191427987 Chr.21: 34182991-34182991 on Build GRCh38
rs73198789 Chr.21: 34182789-34182789 on Build GRCh38
rs1032619737 Chr.21: 34182927-34182927 on Build GRCh38
rs184068622 Chr.21: 34183028-34183028 on Build GRCh38
rs907955696 Chr.21: 34182998-34182998 on Build GRCh38
rs1005996998 Chr.21: 34182942-34182942 on Build GRCh38
rs916868839 Chr.21: 34183103-34183103 on Build GRCh38
rs560704546 Chr.21: 34183203-34183203 on Build GRCh38
rs918167506 Chr.21: 34182750-34182750 on Build GRCh38
rs572958509 Chr.21: 34182934-34182934 on Build GRCh38
rs777093569 Chr.21: 34183235-34183235 on Build GRCh38
rs895583382 Chr.21: 34182819-34182819 on Build GRCh38
rs1056801165 Chr.21: 34183087-34183087 on Build GRCh38
rs996793732 Chr.21: 34183212-34183212 on Build GRCh38
rs892997907 Chr.21: 34182870-34182870 on Build GRCh38
rs532992877 Chr.21: 34182783-34182783 on Build GRCh38
rs1053776160 Chr.21: 34182863-34182863 on Build GRCh38
rs950338147 Chr.21: 34182938-34182938 on Build GRCh38
rs769618053 Chr.21: 34182895-34182895 on Build GRCh38
rs560477364 Chr.21: 34182902-34182902 on Build GRCh38
rs936772737 Chr.21: 34182839-34182839 on Build GRCh38
rs1009903795 Chr.21: 34182845-34182845 on Build GRCh38
rs566864504 Chr.21: 34183118-34183118 on Build GRCh38
rs902726287 Chr.21: 34182884-34182884 on Build GRCh38
rs901759110 Chr.21: 34182915-34182915 on Build GRCh38
rs113295571 Chr.21: 34183029-34183029 on Build GRCh38
rs1041488794 Chr.21: 34183114-34183115 on Build GRCh38
rs949454530 Chr.21: 34182753-34182753 on Build GRCh38
rs188968379 Chr.21: 34183040-34183040 on Build GRCh38
rs550809977 Chr.21: 34183046-34183046 on Build GRCh38
rs149403820 Chr.21: 34183084-34183084 on Build GRCh38
rs761204999 Chr.21: 34183202-34183202 on Build GRCh38
rs943248095 Chr.21: 34183158-34183158 on Build GRCh38
rs745695863 Chr.21: 34182791-34182791 on Build GRCh38
rs996205848 Chr.21: 34182930-34182930 on Build GRCh38
rs558023689 Chr.21: 34183106-34183106 on Build GRCh38
rs911301406 Chr.21: 34183126-34183126 on Build GRCh38
rs61519673 Chr.21: 34182984-34182984 on Build GRCh38
rs79942211 Chr.21: 34183068-34183068 on Build GRCh38
rs1042394971 Chr.21: 34182788-34182788 on Build GRCh38
rs936658178 Chr.21: 34183104-34183104 on Build GRCh38

View Full Product Details