Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 72230119-72230661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC392232
Entrez Gene ID
392232
Gene Name
transient receptor potential cation channel subfamily A member 1 pseudogene
Gene Aliases
-
UniGene
Hs.454043
Gene Chromosome Location
Chr.8: 72202752-72251634 on Build GRCh38
Transcripts
NR_033867.2

SNP Detail

SNP ID Chromosome Location
rs1042016631 Chr.8: 72230427-72230427 on Build GRCh38
rs376384102 Chr.8: 72230142-72230142 on Build GRCh38
rs1012265597 Chr.8: 72230511-72230511 on Build GRCh38
rs990191589 Chr.8: 72230160-72230160 on Build GRCh38
rs898647107 Chr.8: 72230591-72230591 on Build GRCh38
rs895166142 Chr.8: 72230500-72230500 on Build GRCh38
rs190113452 Chr.8: 72230285-72230285 on Build GRCh38
rs940240943 Chr.8: 72230243-72230243 on Build GRCh38
rs987002213 Chr.8: 72230631-72230631 on Build GRCh38
rs1026315240 Chr.8: 72230611-72230611 on Build GRCh38
rs1003578607 Chr.8: 72230185-72230185 on Build GRCh38
rs945927900 Chr.8: 72230418-72230418 on Build GRCh38
rs914387859 Chr.8: 72230408-72230408 on Build GRCh38
rs552775610 Chr.8: 72230299-72230299 on Build GRCh38
rs276657 Chr.8: 72230203-72230203 on Build GRCh38
rs372040306 Chr.8: 72230278-72230278 on Build GRCh38
rs963760440 Chr.8: 72230393-72230393 on Build GRCh38
rs143329289 Chr.8: 72230254-72230254 on Build GRCh38
rs2257653 Chr.8: 72230268-72230268 on Build GRCh38
rs948551867 Chr.8: 72230352-72230352 on Build GRCh38
rs994809427 Chr.8: 72230314-72230314 on Build GRCh38
rs181049248 Chr.8: 72230339-72230339 on Build GRCh38
rs910897821 Chr.8: 72230390-72230390 on Build GRCh38
rs2306505 Chr.8: 72230505-72230505 on Build GRCh38
rs12541758 Chr.8: 72230147-72230147 on Build GRCh38
rs999137698 Chr.8: 72230399-72230399 on Build GRCh38
rs1028483632 Chr.8: 72230171-72230171 on Build GRCh38
rs553906697 Chr.8: 72230293-72230293 on Build GRCh38
rs1011647315 Chr.8: 72230480-72230480 on Build GRCh38
rs1045944421 Chr.8: 72230363-72230363 on Build GRCh38
rs893131823 Chr.8: 72230455-72230455 on Build GRCh38
rs796321892 Chr.8: 72230494-72230494 on Build GRCh38
rs989916763 Chr.8: 72230398-72230398 on Build GRCh38
rs199842987 Chr.8: 72230407-72230407 on Build GRCh38
rs962869760 Chr.8: 72230294-72230294 on Build GRCh38
rs545062416 Chr.8: 72230519-72230519 on Build GRCh38
rs907125344 Chr.8: 72230379-72230379 on Build GRCh38
rs954198316 Chr.8: 72230179-72230179 on Build GRCh38
rs151334289 Chr.8: 72230284-72230284 on Build GRCh38
rs2306506 Chr.8: 72230470-72230470 on Build GRCh38
rs971765283 Chr.8: 72230266-72230266 on Build GRCh38
rs1038441911 Chr.8: 72230562-72230562 on Build GRCh38
rs1041657410 Chr.8: 72230176-72230177 on Build GRCh38
rs2306503 Chr.8: 72230526-72230526 on Build GRCh38
rs775292390 Chr.8: 72230578-72230578 on Build GRCh38
rs928606898 Chr.8: 72230454-72230454 on Build GRCh38
rs564002823 Chr.8: 72230204-72230204 on Build GRCh38
rs1028346364 Chr.8: 72230485-72230485 on Build GRCh38
rs986441957 Chr.8: 72230387-72230387 on Build GRCh38
rs531224955 Chr.8: 72230284-72230284 on Build GRCh38
rs987484161 Chr.8: 72230196-72230196 on Build GRCh38
rs2306504 Chr.8: 72230515-72230515 on Build GRCh38
rs994381742 Chr.8: 72230606-72230606 on Build GRCh38
rs576105412 Chr.8: 72230462-72230462 on Build GRCh38
rs58254809 Chr.8: 72230492-72230492 on Build GRCh38
rs954105826 Chr.8: 72230619-72230619 on Build GRCh38
rs398086694 Chr.8: 72230493-72230493 on Build GRCh38
rs892987698 Chr.8: 72230152-72230152 on Build GRCh38
rs573770893 Chr.8: 72230305-72230305 on Build GRCh38

View Full Product Details