Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 72251330-72251883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC392232
Entrez Gene ID
392232
Gene Name
transient receptor potential cation channel subfamily A member 1 pseudogene
Gene Aliases
-
UniGene
Hs.454043
Gene Chromosome Location
Chr.8: 72202752-72251634 on Build GRCh38
Transcripts
NR_033867.2

SNP Detail

SNP ID Chromosome Location
rs951137191 Chr.8: 72251363-72251363 on Build GRCh38
rs560243729 Chr.8: 72251684-72251684 on Build GRCh38
rs1028120917 Chr.8: 72251639-72251639 on Build GRCh38
rs912305864 Chr.8: 72251410-72251410 on Build GRCh38
rs1029633143 Chr.8: 72251726-72251726 on Build GRCh38
rs1020549097 Chr.8: 72251574-72251574 on Build GRCh38
rs371848525 Chr.8: 72251496-72251496 on Build GRCh38
rs185118418 Chr.8: 72251490-72251490 on Build GRCh38
rs768983920 Chr.8: 72251751-72251751 on Build GRCh38
rs1043757650 Chr.8: 72251484-72251484 on Build GRCh38
rs895454308 Chr.8: 72251661-72251661 on Build GRCh38
rs878876520 Chr.8: 72251598-72251598 on Build GRCh38
rs551485375 Chr.8: 72251380-72251380 on Build GRCh38
rs757322808 Chr.8: 72251619-72251619 on Build GRCh38
rs544432890 Chr.8: 72251641-72251641 on Build GRCh38
rs964045813 Chr.8: 72251602-72251602 on Build GRCh38
rs76471014 Chr.8: 72251613-72251613 on Build GRCh38
rs1027190766 Chr.8: 72251593-72251593 on Build GRCh38
rs967213745 Chr.8: 72251575-72251575 on Build GRCh38
rs531574185 Chr.8: 72251364-72251364 on Build GRCh38
rs941622791 Chr.8: 72251488-72251488 on Build GRCh38
rs1047657374 Chr.8: 72251462-72251462 on Build GRCh38
rs1034135362 Chr.8: 72251508-72251508 on Build GRCh38
rs1000023050 Chr.8: 72251683-72251683 on Build GRCh38
rs938256400 Chr.8: 72251800-72251800 on Build GRCh38
rs147567355 Chr.8: 72251632-72251632 on Build GRCh38
rs369722801 Chr.8: 72251657-72251657 on Build GRCh38
rs887397543 Chr.8: 72251480-72251480 on Build GRCh38
rs542550484 Chr.8: 72251763-72251763 on Build GRCh38
rs531385024 Chr.8: 72251797-72251797 on Build GRCh38
rs542217791 Chr.8: 72251742-72251744 on Build GRCh38
rs896557411 Chr.8: 72251509-72251509 on Build GRCh38
rs1905089 Chr.8: 72251682-72251682 on Build GRCh38
rs975474815 Chr.8: 72251376-72251376 on Build GRCh38
rs552718223 Chr.8: 72251586-72251586 on Build GRCh38
rs746028871 Chr.8: 72251861-72251861 on Build GRCh38
rs1014116073 Chr.8: 72251552-72251552 on Build GRCh38
rs571303867 Chr.8: 72251595-72251595 on Build GRCh38
rs931366614 Chr.8: 72251640-72251658 on Build GRCh38
rs577684062 Chr.8: 72251678-72251678 on Build GRCh38
rs565292441 Chr.8: 72251676-72251676 on Build GRCh38
rs529262776 Chr.8: 72251779-72251779 on Build GRCh38
rs949411399 Chr.8: 72251823-72251823 on Build GRCh38
rs955917823 Chr.8: 72251806-72251806 on Build GRCh38
rs531048992 Chr.8: 72251506-72251506 on Build GRCh38
rs377017864 Chr.8: 72251725-72251725 on Build GRCh38
rs752337936 Chr.8: 72251432-72251432 on Build GRCh38
rs546234508 Chr.8: 72251756-72251756 on Build GRCh38
rs369344371 Chr.8: 72251479-72251479 on Build GRCh38
rs1026580074 Chr.8: 72251358-72251358 on Build GRCh38
rs955380433 Chr.8: 72251731-72251731 on Build GRCh38
rs895567557 Chr.8: 72251517-72251517 on Build GRCh38
rs963909503 Chr.8: 72251690-72251690 on Build GRCh38
rs753845207 Chr.8: 72251607-72251607 on Build GRCh38
rs975253293 Chr.8: 72251457-72251457 on Build GRCh38
rs1014194985 Chr.8: 72251521-72251523 on Build GRCh38
rs995676879 Chr.8: 72251589-72251589 on Build GRCh38
rs1055454772 Chr.8: 72251658-72251658 on Build GRCh38
rs554534874 Chr.8: 72251826-72251826 on Build GRCh38
rs1039035507 Chr.8: 72251649-72251649 on Build GRCh38
rs1041427016 Chr.8: 72251679-72251679 on Build GRCh38
rs908724595 Chr.8: 72251638-72251638 on Build GRCh38
rs1038596630 Chr.8: 72251489-72251489 on Build GRCh38
rs926855436 Chr.8: 72251652-72251652 on Build GRCh38
rs975663731 Chr.8: 72251633-72251633 on Build GRCh38
rs898834837 Chr.8: 72251493-72251493 on Build GRCh38
rs140246012 Chr.8: 72251822-72251822 on Build GRCh38
rs781410415 Chr.8: 72251799-72251799 on Build GRCh38
rs988561162 Chr.8: 72251810-72251810 on Build GRCh38
rs529337693 Chr.8: 72251697-72251697 on Build GRCh38
rs111769798 Chr.8: 72251644-72251644 on Build GRCh38
rs556067156 Chr.8: 72251618-72251618 on Build GRCh38
rs974266361 Chr.8: 72251798-72251798 on Build GRCh38

View Full Product Details