Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15759173-15759721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F24P
Entrez Gene ID
388514
Gene Name
cytochrome P450 family 4 subfamily F member 24, pseudogene
Gene Aliases
-
UniGene
Hs.438274
Gene Chromosome Location
Chr.19: 15759334-15779988 on Build GRCh38
Transcripts
NR_033864.1

SNP Detail

SNP ID Chromosome Location
rs1020071405 Chr.19: 15759586-15759586 on Build GRCh38
rs572045526 Chr.19: 15759271-15759271 on Build GRCh38
rs897143302 Chr.19: 15759467-15759467 on Build GRCh38
rs796849364 Chr.19: 15759592-15759592 on Build GRCh38
rs1023987779 Chr.19: 15759276-15759276 on Build GRCh38
rs1037111709 Chr.19: 15759464-15759464 on Build GRCh38
rs908763761 Chr.19: 15759675-15759675 on Build GRCh38
rs969797379 Chr.19: 15759306-15759306 on Build GRCh38
rs1031996148 Chr.19: 15759512-15759512 on Build GRCh38
rs111458596 Chr.19: 15759670-15759670 on Build GRCh38
rs921811130 Chr.19: 15759659-15759659 on Build GRCh38
rs150308611 Chr.19: 15759442-15759442 on Build GRCh38
rs984776677 Chr.19: 15759347-15759347 on Build GRCh38
rs550262514 Chr.19: 15759333-15759333 on Build GRCh38
rs891732801 Chr.19: 15759520-15759520 on Build GRCh38
rs112758174 Chr.19: 15759569-15759569 on Build GRCh38
rs979823214 Chr.19: 15759649-15759649 on Build GRCh38
rs189413013 Chr.19: 15759686-15759686 on Build GRCh38
rs1002068249 Chr.19: 15759539-15759539 on Build GRCh38
rs912892283 Chr.19: 15759335-15759335 on Build GRCh38
rs931780625 Chr.19: 15759396-15759396 on Build GRCh38
rs535387839 Chr.19: 15759485-15759485 on Build GRCh38
rs1042979374 Chr.19: 15759360-15759360 on Build GRCh38
rs908827927 Chr.19: 15759353-15759353 on Build GRCh38
rs529991913 Chr.19: 15759309-15759309 on Build GRCh38
rs1000525042 Chr.19: 15759234-15759234 on Build GRCh38
rs79233950 Chr.19: 15759555-15759555 on Build GRCh38
rs980876035 Chr.19: 15759308-15759308 on Build GRCh38
rs1031629112 Chr.19: 15759237-15759237 on Build GRCh38
rs959817971 Chr.19: 15759212-15759212 on Build GRCh38
rs1051613735 Chr.19: 15759404-15759404 on Build GRCh38
rs901230327 Chr.19: 15759402-15759402 on Build GRCh38
rs372267279 Chr.19: 15759645-15759645 on Build GRCh38
rs1000068987 Chr.19: 15759499-15759499 on Build GRCh38
rs995485444 Chr.19: 15759477-15759477 on Build GRCh38
rs904406377 Chr.19: 15759220-15759220 on Build GRCh38
rs576085866 Chr.19: 15759685-15759685 on Build GRCh38
rs928337397 Chr.19: 15759194-15759194 on Build GRCh38
rs540920205 Chr.19: 15759283-15759283 on Build GRCh38
rs7245638 Chr.19: 15759240-15759240 on Build GRCh38
rs560995393 Chr.19: 15759289-15759289 on Build GRCh38
rs3764557 Chr.19: 15759436-15759436 on Build GRCh38
rs7248867 Chr.19: 15759394-15759394 on Build GRCh38
rs137957191 Chr.19: 15759653-15759653 on Build GRCh38
rs931542484 Chr.19: 15759328-15759328 on Build GRCh38
rs73929638 Chr.19: 15759357-15759357 on Build GRCh38
rs926800815 Chr.19: 15759325-15759325 on Build GRCh38
rs906122615 Chr.19: 15759524-15759524 on Build GRCh38
rs992533660 Chr.19: 15759263-15759263 on Build GRCh38
rs756654772 Chr.19: 15759684-15759684 on Build GRCh38
rs1035857319 Chr.19: 15759543-15759543 on Build GRCh38

View Full Product Details