Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15771724-15772308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F24P
Entrez Gene ID
388514
Gene Name
cytochrome P450 family 4 subfamily F member 24, pseudogene
Gene Aliases
-
UniGene
Hs.438274
Gene Chromosome Location
Chr.19: 15759334-15779988 on Build GRCh38
Transcripts
NR_033864.1

SNP Detail

SNP ID Chromosome Location
rs3752147 Chr.19: 15771953-15771953 on Build GRCh38
rs1029493933 Chr.19: 15771829-15771829 on Build GRCh38
rs749147606 Chr.19: 15772051-15772051 on Build GRCh38
rs761464870 Chr.19: 15772115-15772115 on Build GRCh38
rs184121684 Chr.19: 15771898-15771898 on Build GRCh38
rs779870421 Chr.19: 15772050-15772050 on Build GRCh38
rs751052284 Chr.19: 15772034-15772034 on Build GRCh38
rs370898404 Chr.19: 15772171-15772171 on Build GRCh38
rs76065100 Chr.19: 15771890-15771890 on Build GRCh38
rs773189575 Chr.19: 15772091-15772091 on Build GRCh38
rs754450777 Chr.19: 15772008-15772008 on Build GRCh38
rs2285889 Chr.19: 15771986-15771986 on Build GRCh38
rs140720527 Chr.19: 15772018-15772018 on Build GRCh38
rs749936201 Chr.19: 15772192-15772192 on Build GRCh38
rs761102211 Chr.19: 15772012-15772012 on Build GRCh38
rs1046604904 Chr.19: 15771908-15771908 on Build GRCh38
rs751142474 Chr.19: 15772112-15772112 on Build GRCh38
rs766714806 Chr.19: 15772017-15772017 on Build GRCh38
rs754753777 Chr.19: 15772137-15772137 on Build GRCh38
rs904325349 Chr.19: 15772266-15772266 on Build GRCh38
rs954089806 Chr.19: 15771824-15771824 on Build GRCh38
rs567378641 Chr.19: 15771770-15771770 on Build GRCh38
rs558572774 Chr.19: 15771952-15771952 on Build GRCh38
rs188199948 Chr.19: 15772061-15772061 on Build GRCh38
rs895731895 Chr.19: 15771993-15771993 on Build GRCh38
rs138235179 Chr.19: 15772177-15772177 on Build GRCh38
rs768135138 Chr.19: 15771998-15771998 on Build GRCh38
rs748386373 Chr.19: 15772085-15772085 on Build GRCh38
rs777876153 Chr.19: 15772158-15772158 on Build GRCh38
rs986075866 Chr.19: 15771828-15771828 on Build GRCh38
rs945937394 Chr.19: 15772247-15772247 on Build GRCh38
rs753774905 Chr.19: 15772206-15772206 on Build GRCh38
rs386807245 Chr.19: 15772170-15772171 on Build GRCh38
rs761202150 Chr.19: 15772094-15772094 on Build GRCh38
rs200914227 Chr.19: 15772170-15772171 on Build GRCh38
rs775852037 Chr.19: 15772178-15772178 on Build GRCh38
rs912591604 Chr.19: 15772110-15772110 on Build GRCh38
rs897195171 Chr.19: 15772239-15772239 on Build GRCh38
rs3752143 Chr.19: 15772170-15772170 on Build GRCh38
rs1037198998 Chr.19: 15771852-15771852 on Build GRCh38
rs912994098 Chr.19: 15771899-15771899 on Build GRCh38
rs759952379 Chr.19: 15772100-15772100 on Build GRCh38
rs780568082 Chr.19: 15772038-15772038 on Build GRCh38
rs3752146 Chr.19: 15771967-15771967 on Build GRCh38
rs944302994 Chr.19: 15771848-15771848 on Build GRCh38
rs925596385 Chr.19: 15771875-15771875 on Build GRCh38
rs766833831 Chr.19: 15772095-15772095 on Build GRCh38
rs1001217936 Chr.19: 15771941-15771941 on Build GRCh38
rs1056783156 Chr.19: 15772232-15772232 on Build GRCh38
rs763785005 Chr.19: 15772033-15772033 on Build GRCh38
rs925244835 Chr.19: 15771818-15771818 on Build GRCh38
rs770732669 Chr.19: 15771928-15771928 on Build GRCh38
rs115251433 Chr.19: 15772118-15772118 on Build GRCh38
rs1041633629 Chr.19: 15772265-15772265 on Build GRCh38
rs374223714 Chr.19: 15771756-15771756 on Build GRCh38
rs953960674 Chr.19: 15771853-15771853 on Build GRCh38
rs774554947 Chr.19: 15771995-15771995 on Build GRCh38
rs961164298 Chr.19: 15771787-15771787 on Build GRCh38
rs773703577 Chr.19: 15772009-15772009 on Build GRCh38
rs938238261 Chr.19: 15771923-15771923 on Build GRCh38
rs772223143 Chr.19: 15772086-15772086 on Build GRCh38
rs760170627 Chr.19: 15772193-15772193 on Build GRCh38
rs906741048 Chr.19: 15771922-15771922 on Build GRCh38
rs778866504 Chr.19: 15772140-15772140 on Build GRCh38
rs1048164044 Chr.19: 15771916-15771916 on Build GRCh38
rs762042955 Chr.19: 15771997-15771997 on Build GRCh38
rs772212209 Chr.19: 15772156-15772156 on Build GRCh38
rs747106177 Chr.19: 15772167-15772167 on Build GRCh38
rs949780324 Chr.19: 15772129-15772129 on Build GRCh38
rs1051147413 Chr.19: 15772261-15772261 on Build GRCh38
rs779917868 Chr.19: 15772121-15772121 on Build GRCh38
rs985428149 Chr.19: 15771865-15771865 on Build GRCh38
rs566553456 Chr.19: 15771868-15771868 on Build GRCh38
rs754554469 Chr.19: 15772248-15772248 on Build GRCh38
rs767126018 Chr.19: 15772180-15772180 on Build GRCh38
rs753681287 Chr.19: 15772127-15772127 on Build GRCh38
rs3752142 Chr.19: 15772171-15772171 on Build GRCh38
rs912819477 Chr.19: 15771843-15771843 on Build GRCh38
rs745758062 Chr.19: 15772041-15772041 on Build GRCh38
rs555905857 Chr.19: 15771832-15771832 on Build GRCh38
rs561408250 Chr.19: 15771983-15771983 on Build GRCh38
rs765922623 Chr.19: 15772204-15772204 on Build GRCh38
rs576989340 Chr.19: 15771861-15771861 on Build GRCh38
rs1032948155 Chr.19: 15772073-15772073 on Build GRCh38
rs1037030333 Chr.19: 15772238-15772238 on Build GRCh38
rs1020061144 Chr.19: 15772010-15772010 on Build GRCh38
rs758013600 Chr.19: 15772022-15772022 on Build GRCh38
rs895515706 Chr.19: 15772243-15772243 on Build GRCh38
rs762397191 Chr.19: 15772026-15772026 on Build GRCh38
rs553035710 Chr.19: 15771940-15771940 on Build GRCh38
rs3752145 Chr.19: 15771978-15771978 on Build GRCh38
rs761425870 Chr.19: 15772179-15772179 on Build GRCh38
rs756040136 Chr.19: 15772042-15772042 on Build GRCh38
rs3752144 Chr.19: 15772074-15772074 on Build GRCh38
rs573173389 Chr.19: 15771946-15771946 on Build GRCh38
rs756823676 Chr.19: 15772035-15772035 on Build GRCh38
rs577812728 Chr.19: 15772120-15772120 on Build GRCh38
rs976560764 Chr.19: 15771823-15771823 on Build GRCh38
rs1029441777 Chr.19: 15772030-15772030 on Build GRCh38
rs115673553 Chr.19: 15771781-15771781 on Build GRCh38
rs547049680 Chr.19: 15772275-15772275 on Build GRCh38
rs763727884 Chr.19: 15772111-15772111 on Build GRCh38
rs748333139 Chr.19: 15772144-15772144 on Build GRCh38

View Full Product Details