Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39707089-39707219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBB2
Entrez Gene ID
2064
Gene Name
erb-b2 receptor tyrosine kinase 2
Gene Aliases
CD340, HER-2, HER-2/neu, HER2, MLN 19, NEU, NGL, TKR1
UniGene
Hs.446352
Gene Chromosome Location
Chr.17: 39688084-39728662 on Build GRCh38
Transcripts
NM_001005862.2 NM_001289936.1 NM_001289937.1 NM_001289938.1 NM_004448.3 NR_110535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6655692 c.180+8C>T p.? Chr.17: 39707149-39707149 on Build GRCh38
6655691 c.225+8C>T p.? Chr.17: 39707149-39707149 on Build GRCh38
5441271 c.132C>A p.N44K Chr.17: 39707093-39707093 on Build GRCh38
7343958 c.178C>G p.L60V Chr.17: 39707094-39707094 on Build GRCh38
5441270 c.177C>A p.N59K Chr.17: 39707093-39707093 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778575008 Chr.17: 39707115-39707115 on Build GRCh38
rs754691039 Chr.17: 39707126-39707126 on Build GRCh38
rs752339377 Chr.17: 39707116-39707116 on Build GRCh38
rs766351280 Chr.17: 39707197-39707197 on Build GRCh38
rs780661804 Chr.17: 39707141-39707141 on Build GRCh38
rs768897680 Chr.17: 39707192-39707192 on Build GRCh38
rs754278477 Chr.17: 39707145-39707165 on Build GRCh38
rs761750231 Chr.17: 39707194-39707194 on Build GRCh38
rs777347427 Chr.17: 39707122-39707122 on Build GRCh38
rs757899978 Chr.17: 39707119-39707119 on Build GRCh38
rs763250146 Chr.17: 39707191-39707191 on Build GRCh38
rs746278619 Chr.17: 39707165-39707165 on Build GRCh38
rs770279585 Chr.17: 39707171-39707171 on Build GRCh38
rs775914701 Chr.17: 39707174-39707174 on Build GRCh38
rs774519406 Chr.17: 39707193-39707193 on Build GRCh38
rs371270269 Chr.17: 39707151-39707151 on Build GRCh38
rs529042839 Chr.17: 39707155-39707155 on Build GRCh38
rs928795169 Chr.17: 39707114-39707114 on Build GRCh38

View Full Product Details