Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36228961-36229218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD3
Entrez Gene ID
143458
Gene Name
low density lipoprotein receptor class A domain containing 3
Gene Aliases
LRAD3
UniGene
Hs.745134
Gene Chromosome Location
Chr.11: 35943981-36232136 on Build GRCh38
Transcripts
NM_001304263.1 NM_001304264.1 NM_174902.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1978140 c.807G>A p.A269A Chr.11: 36229166-36229166 on Build GRCh38
7106924 c.828G>A p.P276P Chr.11: 36229187-36229187 on Build GRCh38
6068483 c.807G>T p.A269A Chr.11: 36229166-36229166 on Build GRCh38
3447031 c.826C>T p.P276S Chr.11: 36229185-36229185 on Build GRCh38
4032596 c.827C>T p.P276L Chr.11: 36229186-36229186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775330520 Chr.11: 36229046-36229046 on Build GRCh38
rs78027431 Chr.11: 36229141-36229141 on Build GRCh38
rs913958460 Chr.11: 36229041-36229041 on Build GRCh38
rs563212236 Chr.11: 36228988-36228988 on Build GRCh38
rs564722286 Chr.11: 36229062-36229062 on Build GRCh38
rs183702391 Chr.11: 36229050-36229050 on Build GRCh38
rs891302653 Chr.11: 36229080-36229080 on Build GRCh38
rs934453334 Chr.11: 36228983-36228983 on Build GRCh38
rs369173678 Chr.11: 36229117-36229117 on Build GRCh38
rs763477666 Chr.11: 36229131-36229131 on Build GRCh38
rs149451273 Chr.11: 36229165-36229165 on Build GRCh38
rs755767754 Chr.11: 36229142-36229142 on Build GRCh38
rs778689642 Chr.11: 36229160-36229160 on Build GRCh38
rs749100906 Chr.11: 36229151-36229151 on Build GRCh38
rs1047411723 Chr.11: 36229015-36229015 on Build GRCh38
rs543216259 Chr.11: 36228987-36228987 on Build GRCh38
rs756975237 Chr.11: 36229153-36229153 on Build GRCh38
rs748234560 Chr.11: 36229171-36229171 on Build GRCh38
rs745754811 Chr.11: 36229164-36229164 on Build GRCh38
rs900005584 Chr.11: 36229101-36229101 on Build GRCh38
rs777348525 Chr.11: 36229143-36229143 on Build GRCh38
rs770051772 Chr.11: 36229186-36229186 on Build GRCh38
rs899829684 Chr.11: 36229048-36229048 on Build GRCh38
rs766855250 Chr.11: 36229134-36229134 on Build GRCh38
rs969660023 Chr.11: 36229180-36229180 on Build GRCh38
rs766419149 Chr.11: 36229189-36229189 on Build GRCh38
rs778392274 Chr.11: 36229073-36229073 on Build GRCh38
rs932923410 Chr.11: 36229051-36229051 on Build GRCh38
rs750790800 Chr.11: 36229121-36229121 on Build GRCh38
rs760761455 Chr.11: 36229166-36229166 on Build GRCh38
rs527586595 Chr.11: 36229047-36229047 on Build GRCh38
rs752055780 Chr.11: 36229138-36229138 on Build GRCh38
rs114075493 Chr.11: 36229187-36229187 on Build GRCh38
rs531867272 Chr.11: 36229034-36229034 on Build GRCh38
rs1051313863 Chr.11: 36229078-36229078 on Build GRCh38

View Full Product Details