Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153682377-153682648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPR1
Entrez Gene ID
4881
Gene Name
natriuretic peptide receptor 1
Gene Aliases
ANPRA, ANPa, GUC2A, GUCY2A, NPRA
UniGene
Hs.490330
Gene Chromosome Location
Chr.1: 153678617-153693992 on Build GRCh38
Transcripts
NM_000906.3 XM_005245218.1 XM_017001374.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3474474 c.1226C>T p.S409F Chr.1: 153682552-153682552 on Build GRCh38
5052587 c.1206C>T p.G402G Chr.1: 153682532-153682532 on Build GRCh38
3474475 c.1243C>T p.P415S Chr.1: 153682569-153682569 on Build GRCh38
6058064 c.1252G>T p.G418C Chr.1: 153682578-153682578 on Build GRCh38
5843736 c.1181G>A p.G394E Chr.1: 153682507-153682507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575234899 Chr.1: 153682417-153682417 on Build GRCh38
rs769691983 Chr.1: 153682537-153682537 on Build GRCh38
rs532224804 Chr.1: 153682472-153682472 on Build GRCh38
rs202020768 Chr.1: 153682465-153682465 on Build GRCh38
rs750202886 Chr.1: 153682459-153682459 on Build GRCh38
rs754614053 Chr.1: 153682475-153682475 on Build GRCh38
rs751196634 Chr.1: 153682483-153682483 on Build GRCh38
rs374193089 Chr.1: 153682536-153682536 on Build GRCh38
rs370485762 Chr.1: 153682532-153682532 on Build GRCh38
rs931188553 Chr.1: 153682502-153682502 on Build GRCh38
rs773559889 Chr.1: 153682604-153682604 on Build GRCh38
rs779442067 Chr.1: 153682505-153682505 on Build GRCh38
rs765152343 Chr.1: 153682450-153682450 on Build GRCh38
rs972686139 Chr.1: 153682454-153682454 on Build GRCh38
rs528255169 Chr.1: 153682462-153682462 on Build GRCh38
rs1041093843 Chr.1: 153682538-153682538 on Build GRCh38
rs780697403 Chr.1: 153682477-153682477 on Build GRCh38
rs749649180 Chr.1: 153682486-153682486 on Build GRCh38
rs889760412 Chr.1: 153682533-153682533 on Build GRCh38
rs751327438 Chr.1: 153682565-153682565 on Build GRCh38
rs376178523 Chr.1: 153682595-153682595 on Build GRCh38
rs550468871 Chr.1: 153682484-153682484 on Build GRCh38
rs766728119 Chr.1: 153682582-153682582 on Build GRCh38
rs752156295 Chr.1: 153682479-153682479 on Build GRCh38
rs61806718 Chr.1: 153682438-153682438 on Build GRCh38
rs771463059 Chr.1: 153682488-153682488 on Build GRCh38
rs201243793 Chr.1: 153682468-153682468 on Build GRCh38
rs766117772 Chr.1: 153682559-153682559 on Build GRCh38
rs1012387217 Chr.1: 153682435-153682435 on Build GRCh38
rs201584708 Chr.1: 153682571-153682571 on Build GRCh38
rs74408467 Chr.1: 153682428-153682428 on Build GRCh38
rs756641839 Chr.1: 153682483-153682483 on Build GRCh38
rs761741776 Chr.1: 153682449-153682449 on Build GRCh38
rs762912463 Chr.1: 153682551-153682551 on Build GRCh38
rs966142505 Chr.1: 153682581-153682581 on Build GRCh38
rs563098350 Chr.1: 153682412-153682412 on Build GRCh38
rs928481356 Chr.1: 153682433-153682433 on Build GRCh38
rs113981977 Chr.1: 153682510-153682510 on Build GRCh38
rs561065588 Chr.1: 153682434-153682434 on Build GRCh38
rs773170125 Chr.1: 153682543-153682543 on Build GRCh38
rs370844205 Chr.1: 153682600-153682600 on Build GRCh38

View Full Product Details